Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27312133-27312630 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MPV17
Entrez Gene ID
4358
Gene Name
MPV17, mitochondrial inner membrane protein
Gene Aliases
MTDPS6, SYM1
UniGene
Hs.75659
Gene Chromosome Location
Chr.2: 27309492-27323102 on Build GRCh38
Transcripts
NM_002437.4 XM_005264326.3 XM_006712021.3 XM_017004150.1 XM_017004151.1 XM_017004152.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6326999 c.328G>A p.G110S Chr.2: 27312586-27312586 on Build GRCh38
3713759 c.336C>T p.A112A Chr.2: 27312578-27312578 on Build GRCh38
6990122 c.378T>A p.D126E Chr.2: 27312244-27312244 on Build GRCh38
6990120 c.381T>A p.Y127* Chr.2: 27312241-27312241 on Build GRCh38
3580601 c.360C>T p.L120L Chr.2: 27312554-27312554 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761079345 Chr.2: 27312387-27312387 on Build GRCh38
rs776344322 Chr.2: 27312392-27312392 on Build GRCh38
rs369706595 Chr.2: 27312568-27312568 on Build GRCh38
rs886044454 Chr.2: 27312585-27312586 on Build GRCh38
rs764842864 Chr.2: 27312397-27312397 on Build GRCh38
rs988733706 Chr.2: 27312542-27312542 on Build GRCh38
rs374763606 Chr.2: 27312456-27312456 on Build GRCh38
rs201532533 Chr.2: 27312453-27312453 on Build GRCh38
rs754100850 Chr.2: 27312424-27312424 on Build GRCh38
rs751347687 Chr.2: 27312410-27312410 on Build GRCh38
rs772621135 Chr.2: 27312435-27312435 on Build GRCh38
rs397775769 Chr.2: 27312445-27312446 on Build GRCh38
rs368900406 Chr.2: 27312255-27312255 on Build GRCh38
rs869046158 Chr.2: 27312447-27312448 on Build GRCh38
rs149309794 Chr.2: 27312224-27312224 on Build GRCh38
rs559110607 Chr.2: 27312277-27312277 on Build GRCh38
rs761288514 Chr.2: 27312395-27312395 on Build GRCh38
rs374692755 Chr.2: 27312215-27312215 on Build GRCh38
rs559746258 Chr.2: 27312322-27312322 on Build GRCh38
rs137975836 Chr.2: 27312512-27312512 on Build GRCh38
rs758469630 Chr.2: 27312556-27312556 on Build GRCh38
rs767352327 Chr.2: 27312504-27312504 on Build GRCh38
rs749911111 Chr.2: 27312399-27312399 on Build GRCh38
rs776812146 Chr.2: 27312442-27312443 on Build GRCh38
rs768234149 Chr.2: 27312391-27312391 on Build GRCh38
rs758343007 Chr.2: 27312271-27312271 on Build GRCh38
rs770017331 Chr.2: 27312191-27312191 on Build GRCh38
rs113055360 Chr.2: 27312248-27312248 on Build GRCh38
rs182015952 Chr.2: 27312442-27312442 on Build GRCh38
rs1054997754 Chr.2: 27312492-27312492 on Build GRCh38
rs766160589 Chr.2: 27312584-27312585 on Build GRCh38
rs746381745 Chr.2: 27312294-27312294 on Build GRCh38
rs542512563 Chr.2: 27312373-27312373 on Build GRCh38
rs187496263 Chr.2: 27312282-27312282 on Build GRCh38
rs142286079 Chr.2: 27312443-27312443 on Build GRCh38
rs764173266 Chr.2: 27312428-27312428 on Build GRCh38
rs778888514 Chr.2: 27312171-27312171 on Build GRCh38
rs772896372 Chr.2: 27312476-27312476 on Build GRCh38
rs553828539 Chr.2: 27312161-27312161 on Build GRCh38
rs1057379273 Chr.2: 27312318-27312318 on Build GRCh38
rs372125579 Chr.2: 27312281-27312281 on Build GRCh38
rs750541759 Chr.2: 27312540-27312540 on Build GRCh38
rs772204532 Chr.2: 27312267-27312267 on Build GRCh38
rs751072070 Chr.2: 27312305-27312305 on Build GRCh38
rs531025023 Chr.2: 27312426-27312426 on Build GRCh38
rs267607257 Chr.2: 27312589-27312589 on Build GRCh38
rs780507661 Chr.2: 27312441-27312441 on Build GRCh38
rs747567003 Chr.2: 27312575-27312575 on Build GRCh38
rs902269508 Chr.2: 27312559-27312559 on Build GRCh38
rs938889725 Chr.2: 27312304-27312304 on Build GRCh38
rs987343087 Chr.2: 27312170-27312170 on Build GRCh38
rs781636695 Chr.2: 27312590-27312590 on Build GRCh38
rs745902338 Chr.2: 27312434-27312434 on Build GRCh38
rs760987542 Chr.2: 27312370-27312371 on Build GRCh38
rs121909724 Chr.2: 27312510-27312510 on Build GRCh38
rs778998249 Chr.2: 27312429-27312429 on Build GRCh38
rs376678921 Chr.2: 27312600-27312600 on Build GRCh38
rs763693721 Chr.2: 27312266-27312266 on Build GRCh38
rs184194542 Chr.2: 27312278-27312278 on Build GRCh38
rs1030845017 Chr.2: 27312488-27312488 on Build GRCh38
rs199690638 Chr.2: 27312218-27312218 on Build GRCh38
rs112170670 Chr.2: 27312496-27312496 on Build GRCh38
rs927475461 Chr.2: 27312284-27312284 on Build GRCh38
rs757495960 Chr.2: 27312532-27312532 on Build GRCh38
rs777912572 Chr.2: 27312420-27312420 on Build GRCh38
rs980207020 Chr.2: 27312586-27312586 on Build GRCh38
rs753339511 Chr.2: 27312267-27312267 on Build GRCh38
rs760281019 Chr.2: 27312232-27312232 on Build GRCh38
rs779935325 Chr.2: 27312189-27312189 on Build GRCh38
rs746360637 Chr.2: 27312181-27312183 on Build GRCh38
rs368503416 Chr.2: 27312167-27312167 on Build GRCh38
rs898872925 Chr.2: 27312394-27312394 on Build GRCh38
rs774833271 Chr.2: 27312217-27312217 on Build GRCh38
rs749361266 Chr.2: 27312213-27312213 on Build GRCh38
rs575516637 Chr.2: 27312186-27312186 on Build GRCh38
rs766486395 Chr.2: 27312495-27312495 on Build GRCh38
rs949261135 Chr.2: 27312507-27312507 on Build GRCh38
rs745699358 Chr.2: 27312185-27312185 on Build GRCh38
rs55689147 Chr.2: 27312538-27312538 on Build GRCh38
rs754794889 Chr.2: 27312418-27312418 on Build GRCh38
rs368819139 Chr.2: 27312457-27312457 on Build GRCh38
rs750374833 Chr.2: 27312270-27312270 on Build GRCh38
rs563702392 Chr.2: 27312401-27312401 on Build GRCh38
rs775364154 Chr.2: 27312367-27312368 on Build GRCh38
rs191656934 Chr.2: 27312337-27312337 on Build GRCh38
rs780025038 Chr.2: 27312272-27312272 on Build GRCh38
rs748384037 Chr.2: 27312190-27312190 on Build GRCh38
rs773187260 Chr.2: 27312207-27312207 on Build GRCh38
rs1037041622 Chr.2: 27312449-27312449 on Build GRCh38
rs371775551 Chr.2: 27312481-27312481 on Build GRCh38
rs267607258 Chr.2: 27312576-27312576 on Build GRCh38
rs863224074 Chr.2: 27312499-27312499 on Build GRCh38
rs748500676 Chr.2: 27312591-27312591 on Build GRCh38
rs772990781 Chr.2: 27312603-27312603 on Build GRCh38
rs775768004 Chr.2: 27312226-27312226 on Build GRCh38
rs772531611 Chr.2: 27312360-27312360 on Build GRCh38
rs397984156 Chr.2: 27312446-27312447 on Build GRCh38
rs373571379 Chr.2: 27312579-27312579 on Build GRCh38
rs763757825 Chr.2: 27312249-27312249 on Build GRCh38
rs990783785 Chr.2: 27312307-27312307 on Build GRCh38
rs138004746 Chr.2: 27312157-27312157 on Build GRCh38
rs780485901 Chr.2: 27312564-27312564 on Build GRCh38
rs778021945 Chr.2: 27312279-27312279 on Build GRCh38

View Full Product Details