Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27368280-27368791 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF2B4
Entrez Gene ID
8890
Gene Name
eukaryotic translation initiation factor 2B subunit delta
Gene Aliases
EIF-2B, EIF2B, EIF2Bdelta
UniGene
Hs.169474
Gene Chromosome Location
Chr.2: 27364352-27370457 on Build GRCh38
Transcripts
NM_001034116.1 NM_001318965.1 NM_001318966.1 NM_001318967.1 NM_001318968.1 NM_001318969.1 NM_015636.3 NM_172195.3 XM_006712132.1 XM_011533147.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1641896 c.526A>G p.K176E Chr.2: 27368436-27368436 on Build GRCh38
3839280 c.544C>T p.H182Y Chr.2: 27368418-27368418 on Build GRCh38
6333237 c.558C>A p.Y186* Chr.2: 27368404-27368404 on Build GRCh38
6273524 c.584C>A p.S195Y Chr.2: 27368438-27368438 on Build GRCh38
4983035 c.505C>G p.Q169E Chr.2: 27368707-27368707 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs944661678 Chr.2: 27368511-27368511 on Build GRCh38
rs566287769 Chr.2: 27368495-27368495 on Build GRCh38
rs913265804 Chr.2: 27368538-27368538 on Build GRCh38
rs752260211 Chr.2: 27368745-27368745 on Build GRCh38
rs547695043 Chr.2: 27368489-27368489 on Build GRCh38
rs201042063 Chr.2: 27368640-27368640 on Build GRCh38
rs754559917 Chr.2: 27368599-27368599 on Build GRCh38
rs758627794 Chr.2: 27368582-27368582 on Build GRCh38
rs752818834 Chr.2: 27368379-27368379 on Build GRCh38
rs947539380 Chr.2: 27368570-27368570 on Build GRCh38
rs866357253 Chr.2: 27368670-27368670 on Build GRCh38
rs756090304 Chr.2: 27368392-27368392 on Build GRCh38
rs751135430 Chr.2: 27368588-27368588 on Build GRCh38
rs559965006 Chr.2: 27368763-27368763 on Build GRCh38
rs758641876 Chr.2: 27368535-27368535 on Build GRCh38
rs374326886 Chr.2: 27368516-27368516 on Build GRCh38
rs911814858 Chr.2: 27368498-27368498 on Build GRCh38
rs113309679 Chr.2: 27368320-27368320 on Build GRCh38
rs370256543 Chr.2: 27368513-27368513 on Build GRCh38
rs761726266 Chr.2: 27368351-27368351 on Build GRCh38
rs761810644 Chr.2: 27368711-27368711 on Build GRCh38
rs770591078 Chr.2: 27368661-27368661 on Build GRCh38
rs770699036 Chr.2: 27368746-27368746 on Build GRCh38
rs914750494 Chr.2: 27368562-27368562 on Build GRCh38
rs775819795 Chr.2: 27368662-27368662 on Build GRCh38
rs779235094 Chr.2: 27368744-27368746 on Build GRCh38
rs371511880 Chr.2: 27368748-27368748 on Build GRCh38
rs886055902 Chr.2: 27368447-27368447 on Build GRCh38
rs529735415 Chr.2: 27368639-27368639 on Build GRCh38
rs758061891 Chr.2: 27368443-27368443 on Build GRCh38
rs768638665 Chr.2: 27368346-27368346 on Build GRCh38
rs754390217 Chr.2: 27368509-27368509 on Build GRCh38
rs147672203 Chr.2: 27368669-27368669 on Build GRCh38
rs373696834 Chr.2: 27368352-27368352 on Build GRCh38
rs149121402 Chr.2: 27368723-27368723 on Build GRCh38
rs772991139 Chr.2: 27368390-27368390 on Build GRCh38
rs946111331 Chr.2: 27368552-27368552 on Build GRCh38
rs779782574 Chr.2: 27368583-27368583 on Build GRCh38
rs748069510 Chr.2: 27368469-27368469 on Build GRCh38
rs748155307 Chr.2: 27368634-27368634 on Build GRCh38
rs928893380 Chr.2: 27368500-27368500 on Build GRCh38
rs535744717 Chr.2: 27368332-27368332 on Build GRCh38
rs750788095 Chr.2: 27368579-27368579 on Build GRCh38
rs768779136 Chr.2: 27368685-27368685 on Build GRCh38
rs776685186 Chr.2: 27368695-27368695 on Build GRCh38
rs369686466 Chr.2: 27368324-27368324 on Build GRCh38
rs201503266 Chr.2: 27368666-27368666 on Build GRCh38
rs769763044 Chr.2: 27368474-27368474 on Build GRCh38
rs746547024 Chr.2: 27368457-27368457 on Build GRCh38
rs569664227 Chr.2: 27368697-27368697 on Build GRCh38
rs879271655 Chr.2: 27368415-27368415 on Build GRCh38
rs374915492 Chr.2: 27368322-27368322 on Build GRCh38
rs1008673121 Chr.2: 27368647-27368647 on Build GRCh38
rs776656241 Chr.2: 27368350-27368350 on Build GRCh38
rs375458330 Chr.2: 27368396-27368396 on Build GRCh38
rs777786550 Chr.2: 27368644-27368644 on Build GRCh38
rs565245777 Chr.2: 27368427-27368427 on Build GRCh38
rs757657538 Chr.2: 27368416-27368416 on Build GRCh38
rs750850634 Chr.2: 27368719-27368719 on Build GRCh38
rs542713478 Chr.2: 27368312-27368314 on Build GRCh38
rs886038382 Chr.2: 27368617-27368617 on Build GRCh38
rs776743727 Chr.2: 27368467-27368467 on Build GRCh38
rs749149001 Chr.2: 27368752-27368752 on Build GRCh38
rs746981613 Chr.2: 27368333-27368333 on Build GRCh38
rs779565150 Chr.2: 27368588-27368589 on Build GRCh38
rs570777761 Chr.2: 27368740-27368740 on Build GRCh38
rs534790807 Chr.2: 27368750-27368750 on Build GRCh38
rs370413772 Chr.2: 27368650-27368650 on Build GRCh38
rs778566686 Chr.2: 27368323-27368323 on Build GRCh38
rs961728137 Chr.2: 27368747-27368747 on Build GRCh38
rs779502336 Chr.2: 27368444-27368444 on Build GRCh38
rs756334195 Chr.2: 27368317-27368317 on Build GRCh38
rs1014356484 Chr.2: 27368341-27368341 on Build GRCh38
rs765743112 Chr.2: 27368714-27368714 on Build GRCh38
rs990980109 Chr.2: 27368361-27368361 on Build GRCh38
rs370123021 Chr.2: 27368479-27368479 on Build GRCh38
rs764126847 Chr.2: 27368506-27368506 on Build GRCh38
rs377475649 Chr.2: 27368493-27368493 on Build GRCh38
rs377518092 Chr.2: 27368620-27368620 on Build GRCh38
rs768826695 Chr.2: 27368342-27368342 on Build GRCh38
rs754665816 Chr.2: 27368725-27368725 on Build GRCh38
rs745575196 Chr.2: 27368430-27368430 on Build GRCh38
rs368243788 Chr.2: 27368724-27368724 on Build GRCh38
rs770808088 Chr.2: 27368765-27368765 on Build GRCh38
rs376400542 Chr.2: 27368330-27368330 on Build GRCh38
rs373303281 Chr.2: 27368488-27368488 on Build GRCh38
rs191734334 Chr.2: 27368355-27368355 on Build GRCh38
rs904731151 Chr.2: 27368319-27368319 on Build GRCh38
rs920428422 Chr.2: 27368439-27368439 on Build GRCh38
rs767553031 Chr.2: 27368375-27368375 on Build GRCh38
rs1053334796 Chr.2: 27368540-27368540 on Build GRCh38

View Full Product Details