Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 30561758-30562264 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LCLAT1
Entrez Gene ID
253558
Gene Name
lysocardiolipin acyltransferase 1
Gene Aliases
1AGPAT8, AGPAT8, ALCAT1, HSRG1849, LYCAT, UNQ1849
UniGene
Hs.468048
Gene Chromosome Location
Chr.2: 30447226-30644225 on Build GRCh38
Transcripts
NM_001002257.2 NM_001304445.1 NM_001304446.1 NM_182551.4 XM_005264244.1 XM_005264245.3 XM_011532741.2 XM_011532742.2 XM_011532743.2 XM_011532744.2 XM_017003746.1 XM_017003747.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5634749 c.579C>T p.H193H Chr.2: 30562246-30562246 on Build GRCh38
5681907 c.529G>A p.D177N Chr.2: 30562196-30562196 on Build GRCh38
4312757 c.495T>A p.A165A Chr.2: 30562162-30562162 on Build GRCh38
6835529 c.490C>T p.Q164* Chr.2: 30562157-30562157 on Build GRCh38
185059 c.547G>A p.E183K Chr.2: 30562214-30562214 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs897143612 Chr.2: 30561974-30561974 on Build GRCh38
rs531694011 Chr.2: 30561841-30561841 on Build GRCh38
rs754740898 Chr.2: 30562234-30562234 on Build GRCh38
rs777896228 Chr.2: 30562019-30562019 on Build GRCh38
rs1031698644 Chr.2: 30562220-30562220 on Build GRCh38
rs1036714139 Chr.2: 30561915-30561915 on Build GRCh38
rs571519822 Chr.2: 30561903-30561903 on Build GRCh38
rs547645100 Chr.2: 30561999-30561999 on Build GRCh38
rs527681258 Chr.2: 30561926-30561926 on Build GRCh38
rs368421909 Chr.2: 30562097-30562097 on Build GRCh38
rs764465122 Chr.2: 30562114-30562114 on Build GRCh38
rs915528213 Chr.2: 30561811-30561811 on Build GRCh38
rs183362062 Chr.2: 30561825-30561825 on Build GRCh38
rs551867085 Chr.2: 30561902-30561902 on Build GRCh38
rs1014816905 Chr.2: 30561960-30561960 on Build GRCh38
rs780206248 Chr.2: 30562214-30562214 on Build GRCh38
rs771156495 Chr.2: 30562154-30562154 on Build GRCh38
rs989760975 Chr.2: 30561810-30561810 on Build GRCh38
rs902693182 Chr.2: 30561832-30561832 on Build GRCh38
rs781020487 Chr.2: 30562193-30562193 on Build GRCh38
rs762367929 Chr.2: 30562127-30562127 on Build GRCh38
rs750575432 Chr.2: 30562172-30562172 on Build GRCh38
rs953125048 Chr.2: 30562051-30562051 on Build GRCh38
rs978767363 Chr.2: 30562240-30562240 on Build GRCh38
rs758595183 Chr.2: 30562161-30562161 on Build GRCh38
rs1022389710 Chr.2: 30562182-30562182 on Build GRCh38
rs754420718 Chr.2: 30562115-30562115 on Build GRCh38
rs945702752 Chr.2: 30562140-30562140 on Build GRCh38
rs776794409 Chr.2: 30562162-30562162 on Build GRCh38
rs370427576 Chr.2: 30562112-30562112 on Build GRCh38
rs897755491 Chr.2: 30561893-30561893 on Build GRCh38
rs758487673 Chr.2: 30562213-30562213 on Build GRCh38
rs143382745 Chr.2: 30562055-30562055 on Build GRCh38
rs1005218830 Chr.2: 30561922-30561922 on Build GRCh38
rs887741318 Chr.2: 30561921-30561921 on Build GRCh38
rs749097869 Chr.2: 30561873-30561873 on Build GRCh38
rs946960756 Chr.2: 30561819-30561819 on Build GRCh38
rs774052270 Chr.2: 30562107-30562107 on Build GRCh38
rs1028729256 Chr.2: 30562001-30562001 on Build GRCh38
rs539845326 Chr.2: 30561890-30561890 on Build GRCh38
rs567610025 Chr.2: 30562017-30562017 on Build GRCh38
rs367665468 Chr.2: 30562223-30562223 on Build GRCh38

View Full Product Details