Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 73762321-73762850 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUSP11
Entrez Gene ID
8446
Gene Name
dual specificity phosphatase 11
Gene Aliases
PIR1
UniGene
Hs.14611
Gene Chromosome Location
Chr.2: 73762184-73780157 on Build GRCh38
Transcripts
NM_003584.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7106354 c.1024T>C p.S342P Chr.2: 73762771-73762771 on Build GRCh38
2998684 c.948G>T p.E316D Chr.2: 73762847-73762847 on Build GRCh38
4679702 c.909T>C p.S303S Chr.2: 73762745-73762745 on Build GRCh38
7106353 c.883T>C p.S295P Chr.2: 73762771-73762771 on Build GRCh38
4568046 c.850_851CC>TT p.P284F Chr.2: 73762803-73762804 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs11538238 Chr.2: 73762367-73762367 on Build GRCh38
rs769539660 Chr.2: 73762679-73762679 on Build GRCh38
rs58518125 Chr.2: 73762798-73762798 on Build GRCh38
rs779685569 Chr.2: 73762687-73762687 on Build GRCh38
rs755916818 Chr.2: 73762817-73762817 on Build GRCh38
rs759908477 Chr.2: 73762756-73762756 on Build GRCh38
rs769093589 Chr.2: 73762691-73762691 on Build GRCh38
rs1021562696 Chr.2: 73762640-73762640 on Build GRCh38
rs766735751 Chr.2: 73762628-73762628 on Build GRCh38
rs931556633 Chr.2: 73762740-73762740 on Build GRCh38
rs756497511 Chr.2: 73762619-73762619 on Build GRCh38
rs1023366492 Chr.2: 73762694-73762694 on Build GRCh38
rs780558997 Chr.2: 73762618-73762619 on Build GRCh38
rs140629191 Chr.2: 73762803-73762803 on Build GRCh38
rs768638383 Chr.2: 73762550-73762550 on Build GRCh38
rs537313094 Chr.2: 73762802-73762802 on Build GRCh38
rs145553625 Chr.2: 73762358-73762358 on Build GRCh38
rs933057517 Chr.2: 73762432-73762432 on Build GRCh38
rs372817267 Chr.2: 73762355-73762355 on Build GRCh38
rs757184899 Chr.2: 73762672-73762672 on Build GRCh38
rs1028158704 Chr.2: 73762383-73762384 on Build GRCh38
rs200735058 Chr.2: 73762748-73762748 on Build GRCh38
rs779883515 Chr.2: 73762820-73762820 on Build GRCh38
rs565676184 Chr.2: 73762585-73762585 on Build GRCh38
rs79821643 Chr.2: 73762788-73762788 on Build GRCh38
rs550115739 Chr.2: 73762547-73762547 on Build GRCh38
rs1802053 Chr.2: 73762462-73762462 on Build GRCh38
rs1051237775 Chr.2: 73762461-73762461 on Build GRCh38
rs61485894 Chr.2: 73762799-73762799 on Build GRCh38
rs893680044 Chr.2: 73762516-73762516 on Build GRCh38
rs768350167 Chr.2: 73762697-73762697 on Build GRCh38
rs745687382 Chr.2: 73762674-73762674 on Build GRCh38
rs773725945 Chr.2: 73762744-73762744 on Build GRCh38
rs768242430 Chr.2: 73762822-73762822 on Build GRCh38
rs781169905 Chr.2: 73762673-73762673 on Build GRCh38
rs773774405 Chr.2: 73762710-73762710 on Build GRCh38
rs547464771 Chr.2: 73762579-73762579 on Build GRCh38
rs992034593 Chr.2: 73762711-73762711 on Build GRCh38
rs368196762 Chr.2: 73762658-73762658 on Build GRCh38
rs536357787 Chr.2: 73762746-73762746 on Build GRCh38
rs571449114 Chr.2: 73762553-73762553 on Build GRCh38
rs748259649 Chr.2: 73762667-73762667 on Build GRCh38
rs567574129 Chr.2: 73762800-73762800 on Build GRCh38
rs35245567 Chr.2: 73762400-73762401 on Build GRCh38
rs754044455 Chr.2: 73762636-73762636 on Build GRCh38
rs761361546 Chr.2: 73762717-73762717 on Build GRCh38
rs749051316 Chr.2: 73762821-73762821 on Build GRCh38
rs538861797 Chr.2: 73762563-73762563 on Build GRCh38
rs748882012 Chr.2: 73762696-73762696 on Build GRCh38
rs772476877 Chr.2: 73762729-73762729 on Build GRCh38
rs983380919 Chr.2: 73762810-73762810 on Build GRCh38
rs754238468 Chr.2: 73762754-73762754 on Build GRCh38
rs977406554 Chr.2: 73762659-73762659 on Build GRCh38

View Full Product Details