Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 73766320-73766847 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUSP11
Entrez Gene ID
8446
Gene Name
dual specificity phosphatase 11
Gene Aliases
PIR1
UniGene
Hs.14611
Gene Chromosome Location
Chr.2: 73762184-73780157 on Build GRCh38
Transcripts
NM_003584.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1022858 c.752G>A p.R251Q Chr.2: 73766460-73766460 on Build GRCh38
7152549 c.729G>C p.Q243H Chr.2: 73766483-73766483 on Build GRCh38
7143577 c.759G>A p.K253K Chr.2: 73766594-73766594 on Build GRCh38
7143576 c.618G>A p.K206K Chr.2: 73766594-73766594 on Build GRCh38
6158892 c.935+1G>A p.? Chr.2: 73766417-73766417 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs187369281 Chr.2: 73766626-73766626 on Build GRCh38
rs746542447 Chr.2: 73766484-73766484 on Build GRCh38
rs746489471 Chr.2: 73766406-73766406 on Build GRCh38
rs762677137 Chr.2: 73766347-73766347 on Build GRCh38
rs773148624 Chr.2: 73766545-73766545 on Build GRCh38
rs768887234 Chr.2: 73766523-73766523 on Build GRCh38
rs771618056 Chr.2: 73766407-73766407 on Build GRCh38
rs866489262 Chr.2: 73766729-73766729 on Build GRCh38
rs200079097 Chr.2: 73766817-73766817 on Build GRCh38
rs918915781 Chr.2: 73766419-73766419 on Build GRCh38
rs1002339411 Chr.2: 73766355-73766355 on Build GRCh38
rs746595455 Chr.2: 73766487-73766487 on Build GRCh38
rs925810973 Chr.2: 73766543-73766543 on Build GRCh38
rs759583420 Chr.2: 73766574-73766574 on Build GRCh38
rs780906902 Chr.2: 73766621-73766621 on Build GRCh38
rs749592587 Chr.2: 73766519-73766519 on Build GRCh38
rs965137039 Chr.2: 73766464-73766464 on Build GRCh38
rs772272952 Chr.2: 73766791-73766791 on Build GRCh38
rs116238730 Chr.2: 73766466-73766466 on Build GRCh38
rs753655561 Chr.2: 73766367-73766367 on Build GRCh38
rs542695914 Chr.2: 73766448-73766448 on Build GRCh38
rs373776486 Chr.2: 73766644-73766644 on Build GRCh38
rs777549183 Chr.2: 73766472-73766472 on Build GRCh38
rs575155287 Chr.2: 73766424-73766424 on Build GRCh38
rs780375528 Chr.2: 73766634-73766634 on Build GRCh38
rs774516556 Chr.2: 73766535-73766535 on Build GRCh38
rs761885998 Chr.2: 73766540-73766540 on Build GRCh38
rs777167284 Chr.2: 73766439-73766440 on Build GRCh38
rs768942569 Chr.2: 73766642-73766642 on Build GRCh38
rs539353964 Chr.2: 73766458-73766458 on Build GRCh38
rs533622281 Chr.2: 73766788-73766788 on Build GRCh38
rs775991402 Chr.2: 73766510-73766510 on Build GRCh38
rs189175079 Chr.2: 73766740-73766740 on Build GRCh38
rs549243023 Chr.2: 73766366-73766366 on Build GRCh38
rs770190511 Chr.2: 73766507-73766507 on Build GRCh38
rs778432155 Chr.2: 73766384-73766384 on Build GRCh38
rs757971485 Chr.2: 73766401-73766401 on Build GRCh38
rs773378774 Chr.2: 73766805-73766805 on Build GRCh38
rs746253641 Chr.2: 73766420-73766420 on Build GRCh38
rs978384574 Chr.2: 73766597-73766597 on Build GRCh38
rs763295258 Chr.2: 73766427-73766427 on Build GRCh38
rs544268073 Chr.2: 73766660-73766660 on Build GRCh38
rs777489945 Chr.2: 73766405-73766405 on Build GRCh38
rs138187423 Chr.2: 73766469-73766469 on Build GRCh38
rs1013674926 Chr.2: 73766369-73766369 on Build GRCh38
rs749693067 Chr.2: 73766639-73766639 on Build GRCh38
rs752590032 Chr.2: 73766580-73766580 on Build GRCh38
rs766896290 Chr.2: 73766719-73766721 on Build GRCh38
rs182289530 Chr.2: 73766353-73766353 on Build GRCh38
rs976431674 Chr.2: 73766465-73766465 on Build GRCh38
rs1055169705 Chr.2: 73766653-73766653 on Build GRCh38
rs775253527 Chr.2: 73766821-73766821 on Build GRCh38
rs141473479 Chr.2: 73766426-73766426 on Build GRCh38
rs143838435 Chr.2: 73766544-73766544 on Build GRCh38
rs987122665 Chr.2: 73766754-73766754 on Build GRCh38
rs974541771 Chr.2: 73766522-73766522 on Build GRCh38
rs756686606 Chr.2: 73766633-73766633 on Build GRCh38
rs371782116 Chr.2: 73766463-73766463 on Build GRCh38
rs762790593 Chr.2: 73766583-73766583 on Build GRCh38
rs766334924 Chr.2: 73766440-73766440 on Build GRCh38
rs376761568 Chr.2: 73766387-73766387 on Build GRCh38
rs756792015 Chr.2: 73766485-73766485 on Build GRCh38
rs774461538 Chr.2: 73766438-73766438 on Build GRCh38
rs376777785 Chr.2: 73766810-73766810 on Build GRCh38
rs754738933 Chr.2: 73766460-73766460 on Build GRCh38
rs35896452 Chr.2: 73766485-73766486 on Build GRCh38
rs746328412 Chr.2: 73766505-73766505 on Build GRCh38
rs1027295210 Chr.2: 73766435-73766435 on Build GRCh38
rs201091075 Chr.2: 73766603-73766603 on Build GRCh38
rs761876917 Chr.2: 73766570-73766570 on Build GRCh38
rs540308027 Chr.2: 73766575-73766575 on Build GRCh38
rs574454673 Chr.2: 73766661-73766662 on Build GRCh38
rs768952657 Chr.2: 73766372-73766372 on Build GRCh38
rs376540529 Chr.2: 73766591-73766591 on Build GRCh38
rs899362492 Chr.2: 73766648-73766648 on Build GRCh38
rs199533069 Chr.2: 73766461-73766461 on Build GRCh38
rs756841272 Chr.2: 73766605-73766605 on Build GRCh38
rs774352521 Chr.2: 73766539-73766539 on Build GRCh38
rs764298408 Chr.2: 73766429-73766429 on Build GRCh38
rs147626836 Chr.2: 73766413-73766413 on Build GRCh38
rs145236121 Chr.2: 73766368-73766368 on Build GRCh38

View Full Product Details