Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 74529025-74529525 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AUP1
Entrez Gene ID
550
Gene Name
ancient ubiquitous protein 1
Gene Aliases
-
UniGene
Hs.411480
Gene Chromosome Location
Chr.2: 74526648-74529897 on Build GRCh38
Transcripts
NM_181575.4 NR_126510.1 NR_126511.1 XM_005264392.3 NM_001321727.1 NM_001321728.1 NM_013247.4 NM_145074.2 NR_135769.1 NR_135770.1 NR_135771.1 NR_135772.1

Gene Symbol
HTRA2
Entrez Gene ID
27429
Gene Name
HtrA serine peptidase 2
Gene Aliases
OMI, PARK13, PRSS25
UniGene
Hs.469045
Gene Chromosome Location
Chr.2: 74529405-74533556 on Build GRCh38
Transcripts
NM_181575.4 NR_126510.1 NR_126511.1 XM_005264392.3 NM_001321727.1 NM_001321728.1 NM_013247.4 NM_145074.2 NR_135769.1 NR_135770.1 NR_135771.1 NR_135772.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7226539 c.51-4C>T p.? Chr.2: 74529503-74529503 on Build GRCh38
3758375 c.129C>G p.L43L Chr.2: 74529421-74529421 on Build GRCh38
4467812 c.150C>T p.F50F Chr.2: 74529400-74529400 on Build GRCh38
5993586 c.293C>T p.T98I Chr.2: 74529178-74529178 on Build GRCh38
4455854 c.92A>T p.Y31F Chr.2: 74529458-74529458 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201225621 Chr.2: 74529372-74529372 on Build GRCh38
rs759469312 Chr.2: 74529424-74529424 on Build GRCh38
rs763774026 Chr.2: 74529101-74529101 on Build GRCh38
rs981710668 Chr.2: 74529236-74529236 on Build GRCh38
rs771797848 Chr.2: 74529397-74529397 on Build GRCh38
rs771283342 Chr.2: 74529258-74529258 on Build GRCh38
rs758110745 Chr.2: 74529089-74529089 on Build GRCh38
rs767474615 Chr.2: 74529217-74529217 on Build GRCh38
rs992389652 Chr.2: 74529307-74529307 on Build GRCh38
rs775457709 Chr.2: 74529166-74529166 on Build GRCh38
rs768146322 Chr.2: 74529272-74529272 on Build GRCh38
rs764787115 Chr.2: 74529080-74529080 on Build GRCh38
rs768134316 Chr.2: 74529280-74529280 on Build GRCh38
rs762527856 Chr.2: 74529279-74529279 on Build GRCh38
rs1017400756 Chr.2: 74529225-74529225 on Build GRCh38
rs376231592 Chr.2: 74529460-74529460 on Build GRCh38
rs749701538 Chr.2: 74529201-74529201 on Build GRCh38
rs770389156 Chr.2: 74529110-74529110 on Build GRCh38
rs769713877 Chr.2: 74529503-74529503 on Build GRCh38
rs773623129 Chr.2: 74529067-74529068 on Build GRCh38
rs771373252 Chr.2: 74529158-74529158 on Build GRCh38
rs992422054 Chr.2: 74529462-74529462 on Build GRCh38
rs916937839 Chr.2: 74529312-74529312 on Build GRCh38
rs775115870 Chr.2: 74529198-74529198 on Build GRCh38
rs998050680 Chr.2: 74529133-74529133 on Build GRCh38
rs750637782 Chr.2: 74529447-74529447 on Build GRCh38
rs774037009 Chr.2: 74529290-74529290 on Build GRCh38
rs145348894 Chr.2: 74529330-74529330 on Build GRCh38
rs773679185 Chr.2: 74529062-74529062 on Build GRCh38
rs200897349 Chr.2: 74529197-74529197 on Build GRCh38
rs773078843 Chr.2: 74529336-74529336 on Build GRCh38
rs891752985 Chr.2: 74529129-74529129 on Build GRCh38
rs201711130 Chr.2: 74529161-74529161 on Build GRCh38
rs776448029 Chr.2: 74529496-74529496 on Build GRCh38
rs11539609 Chr.2: 74529242-74529242 on Build GRCh38
rs763064650 Chr.2: 74529108-74529108 on Build GRCh38
rs73949680 Chr.2: 74529115-74529115 on Build GRCh38
rs751662114 Chr.2: 74529355-74529355 on Build GRCh38
rs764140027 Chr.2: 74529306-74529307 on Build GRCh38
rs747668874 Chr.2: 74529395-74529395 on Build GRCh38
rs770132475 Chr.2: 74529048-74529048 on Build GRCh38
rs752343183 Chr.2: 74529086-74529086 on Build GRCh38
rs781277292 Chr.2: 74529120-74529120 on Build GRCh38
rs377186972 Chr.2: 74529405-74529405 on Build GRCh38
rs773412790 Chr.2: 74529149-74529149 on Build GRCh38
rs898610053 Chr.2: 74529393-74529393 on Build GRCh38
rs761712133 Chr.2: 74529214-74529214 on Build GRCh38
rs745653406 Chr.2: 74529501-74529501 on Build GRCh38
rs756156375 Chr.2: 74529230-74529230 on Build GRCh38
rs766905811 Chr.2: 74529069-74529069 on Build GRCh38
rs568571027 Chr.2: 74529107-74529107 on Build GRCh38
rs374920826 Chr.2: 74529348-74529348 on Build GRCh38
rs756486079 Chr.2: 74529380-74529380 on Build GRCh38
rs767386400 Chr.2: 74529297-74529297 on Build GRCh38
rs749768893 Chr.2: 74529384-74529384 on Build GRCh38
rs766568020 Chr.2: 74529117-74529122 on Build GRCh38
rs71640290 Chr.2: 74529329-74529329 on Build GRCh38
rs774347474 Chr.2: 74529433-74529433 on Build GRCh38
rs1183739 Chr.2: 74529421-74529421 on Build GRCh38
rs777467471 Chr.2: 74529475-74529475 on Build GRCh38
rs760652784 Chr.2: 74529301-74529301 on Build GRCh38
rs755092106 Chr.2: 74529124-74529124 on Build GRCh38
rs750379017 Chr.2: 74529222-74529222 on Build GRCh38
rs770306665 Chr.2: 74529267-74529267 on Build GRCh38
rs772365793 Chr.2: 74529144-74529144 on Build GRCh38
rs2231250 Chr.2: 74529049-74529049 on Build GRCh38
rs367810642 Chr.2: 74529455-74529455 on Build GRCh38
rs76602278 Chr.2: 74529352-74529352 on Build GRCh38
rs371919390 Chr.2: 74529259-74529259 on Build GRCh38
rs370173846 Chr.2: 74529456-74529456 on Build GRCh38
rs372473706 Chr.2: 74529483-74529483 on Build GRCh38
rs752665073 Chr.2: 74529308-74529308 on Build GRCh38
rs958958584 Chr.2: 74529430-74529430 on Build GRCh38
rs551674975 Chr.2: 74529495-74529495 on Build GRCh38
rs761530296 Chr.2: 74529331-74529331 on Build GRCh38
rs753544200 Chr.2: 74529339-74529340 on Build GRCh38
rs749320680 Chr.2: 74529121-74529121 on Build GRCh38
rs180981125 Chr.2: 74529327-74529327 on Build GRCh38
rs71640289 Chr.2: 74529319-74529319 on Build GRCh38
rs989361947 Chr.2: 74529122-74529122 on Build GRCh38
rs760103877 Chr.2: 74529163-74529163 on Build GRCh38
rs767639720 Chr.2: 74529436-74529436 on Build GRCh38
rs759061585 Chr.2: 74529073-74529073 on Build GRCh38
rs765119733 Chr.2: 74529431-74529431 on Build GRCh38
rs376150821 Chr.2: 74529326-74529326 on Build GRCh38
rs113870495 Chr.2: 74529131-74529131 on Build GRCh38
rs200853443 Chr.2: 74529407-74529408 on Build GRCh38
rs376362584 Chr.2: 74529354-74529354 on Build GRCh38
rs746353321 Chr.2: 74529261-74529261 on Build GRCh38
rs758493924 Chr.2: 74529309-74529309 on Build GRCh38
rs760551183 Chr.2: 74529229-74529231 on Build GRCh38
rs762463747 Chr.2: 74529200-74529200 on Build GRCh38
rs755468782 Chr.2: 74529465-74529465 on Build GRCh38
rs779197795 Chr.2: 74529132-74529132 on Build GRCh38
rs963698458 Chr.2: 74529068-74529068 on Build GRCh38
rs757430283 Chr.2: 74529358-74529358 on Build GRCh38
rs758485212 Chr.2: 74529239-74529239 on Build GRCh38
rs373539917 Chr.2: 74529344-74529344 on Build GRCh38
rs752857814 Chr.2: 74529238-74529238 on Build GRCh38
rs779580959 Chr.2: 74529468-74529468 on Build GRCh38
rs766348718 Chr.2: 74529303-74529303 on Build GRCh38
rs937305815 Chr.2: 74529249-74529249 on Build GRCh38
rs757524718 Chr.2: 74529317-74529317 on Build GRCh38
rs1032054779 Chr.2: 74529083-74529083 on Build GRCh38
rs776054411 Chr.2: 74529071-74529071 on Build GRCh38
rs747408018 Chr.2: 74529155-74529155 on Build GRCh38
rs778434732 Chr.2: 74529388-74529388 on Build GRCh38
rs368321899 Chr.2: 74529248-74529248 on Build GRCh38
rs746261566 Chr.2: 74529324-74529324 on Build GRCh38
rs750726884 Chr.2: 74529373-74529373 on Build GRCh38
rs71640291 Chr.2: 74529409-74529409 on Build GRCh38
rs368530188 Chr.2: 74529472-74529472 on Build GRCh38
rs761622172 Chr.2: 74529291-74529291 on Build GRCh38
rs535031131 Chr.2: 74529057-74529057 on Build GRCh38
rs757101193 Chr.2: 74529117-74529117 on Build GRCh38
rs780346944 Chr.2: 74529383-74529383 on Build GRCh38
rs751243813 Chr.2: 74529209-74529209 on Build GRCh38
rs368880053 Chr.2: 74529335-74529335 on Build GRCh38
rs575830403 Chr.2: 74529231-74529231 on Build GRCh38
rs553462817 Chr.2: 74529064-74529064 on Build GRCh38
rs115467431 Chr.2: 74529387-74529387 on Build GRCh38
rs770511647 Chr.2: 74529494-74529494 on Build GRCh38
rs201319345 Chr.2: 74529316-74529316 on Build GRCh38

View Full Product Details