Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 74877062-74877568 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HK2
Entrez Gene ID
3099
Gene Name
hexokinase 2
Gene Aliases
HKII, HXK2
UniGene
Hs.406266
Gene Chromosome Location
Chr.2: 74832655-74893354 on Build GRCh38
Transcripts
NM_000189.4 XM_005264280.1 XM_011532807.1 XM_017003945.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
722457 c.962G>T p.G321V Chr.2: 74877252-74877252 on Build GRCh38
1662152 c.968A>G p.K323R Chr.2: 74877258-74877258 on Build GRCh38
6680587 c.909G>A p.G303G Chr.2: 74877199-74877199 on Build GRCh38
1023105 c.941C>T p.A314V Chr.2: 74877231-74877231 on Build GRCh38
7108567 c.1027G>A p.E343K Chr.2: 74877317-74877317 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775858636 Chr.2: 74877446-74877446 on Build GRCh38
rs374980610 Chr.2: 74877316-74877316 on Build GRCh38
rs781520176 Chr.2: 74877128-74877128 on Build GRCh38
rs771668641 Chr.2: 74877249-74877249 on Build GRCh38
rs577669528 Chr.2: 74877373-74877373 on Build GRCh38
rs28363013 Chr.2: 74877115-74877115 on Build GRCh38
rs377352232 Chr.2: 74877161-74877161 on Build GRCh38
rs3771759 Chr.2: 74877448-74877448 on Build GRCh38
rs1016770948 Chr.2: 74877269-74877269 on Build GRCh38
rs781179001 Chr.2: 74877292-74877292 on Build GRCh38
rs771092061 Chr.2: 74877126-74877126 on Build GRCh38
rs143931243 Chr.2: 74877256-74877256 on Build GRCh38
rs767044426 Chr.2: 74877159-74877159 on Build GRCh38
rs367965703 Chr.2: 74877322-74877322 on Build GRCh38
rs570410813 Chr.2: 74877411-74877411 on Build GRCh38
rs116187651 Chr.2: 74877403-74877403 on Build GRCh38
rs1002065031 Chr.2: 74877397-74877397 on Build GRCh38
rs543436538 Chr.2: 74877489-74877489 on Build GRCh38
rs370773572 Chr.2: 74877199-74877199 on Build GRCh38
rs780579734 Chr.2: 74877238-74877238 on Build GRCh38
rs28363016 Chr.2: 74877282-74877282 on Build GRCh38
rs565854734 Chr.2: 74877241-74877241 on Build GRCh38
rs28363014 Chr.2: 74877202-74877202 on Build GRCh38
rs913494021 Chr.2: 74877495-74877495 on Build GRCh38
rs1016928219 Chr.2: 74877402-74877402 on Build GRCh38
rs367623952 Chr.2: 74877364-74877364 on Build GRCh38
rs760447271 Chr.2: 74877166-74877166 on Build GRCh38
rs373781772 Chr.2: 74877342-74877342 on Build GRCh38
rs28363020 Chr.2: 74877545-74877545 on Build GRCh38
rs61751209 Chr.2: 74877283-74877283 on Build GRCh38
rs960785519 Chr.2: 74877519-74877519 on Build GRCh38
rs939393316 Chr.2: 74877090-74877090 on Build GRCh38
rs762961790 Chr.2: 74877365-74877365 on Build GRCh38
rs779271072 Chr.2: 74877320-74877320 on Build GRCh38
rs28363018 Chr.2: 74877442-74877442 on Build GRCh38
rs538810721 Chr.2: 74877546-74877546 on Build GRCh38
rs916810698 Chr.2: 74877111-74877111 on Build GRCh38
rs537840475 Chr.2: 74877332-74877332 on Build GRCh38
rs777873822 Chr.2: 74877196-74877196 on Build GRCh38
rs571490900 Chr.2: 74877501-74877501 on Build GRCh38
rs770543661 Chr.2: 74877338-74877338 on Build GRCh38
rs765147151 Chr.2: 74877362-74877362 on Build GRCh38
rs749039453 Chr.2: 74877190-74877190 on Build GRCh38
rs781259155 Chr.2: 74877205-74877205 on Build GRCh38
rs763559200 Chr.2: 74877140-74877140 on Build GRCh38
rs994067907 Chr.2: 74877410-74877410 on Build GRCh38
rs993974874 Chr.2: 74877502-74877502 on Build GRCh38
rs756426288 Chr.2: 74877288-74877288 on Build GRCh38
rs955164292 Chr.2: 74877434-74877434 on Build GRCh38
rs754933750 Chr.2: 74877194-74877194 on Build GRCh38
rs750616279 Chr.2: 74877287-74877287 on Build GRCh38
rs559528705 Chr.2: 74877508-74877508 on Build GRCh38
rs918590464 Chr.2: 74877086-74877086 on Build GRCh38
rs773756011 Chr.2: 74877146-74877146 on Build GRCh38
rs935015185 Chr.2: 74877460-74877460 on Build GRCh38
rs761484246 Chr.2: 74877152-74877152 on Build GRCh38
rs200191687 Chr.2: 74877138-74877138 on Build GRCh38
rs777099070 Chr.2: 74877148-74877149 on Build GRCh38
rs749763551 Chr.2: 74877239-74877239 on Build GRCh38
rs906060193 Chr.2: 74877515-74877516 on Build GRCh38
rs752427956 Chr.2: 74877328-74877329 on Build GRCh38
rs36066402 Chr.2: 74877252-74877252 on Build GRCh38
rs10153634 Chr.2: 74877091-74877091 on Build GRCh38
rs768313454 Chr.2: 74877507-74877507 on Build GRCh38
rs774960625 Chr.2: 74877244-74877244 on Build GRCh38
rs199736350 Chr.2: 74877192-74877192 on Build GRCh38
rs772842318 Chr.2: 74877251-74877251 on Build GRCh38
rs28363015 Chr.2: 74877230-74877230 on Build GRCh38
rs767656546 Chr.2: 74877372-74877372 on Build GRCh38
rs772719225 Chr.2: 74877327-74877327 on Build GRCh38
rs779890863 Chr.2: 74877121-74877121 on Build GRCh38
rs543990926 Chr.2: 74877138-74877139 on Build GRCh38
rs770217860 Chr.2: 74877225-74877225 on Build GRCh38
rs1006810437 Chr.2: 74877499-74877499 on Build GRCh38
rs3771760 Chr.2: 74877443-74877443 on Build GRCh38
rs564973701 Chr.2: 74877497-74877497 on Build GRCh38
rs749637190 Chr.2: 74877313-74877313 on Build GRCh38
rs28363017 Chr.2: 74877428-74877428 on Build GRCh38
rs370257597 Chr.2: 74877341-74877341 on Build GRCh38
rs988991955 Chr.2: 74877485-74877485 on Build GRCh38
rs35677390 Chr.2: 74877284-74877284 on Build GRCh38
rs373294336 Chr.2: 74877160-74877160 on Build GRCh38
rs532213676 Chr.2: 74877500-74877500 on Build GRCh38
rs28363019 Chr.2: 74877543-74877543 on Build GRCh38
rs780526388 Chr.2: 74877305-74877305 on Build GRCh38
rs768154230 Chr.2: 74877141-74877141 on Build GRCh38
rs766224616 Chr.2: 74877188-74877188 on Build GRCh38
rs746161634 Chr.2: 74877221-74877221 on Build GRCh38
rs767527984 Chr.2: 74877262-74877264 on Build GRCh38
rs561405547 Chr.2: 74877447-74877447 on Build GRCh38
rs761204336 Chr.2: 74877247-74877247 on Build GRCh38
rs746215216 Chr.2: 74877131-74877131 on Build GRCh38
rs889485412 Chr.2: 74877470-74877470 on Build GRCh38

View Full Product Details