Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 75047114-75047613 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TACR1
Entrez Gene ID
6869
Gene Name
tachykinin receptor 1
Gene Aliases
NK1R, NKIR, SPR, TAC1R
UniGene
Hs.633301
Gene Chromosome Location
Chr.2: 75046463-75199519 on Build GRCh38
Transcripts
NM_001058.3

SNP Detail

SNP ID Chromosome Location
rs958706121 Chr.2: 75047214-75047214 on Build GRCh38
rs1030371643 Chr.2: 75047364-75047367 on Build GRCh38
rs1048719937 Chr.2: 75047406-75047406 on Build GRCh38
rs572923041 Chr.2: 75047160-75047160 on Build GRCh38
rs568056912 Chr.2: 75047165-75047166 on Build GRCh38
rs183901589 Chr.2: 75047503-75047503 on Build GRCh38
rs948415907 Chr.2: 75047522-75047522 on Build GRCh38
rs151139727 Chr.2: 75047436-75047436 on Build GRCh38
rs148625007 Chr.2: 75047245-75047245 on Build GRCh38
rs912714689 Chr.2: 75047352-75047352 on Build GRCh38
rs931640495 Chr.2: 75047395-75047395 on Build GRCh38
rs571044369 Chr.2: 75047555-75047555 on Build GRCh38
rs746817792 Chr.2: 75047454-75047454 on Build GRCh38
rs373800679 Chr.2: 75047348-75047348 on Build GRCh38
rs72824221 Chr.2: 75047216-75047216 on Build GRCh38
rs907058785 Chr.2: 75047521-75047521 on Build GRCh38
rs181354818 Chr.2: 75047218-75047218 on Build GRCh38
rs770033626 Chr.2: 75047544-75047544 on Build GRCh38
rs954909092 Chr.2: 75047371-75047371 on Build GRCh38
rs1056824913 Chr.2: 75047178-75047178 on Build GRCh38
rs774385459 Chr.2: 75047377-75047377 on Build GRCh38
rs944166040 Chr.2: 75047358-75047358 on Build GRCh38
rs1002627378 Chr.2: 75047525-75047525 on Build GRCh38
rs897923505 Chr.2: 75047332-75047332 on Build GRCh38
rs924374422 Chr.2: 75047154-75047154 on Build GRCh38
rs1034366944 Chr.2: 75047164-75047164 on Build GRCh38
rs775525608 Chr.2: 75047587-75047587 on Build GRCh38
rs563483464 Chr.2: 75047361-75047361 on Build GRCh38
rs992614434 Chr.2: 75047491-75047491 on Build GRCh38
rs887725708 Chr.2: 75047412-75047412 on Build GRCh38
rs921648641 Chr.2: 75047383-75047383 on Build GRCh38
rs1002930738 Chr.2: 75047162-75047162 on Build GRCh38
rs777616491 Chr.2: 75047380-75047380 on Build GRCh38
rs1017962623 Chr.2: 75047387-75047387 on Build GRCh38
rs1006821449 Chr.2: 75047263-75047263 on Build GRCh38
rs142137038 Chr.2: 75047319-75047319 on Build GRCh38
rs1022331721 Chr.2: 75047225-75047225 on Build GRCh38

View Full Product Details