Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 86211047-86211547 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MRPL35
Entrez Gene ID
51318
Gene Name
mitochondrial ribosomal protein L35
Gene Aliases
L35mt, MRP-L35
UniGene
Hs.433439
Gene Chromosome Location
Chr.2: 86199355-86213794 on Build GRCh38
Transcripts
NM_016622.3 NM_145644.2 XM_005264361.2

SNP Detail

SNP ID Chromosome Location
rs1027896775 Chr.2: 86211520-86211520 on Build GRCh38
rs1046967400 Chr.2: 86211349-86211358 on Build GRCh38
rs995620746 Chr.2: 86211513-86211513 on Build GRCh38
rs955790105 Chr.2: 86211496-86211496 on Build GRCh38
rs1038122434 Chr.2: 86211521-86211521 on Build GRCh38
rs978366478 Chr.2: 86211154-86211154 on Build GRCh38
rs372622357 Chr.2: 86211308-86211308 on Build GRCh38
rs991649668 Chr.2: 86211122-86211122 on Build GRCh38
rs878940443 Chr.2: 86211229-86211229 on Build GRCh38
rs1002792824 Chr.2: 86211367-86211367 on Build GRCh38
rs911411983 Chr.2: 86211237-86211237 on Build GRCh38
rs779588153 Chr.2: 86211397-86211397 on Build GRCh38
rs907080606 Chr.2: 86211350-86211350 on Build GRCh38
rs754486323 Chr.2: 86211435-86211435 on Build GRCh38
rs911380912 Chr.2: 86211515-86211515 on Build GRCh38
rs564726899 Chr.2: 86211478-86211478 on Build GRCh38
rs766931412 Chr.2: 86211386-86211386 on Build GRCh38
rs1034119857 Chr.2: 86211381-86211381 on Build GRCh38
rs898144797 Chr.2: 86211280-86211280 on Build GRCh38
rs541471869 Chr.2: 86211455-86211455 on Build GRCh38
rs950157918 Chr.2: 86211150-86211150 on Build GRCh38
rs958860235 Chr.2: 86211091-86211091 on Build GRCh38
rs755743188 Chr.2: 86211291-86211291 on Build GRCh38
rs958543505 Chr.2: 86211396-86211396 on Build GRCh38
rs763187503 Chr.2: 86211264-86211264 on Build GRCh38
rs920723145 Chr.2: 86211086-86211086 on Build GRCh38
rs765570427 Chr.2: 86211106-86211106 on Build GRCh38
rs748766989 Chr.2: 86211475-86211475 on Build GRCh38
rs17438451 Chr.2: 86211477-86211477 on Build GRCh38
rs924224743 Chr.2: 86211155-86211155 on Build GRCh38
rs924647745 Chr.2: 86211193-86211193 on Build GRCh38
rs917317428 Chr.2: 86211137-86211137 on Build GRCh38
rs532827614 Chr.2: 86211457-86211457 on Build GRCh38
rs934270802 Chr.2: 86211164-86211164 on Build GRCh38
rs890855695 Chr.2: 86211440-86211440 on Build GRCh38
rs1012270731 Chr.2: 86211419-86211419 on Build GRCh38
rs146749477 Chr.2: 86211190-86211190 on Build GRCh38
rs942399455 Chr.2: 86211238-86211238 on Build GRCh38
rs899783501 Chr.2: 86211320-86211320 on Build GRCh38
rs556041820 Chr.2: 86211443-86211443 on Build GRCh38
rs79849456 Chr.2: 86211420-86211420 on Build GRCh38
rs117827858 Chr.2: 86211466-86211466 on Build GRCh38
rs532316692 Chr.2: 86211076-86211076 on Build GRCh38
rs555701177 Chr.2: 86211251-86211251 on Build GRCh38
rs560601265 Chr.2: 86211461-86211461 on Build GRCh38
rs968477807 Chr.2: 86211138-86211138 on Build GRCh38

View Full Product Details