Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 88729188-88729700 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPIA
Entrez Gene ID
22934
Gene Name
ribose 5-phosphate isomerase A
Gene Aliases
RPI, RPIAD
UniGene
Hs.469264
Gene Chromosome Location
Chr.2: 88691658-88750935 on Build GRCh38
Transcripts
NM_144563.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4722584 c.423G>A p.Q141Q Chr.2: 88729298-88729298 on Build GRCh38
76286 c.428G>A p.G143D Chr.2: 88729303-88729303 on Build GRCh38
1224200 c.406C>T p.R136C Chr.2: 88729281-88729281 on Build GRCh38
1023617 c.452G>A p.R151Q Chr.2: 88729327-88729327 on Build GRCh38
7388123 c.423G>T p.Q141H Chr.2: 88729298-88729298 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs975950516 Chr.2: 88729600-88729600 on Build GRCh38
rs370133682 Chr.2: 88729648-88729648 on Build GRCh38
rs558813228 Chr.2: 88729624-88729624 on Build GRCh38
rs878874101 Chr.2: 88729433-88729433 on Build GRCh38
rs931608761 Chr.2: 88729329-88729329 on Build GRCh38
rs374038384 Chr.2: 88729516-88729517 on Build GRCh38
rs121918591 Chr.2: 88729279-88729279 on Build GRCh38
rs767607229 Chr.2: 88729250-88729250 on Build GRCh38
rs771026725 Chr.2: 88729334-88729339 on Build GRCh38
rs1009551971 Chr.2: 88729561-88729561 on Build GRCh38
rs773038072 Chr.2: 88729318-88729318 on Build GRCh38
rs760452079 Chr.2: 88729326-88729326 on Build GRCh38
rs550841076 Chr.2: 88729261-88729261 on Build GRCh38
rs1021091816 Chr.2: 88729573-88729573 on Build GRCh38
rs371893786 Chr.2: 88729369-88729369 on Build GRCh38
rs375171924 Chr.2: 88729248-88729248 on Build GRCh38
rs537469091 Chr.2: 88729526-88729526 on Build GRCh38
rs763137724 Chr.2: 88729338-88729338 on Build GRCh38
rs1036099371 Chr.2: 88729382-88729382 on Build GRCh38
rs566953511 Chr.2: 88729439-88729439 on Build GRCh38
rs374027178 Chr.2: 88729635-88729635 on Build GRCh38
rs1025008186 Chr.2: 88729590-88729590 on Build GRCh38
rs572164680 Chr.2: 88729609-88729609 on Build GRCh38
rs773059328 Chr.2: 88729251-88729251 on Build GRCh38
rs1054455133 Chr.2: 88729549-88729549 on Build GRCh38
rs370696470 Chr.2: 88729274-88729274 on Build GRCh38
rs553983149 Chr.2: 88729601-88729601 on Build GRCh38
rs548333806 Chr.2: 88729402-88729402 on Build GRCh38
rs145397758 Chr.2: 88729470-88729470 on Build GRCh38
rs368173743 Chr.2: 88729264-88729264 on Build GRCh38
rs1009622311 Chr.2: 88729350-88729350 on Build GRCh38
rs532436876 Chr.2: 88729224-88729224 on Build GRCh38
rs75541898 Chr.2: 88729620-88729620 on Build GRCh38
rs967198024 Chr.2: 88729577-88729577 on Build GRCh38
rs772127025 Chr.2: 88729307-88729307 on Build GRCh38
rs764179937 Chr.2: 88729266-88729266 on Build GRCh38
rs879151446 Chr.2: 88729345-88729345 on Build GRCh38
rs747731446 Chr.2: 88729301-88729301 on Build GRCh38
rs35747781 Chr.2: 88729311-88729311 on Build GRCh38
rs1009640094 Chr.2: 88729563-88729564 on Build GRCh38
rs751686558 Chr.2: 88729267-88729267 on Build GRCh38
rs565250547 Chr.2: 88729223-88729223 on Build GRCh38
rs974397580 Chr.2: 88729409-88729409 on Build GRCh38
rs374907215 Chr.2: 88729380-88729380 on Build GRCh38
rs773970466 Chr.2: 88729300-88729300 on Build GRCh38
rs367917981 Chr.2: 88729327-88729327 on Build GRCh38
rs775733975 Chr.2: 88729332-88729332 on Build GRCh38
rs774389187 Chr.2: 88729238-88729238 on Build GRCh38
rs540395081 Chr.2: 88729218-88729218 on Build GRCh38
rs892447440 Chr.2: 88729550-88729550 on Build GRCh38
rs750895646 Chr.2: 88729389-88729389 on Build GRCh38
rs56084203 Chr.2: 88729548-88729548 on Build GRCh38
rs570409867 Chr.2: 88729500-88729500 on Build GRCh38
rs982474475 Chr.2: 88729457-88729457 on Build GRCh38
rs749178974 Chr.2: 88729406-88729406 on Build GRCh38
rs757729245 Chr.2: 88729272-88729272 on Build GRCh38
rs535755164 Chr.2: 88729584-88729584 on Build GRCh38
rs919566650 Chr.2: 88729417-88729417 on Build GRCh38
rs768474586 Chr.2: 88729290-88729290 on Build GRCh38
rs774680836 Chr.2: 88729376-88729376 on Build GRCh38
rs375132883 Chr.2: 88729281-88729281 on Build GRCh38
rs191835603 Chr.2: 88729634-88729634 on Build GRCh38
rs905451123 Chr.2: 88729542-88729542 on Build GRCh38
rs371097529 Chr.2: 88729230-88729230 on Build GRCh38
rs186731447 Chr.2: 88729273-88729273 on Build GRCh38
rs530221394 Chr.2: 88729298-88729298 on Build GRCh38
rs115591937 Chr.2: 88729531-88729531 on Build GRCh38
rs1002736065 Chr.2: 88729608-88729608 on Build GRCh38
rs765442781 Chr.2: 88729257-88729257 on Build GRCh38
rs569764920 Chr.2: 88729492-88729492 on Build GRCh38
rs376246983 Chr.2: 88729513-88729513 on Build GRCh38
rs900039030 Chr.2: 88729541-88729541 on Build GRCh38
rs562892316 Chr.2: 88729280-88729280 on Build GRCh38
rs200168324 Chr.2: 88729381-88729381 on Build GRCh38

View Full Product Details