Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 101466686-101467186 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RFX8
Entrez Gene ID
731220
Gene Name
RFX family member 8, lacking RFX DNA binding domain
Gene Aliases
-
UniGene
Hs.662488
Gene Chromosome Location
Chr.2: 101397344-101475179 on Build GRCh38
Transcripts
NM_001145664.1 XM_011511771.2 XM_011511772.1 XM_011511776.1 XM_017004851.1 XM_017004852.1 XM_017004853.1 XM_017004854.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4998217 c.171C>A p.N57K Chr.2: 101466804-101466804 on Build GRCh38
4618076 c.1A>G p.M1V Chr.2: 101466848-101466848 on Build GRCh38
4618077 c.127A>G p.M43V Chr.2: 101466848-101466848 on Build GRCh38
4556670 c.69G>A p.G23G Chr.2: 101466780-101466780 on Build GRCh38
5459795 c.16G>A p.V6M Chr.2: 101466833-101466833 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs564114502 Chr.2: 101466811-101466811 on Build GRCh38
rs976976328 Chr.2: 101467038-101467038 on Build GRCh38
rs753632835 Chr.2: 101466998-101466998 on Build GRCh38
rs750307187 Chr.2: 101466730-101466730 on Build GRCh38
rs776734967 Chr.2: 101466753-101466753 on Build GRCh38
rs924068228 Chr.2: 101466949-101466949 on Build GRCh38
rs954055787 Chr.2: 101467049-101467049 on Build GRCh38
rs949881696 Chr.2: 101466930-101466930 on Build GRCh38
rs115757451 Chr.2: 101466924-101466924 on Build GRCh38
rs111857117 Chr.2: 101466793-101466793 on Build GRCh38
rs973141790 Chr.2: 101466773-101466773 on Build GRCh38
rs945248560 Chr.2: 101467084-101467084 on Build GRCh38
rs1033770993 Chr.2: 101466950-101466950 on Build GRCh38
rs755597802 Chr.2: 101466845-101466845 on Build GRCh38
rs894780476 Chr.2: 101466981-101466981 on Build GRCh38
rs942961178 Chr.2: 101467032-101467032 on Build GRCh38
rs202220240 Chr.2: 101466857-101466857 on Build GRCh38
rs778484548 Chr.2: 101466894-101466894 on Build GRCh38
rs976754468 Chr.2: 101466849-101466849 on Build GRCh38
rs535426317 Chr.2: 101466719-101466719 on Build GRCh38
rs138173220 Chr.2: 101466740-101466740 on Build GRCh38
rs1042317359 Chr.2: 101466896-101466896 on Build GRCh38
rs1015731925 Chr.2: 101467019-101467019 on Build GRCh38
rs749703035 Chr.2: 101466988-101466988 on Build GRCh38
rs921284084 Chr.2: 101467045-101467045 on Build GRCh38
rs578031705 Chr.2: 101466863-101466863 on Build GRCh38
rs1046448488 Chr.2: 101466738-101466738 on Build GRCh38
rs1053852913 Chr.2: 101466875-101466875 on Build GRCh38
rs11680478 Chr.2: 101466931-101466931 on Build GRCh38
rs745534788 Chr.2: 101466844-101466844 on Build GRCh38
rs766614514 Chr.2: 101466750-101466750 on Build GRCh38
rs759594058 Chr.2: 101466754-101466754 on Build GRCh38
rs949518561 Chr.2: 101466735-101466735 on Build GRCh38
rs183254040 Chr.2: 101466792-101466792 on Build GRCh38
rs539779111 Chr.2: 101467069-101467069 on Build GRCh38
rs550080131 Chr.2: 101467146-101467146 on Build GRCh38
rs375204190 Chr.2: 101466836-101466836 on Build GRCh38
rs989998802 Chr.2: 101467066-101467066 on Build GRCh38
rs1005293714 Chr.2: 101467095-101467095 on Build GRCh38
rs528166444 Chr.2: 101466978-101466978 on Build GRCh38
rs901347344 Chr.2: 101467042-101467042 on Build GRCh38
rs371383581 Chr.2: 101466833-101466833 on Build GRCh38
rs991403121 Chr.2: 101466920-101466920 on Build GRCh38
rs915319071 Chr.2: 101467068-101467068 on Build GRCh38
rs1027393848 Chr.2: 101466992-101466992 on Build GRCh38
rs959386701 Chr.2: 101466769-101466769 on Build GRCh38
rs188622884 Chr.2: 101466983-101466983 on Build GRCh38
rs1004381516 Chr.2: 101467001-101467001 on Build GRCh38
rs764083715 Chr.2: 101466796-101466796 on Build GRCh38
rs761190800 Chr.2: 101466731-101466731 on Build GRCh38
rs543718145 Chr.2: 101466871-101466871 on Build GRCh38
rs747323425 Chr.2: 101466923-101466923 on Build GRCh38
rs769269990 Chr.2: 101467133-101467133 on Build GRCh38
rs570691604 Chr.2: 101467028-101467028 on Build GRCh38
rs4851455 Chr.2: 101466721-101466721 on Build GRCh38
rs909540021 Chr.2: 101466883-101466883 on Build GRCh38
rs773847408 Chr.2: 101467083-101467084 on Build GRCh38
rs143815142 Chr.2: 101467154-101467154 on Build GRCh38
rs965629031 Chr.2: 101467037-101467037 on Build GRCh38
rs908787256 Chr.2: 101466834-101466834 on Build GRCh38
rs559409820 Chr.2: 101466968-101466968 on Build GRCh38
rs368285484 Chr.2: 101466893-101466893 on Build GRCh38
rs1018884190 Chr.2: 101466801-101466801 on Build GRCh38

View Full Product Details