Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 102473615-102474114 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC9A4
Entrez Gene ID
389015
Gene Name
solute carrier family 9 member A4
Gene Aliases
NHE4
UniGene
Hs.447686
Gene Chromosome Location
Chr.2: 102473282-102533974 on Build GRCh38
Transcripts
NM_001011552.3 XM_011511158.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3938444 c.225C>T p.L75L Chr.2: 102473984-102473984 on Build GRCh38
5539327 c.163G>A p.E55K Chr.2: 102473922-102473922 on Build GRCh38
1004689 c.18C>T p.F6F Chr.2: 102473777-102473777 on Build GRCh38
714929 c.115C>A p.Q39K Chr.2: 102473874-102473874 on Build GRCh38
6086661 c.139G>T p.A47S Chr.2: 102473898-102473898 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs80335292 Chr.2: 102473778-102473778 on Build GRCh38
rs948559517 Chr.2: 102474085-102474085 on Build GRCh38
rs893005355 Chr.2: 102474074-102474074 on Build GRCh38
rs371028871 Chr.2: 102474032-102474032 on Build GRCh38
rs934553316 Chr.2: 102474056-102474056 on Build GRCh38
rs190587056 Chr.2: 102473811-102473811 on Build GRCh38
rs559642948 Chr.2: 102473808-102473808 on Build GRCh38
rs768345188 Chr.2: 102473791-102473791 on Build GRCh38
rs374368102 Chr.2: 102474015-102474015 on Build GRCh38
rs771496813 Chr.2: 102473804-102473804 on Build GRCh38
rs267598809 Chr.2: 102473844-102473844 on Build GRCh38
rs764448522 Chr.2: 102473931-102473932 on Build GRCh38
rs529743314 Chr.2: 102473660-102473661 on Build GRCh38
rs371139309 Chr.2: 102474030-102474030 on Build GRCh38
rs766505113 Chr.2: 102474039-102474039 on Build GRCh38
rs753958302 Chr.2: 102474001-102474001 on Build GRCh38
rs975470865 Chr.2: 102473882-102473882 on Build GRCh38
rs544930043 Chr.2: 102473656-102473657 on Build GRCh38
rs780847863 Chr.2: 102474074-102474074 on Build GRCh38
rs756810719 Chr.2: 102474062-102474062 on Build GRCh38
rs1040308240 Chr.2: 102473744-102473744 on Build GRCh38
rs369689038 Chr.2: 102473860-102473860 on Build GRCh38
rs377276813 Chr.2: 102473777-102473777 on Build GRCh38
rs754307617 Chr.2: 102473722-102473722 on Build GRCh38
rs149578748 Chr.2: 102473800-102473800 on Build GRCh38
rs760680118 Chr.2: 102474033-102474033 on Build GRCh38
rs775330590 Chr.2: 102473782-102473782 on Build GRCh38
rs56996196 Chr.2: 102474023-102474023 on Build GRCh38
rs767601993 Chr.2: 102473943-102473943 on Build GRCh38
rs748906541 Chr.2: 102473790-102473790 on Build GRCh38
rs140854922 Chr.2: 102473874-102473874 on Build GRCh38
rs144217500 Chr.2: 102473825-102473825 on Build GRCh38
rs370042410 Chr.2: 102473812-102473812 on Build GRCh38
rs747439909 Chr.2: 102473641-102473641 on Build GRCh38
rs754756723 Chr.2: 102473970-102473970 on Build GRCh38
rs547985685 Chr.2: 102473711-102473711 on Build GRCh38
rs776059407 Chr.2: 102473956-102473956 on Build GRCh38
rs752389310 Chr.2: 102474048-102474048 on Build GRCh38
rs765578853 Chr.2: 102473745-102473745 on Build GRCh38
rs780912681 Chr.2: 102474063-102474063 on Build GRCh38
rs77432424 Chr.2: 102473701-102473701 on Build GRCh38
rs777508052 Chr.2: 102473983-102473983 on Build GRCh38
rs768469796 Chr.2: 102473870-102473870 on Build GRCh38
rs148291038 Chr.2: 102473958-102473958 on Build GRCh38
rs762003986 Chr.2: 102473936-102473936 on Build GRCh38
rs771191956 Chr.2: 102473653-102473653 on Build GRCh38
rs985931475 Chr.2: 102474016-102474016 on Build GRCh38
rs570988424 Chr.2: 102473892-102473892 on Build GRCh38
rs770379616 Chr.2: 102473889-102473889 on Build GRCh38
rs777226125 Chr.2: 102474057-102474057 on Build GRCh38
rs756206918 Chr.2: 102473952-102473952 on Build GRCh38
rs549413681 Chr.2: 102473881-102473881 on Build GRCh38
rs543833098 Chr.2: 102474061-102474061 on Build GRCh38
rs537436615 Chr.2: 102473878-102473878 on Build GRCh38
rs777183787 Chr.2: 102473807-102473807 on Build GRCh38
rs202200517 Chr.2: 102473662-102473663 on Build GRCh38
rs761281045 Chr.2: 102473815-102473815 on Build GRCh38
rs753621408 Chr.2: 102474071-102474071 on Build GRCh38
rs776080068 Chr.2: 102474002-102474002 on Build GRCh38
rs765059235 Chr.2: 102474044-102474044 on Build GRCh38
rs140006023 Chr.2: 102473837-102473837 on Build GRCh38
rs903089709 Chr.2: 102473757-102473757 on Build GRCh38
rs757467470 Chr.2: 102474030-102474030 on Build GRCh38
rs920352354 Chr.2: 102473654-102473654 on Build GRCh38
rs183188258 Chr.2: 102474067-102474067 on Build GRCh38
rs760069585 Chr.2: 102473888-102473888 on Build GRCh38
rs1052529996 Chr.2: 102473743-102473743 on Build GRCh38
rs536499682 Chr.2: 102474013-102474013 on Build GRCh38
rs756379306 Chr.2: 102473787-102473787 on Build GRCh38
rs757310998 Chr.2: 102473767-102473767 on Build GRCh38
rs532697069 Chr.2: 102473702-102473702 on Build GRCh38
rs747814840 Chr.2: 102473875-102473875 on Build GRCh38
rs746464265 Chr.2: 102473991-102473991 on Build GRCh38
rs767585881 Chr.2: 102474029-102474029 on Build GRCh38
rs760594857 Chr.2: 102474049-102474049 on Build GRCh38
rs763436561 Chr.2: 102473810-102473810 on Build GRCh38
rs80292047 Chr.2: 102473830-102473830 on Build GRCh38
rs867465296 Chr.2: 102473853-102473853 on Build GRCh38
rs777980900 Chr.2: 102473754-102473754 on Build GRCh38
rs747039324 Chr.2: 102473758-102473758 on Build GRCh38
rs781114165 Chr.2: 102473774-102473774 on Build GRCh38
rs770573550 Chr.2: 102473999-102473999 on Build GRCh38
rs372579387 Chr.2: 102473971-102473971 on Build GRCh38
rs538381964 Chr.2: 102473921-102473921 on Build GRCh38
rs774012299 Chr.2: 102473793-102473793 on Build GRCh38
rs17027255 Chr.2: 102473668-102473668 on Build GRCh38
rs779801889 Chr.2: 102473849-102473849 on Build GRCh38
rs951371081 Chr.2: 102473820-102473820 on Build GRCh38
rs758738818 Chr.2: 102473747-102473747 on Build GRCh38
rs755313341 Chr.2: 102473730-102473730 on Build GRCh38
rs267598810 Chr.2: 102474001-102474001 on Build GRCh38
rs576360655 Chr.2: 102473659-102473659 on Build GRCh38
rs112273277 Chr.2: 102473708-102473708 on Build GRCh38
rs761322485 Chr.2: 102473795-102473795 on Build GRCh38
rs79378995 Chr.2: 102473809-102473809 on Build GRCh38
rs867854675 Chr.2: 102473650-102473650 on Build GRCh38
rs774615631 Chr.2: 102474020-102474020 on Build GRCh38
rs116706610 Chr.2: 102473946-102473946 on Build GRCh38
rs886950229 Chr.2: 102474076-102474076 on Build GRCh38
rs764564863 Chr.2: 102473898-102473898 on Build GRCh38
rs141912003 Chr.2: 102473975-102473975 on Build GRCh38
rs373684225 Chr.2: 102474040-102474040 on Build GRCh38
rs566055611 Chr.2: 102473661-102473662 on Build GRCh38

View Full Product Details