Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 154855374-154855896 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNJ3
Entrez Gene ID
3760
Gene Name
potassium voltage-gated channel subfamily J member 3
Gene Aliases
GIRK1, KGA, KIR3.1
UniGene
Hs.591606
Gene Chromosome Location
Chr.2: 154698581-154858352 on Build GRCh38
Transcripts
NM_001260508.1 NM_002239.3

SNP Detail

SNP ID Chromosome Location
rs907829663 Chr.2: 154855759-154855770 on Build GRCh38
rs770963822 Chr.2: 154855462-154855462 on Build GRCh38
rs1043835883 Chr.2: 154855778-154855778 on Build GRCh38
rs1006428074 Chr.2: 154855647-154855647 on Build GRCh38
rs953668259 Chr.2: 154855739-154855739 on Build GRCh38
rs969876543 Chr.2: 154855430-154855430 on Build GRCh38
rs552120206 Chr.2: 154855731-154855732 on Build GRCh38
rs1050576202 Chr.2: 154855416-154855416 on Build GRCh38
rs931766915 Chr.2: 154855774-154855774 on Build GRCh38
rs1041811676 Chr.2: 154855457-154855457 on Build GRCh38
rs975948763 Chr.2: 154855765-154855766 on Build GRCh38
rs947581596 Chr.2: 154855784-154855784 on Build GRCh38
rs1043400891 Chr.2: 154855705-154855705 on Build GRCh38
rs535624240 Chr.2: 154855517-154855517 on Build GRCh38
rs759457072 Chr.2: 154855731-154855731 on Build GRCh38
rs557465320 Chr.2: 154855808-154855808 on Build GRCh38
rs896726254 Chr.2: 154855732-154855739 on Build GRCh38
rs924731128 Chr.2: 154855807-154855807 on Build GRCh38
rs903226697 Chr.2: 154855460-154855460 on Build GRCh38
rs1014697354 Chr.2: 154855683-154855683 on Build GRCh38
rs1002893650 Chr.2: 154855487-154855487 on Build GRCh38
rs951333462 Chr.2: 154855747-154855747 on Build GRCh38
rs1012168494 Chr.2: 154855621-154855621 on Build GRCh38
rs776432935 Chr.2: 154855711-154855711 on Build GRCh38
rs1023746267 Chr.2: 154855429-154855429 on Build GRCh38
rs757452826 Chr.2: 154855707-154855708 on Build GRCh38
rs537129410 Chr.2: 154855761-154855761 on Build GRCh38
rs896778652 Chr.2: 154855809-154855809 on Build GRCh38
rs146315383 Chr.2: 154855866-154855867 on Build GRCh38
rs914741772 Chr.2: 154855427-154855427 on Build GRCh38
rs935020287 Chr.2: 154855786-154855787 on Build GRCh38
rs550514122 Chr.2: 154855764-154855765 on Build GRCh38
rs569891713 Chr.2: 154855422-154855422 on Build GRCh38
rs33975324 Chr.2: 154855616-154855617 on Build GRCh38
rs879211839 Chr.2: 154855567-154855567 on Build GRCh38
rs568249551 Chr.2: 154855791-154855791 on Build GRCh38
rs921736208 Chr.2: 154855633-154855638 on Build GRCh38
rs938734777 Chr.2: 154855837-154855837 on Build GRCh38
rs200111474 Chr.2: 154855623-154855623 on Build GRCh38
rs896432589 Chr.2: 154855586-154855586 on Build GRCh38
rs986336475 Chr.2: 154855741-154855741 on Build GRCh38
rs944553050 Chr.2: 154855623-154855627 on Build GRCh38
rs546939089 Chr.2: 154855787-154855790 on Build GRCh38
rs914773001 Chr.2: 154855743-154855743 on Build GRCh38
rs201284261 Chr.2: 154855625-154855625 on Build GRCh38
rs931690616 Chr.2: 154855678-154855678 on Build GRCh38
rs527298420 Chr.2: 154855733-154855733 on Build GRCh38
rs947490175 Chr.2: 154855440-154855440 on Build GRCh38
rs548745467 Chr.2: 154855756-154855756 on Build GRCh38
rs371911954 Chr.2: 154855726-154855726 on Build GRCh38
rs5835552 Chr.2: 154855623-154855624 on Build GRCh38
rs921722294 Chr.2: 154855773-154855782 on Build GRCh38
rs869271535 Chr.2: 154855624-154855625 on Build GRCh38
rs549181236 Chr.2: 154855591-154855591 on Build GRCh38
rs1057125490 Chr.2: 154855505-154855505 on Build GRCh38
rs1042255397 Chr.2: 154855787-154855787 on Build GRCh38

View Full Product Details