Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 201078637-201079092 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NDUFB3
Entrez Gene ID
4709
Gene Name
NADH:ubiquinone oxidoreductase subunit B3
Gene Aliases
B12, CI-B12
UniGene
Hs.109760
Gene Chromosome Location
Chr.2: 201071739-201085750 on Build GRCh38
Transcripts
NM_001257102.1 NM_002491.2 XM_011511230.2 XM_017004186.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4441197 c.122T>C p.L41P Chr.2: 201079004-201079004 on Build GRCh38
4090156 c.140G>A p.R47H Chr.2: 201079022-201079022 on Build GRCh38
1404444 c.31C>T p.H11Y Chr.2: 201078913-201078913 on Build GRCh38
6320903 c.4G>T p.A2S Chr.2: 201078886-201078886 on Build GRCh38
4001465 c.110C>T p.A37V Chr.2: 201078992-201078992 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777036844 Chr.2: 201078966-201078966 on Build GRCh38
rs543676906 Chr.2: 201078685-201078685 on Build GRCh38
rs761452789 Chr.2: 201078855-201078855 on Build GRCh38
rs142609245 Chr.2: 201078946-201078946 on Build GRCh38
rs943010457 Chr.2: 201078712-201078712 on Build GRCh38
rs755829281 Chr.2: 201078896-201078896 on Build GRCh38
rs978701792 Chr.2: 201078886-201078886 on Build GRCh38
rs989884081 Chr.2: 201078702-201078702 on Build GRCh38
rs182420107 Chr.2: 201078916-201078916 on Build GRCh38
rs754809738 Chr.2: 201079049-201079049 on Build GRCh38
rs925474051 Chr.2: 201078891-201078891 on Build GRCh38
rs779174380 Chr.2: 201078914-201078914 on Build GRCh38
rs374448057 Chr.2: 201078730-201078730 on Build GRCh38
rs767542169 Chr.2: 201078890-201078890 on Build GRCh38
rs774198937 Chr.2: 201078997-201078997 on Build GRCh38
rs763947691 Chr.2: 201078963-201078963 on Build GRCh38
rs772796020 Chr.2: 201078720-201078720 on Build GRCh38
rs879088664 Chr.2: 201078876-201078876 on Build GRCh38
rs754835124 Chr.2: 201079021-201079024 on Build GRCh38
rs991799869 Chr.2: 201078801-201078801 on Build GRCh38
rs547582753 Chr.2: 201078767-201078767 on Build GRCh38
rs199607830 Chr.2: 201078878-201078878 on Build GRCh38
rs765393281 Chr.2: 201079058-201079058 on Build GRCh38
rs767171840 Chr.2: 201079019-201079019 on Build GRCh38
rs772960455 Chr.2: 201079021-201079021 on Build GRCh38
rs529243053 Chr.2: 201078742-201078742 on Build GRCh38
rs747554855 Chr.2: 201078953-201078953 on Build GRCh38
rs879077765 Chr.2: 201078877-201078877 on Build GRCh38
rs1047992025 Chr.2: 201078812-201078812 on Build GRCh38
rs755181080 Chr.2: 201078904-201078904 on Build GRCh38
rs761360526 Chr.2: 201079005-201079005 on Build GRCh38
rs11887751 Chr.2: 201078708-201078708 on Build GRCh38
rs11552469 Chr.2: 201078933-201078933 on Build GRCh38
rs772160306 Chr.2: 201078937-201078937 on Build GRCh38
rs376059572 Chr.2: 201079036-201079036 on Build GRCh38
rs368674247 Chr.2: 201078776-201078776 on Build GRCh38
rs375279387 Chr.2: 201078847-201078847 on Build GRCh38
rs972030737 Chr.2: 201079015-201079015 on Build GRCh38
rs768388896 Chr.2: 201078985-201078985 on Build GRCh38
rs144513268 Chr.2: 201078901-201078901 on Build GRCh38
rs373095347 Chr.2: 201079022-201079022 on Build GRCh38
rs751392490 Chr.2: 201078973-201078974 on Build GRCh38
rs752948093 Chr.2: 201079063-201079063 on Build GRCh38
rs1038729119 Chr.2: 201078762-201078762 on Build GRCh38
rs933271379 Chr.2: 201078782-201078782 on Build GRCh38
rs750401775 Chr.2: 201078894-201078894 on Build GRCh38
rs376733237 Chr.2: 201078838-201078838 on Build GRCh38
rs537946121 Chr.2: 201078810-201078810 on Build GRCh38
rs370768077 Chr.2: 201079035-201079035 on Build GRCh38
rs1022967616 Chr.2: 201078996-201078996 on Build GRCh38
rs778982161 Chr.2: 201078902-201078902 on Build GRCh38
rs763894617 Chr.2: 201078846-201078846 on Build GRCh38
rs781198264 Chr.2: 201079043-201079046 on Build GRCh38
rs779904920 Chr.2: 201078897-201078897 on Build GRCh38
rs369669842 Chr.2: 201078964-201078964 on Build GRCh38
rs540436295 Chr.2: 201078680-201078680 on Build GRCh38
rs568058126 Chr.2: 201078773-201078773 on Build GRCh38
rs538153415 Chr.2: 201078978-201078978 on Build GRCh38

View Full Product Details