Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 201195652-201196159 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASP10
Entrez Gene ID
843
Gene Name
caspase 10
Gene Aliases
ALPS2, FLICE2, MCH4
UniGene
Hs.5353
Gene Chromosome Location
Chr.2: 201182881-201229406 on Build GRCh38
Transcripts
NM_001206524.1 NM_001206542.1 NM_001230.4 NM_001306083.1 NM_032974.4 NM_032976.3 NM_032977.3 XM_005246907.2 XM_006712796.3 XM_011511990.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4822036 c.625C>G p.Q209E Chr.2: 201195889-201195889 on Build GRCh38
1318120 c.593C>A p.T198K Chr.2: 201195857-201195857 on Build GRCh38
3909326 c.599C>T p.P200L Chr.2: 201195863-201195863 on Build GRCh38
719803 c.620C>T p.S207L Chr.2: 201195884-201195884 on Build GRCh38
4822037 c.625C>G p.Q209E Chr.2: 201195889-201195889 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747896320 Chr.2: 201195792-201195792 on Build GRCh38
rs767731671 Chr.2: 201195850-201195851 on Build GRCh38
rs765626187 Chr.2: 201195840-201195840 on Build GRCh38
rs374339540 Chr.2: 201195952-201195952 on Build GRCh38
rs562571641 Chr.2: 201196081-201196081 on Build GRCh38
rs774349941 Chr.2: 201195791-201195791 on Build GRCh38
rs1030623554 Chr.2: 201195680-201195680 on Build GRCh38
rs1052572966 Chr.2: 201196101-201196101 on Build GRCh38
rs576476434 Chr.2: 201196047-201196047 on Build GRCh38
rs776556990 Chr.2: 201195825-201195825 on Build GRCh38
rs1046681189 Chr.2: 201196058-201196058 on Build GRCh38
rs3731713 Chr.2: 201196103-201196103 on Build GRCh38
rs552345754 Chr.2: 201195731-201195731 on Build GRCh38
rs374490783 Chr.2: 201195802-201195802 on Build GRCh38
rs935506949 Chr.2: 201195971-201195971 on Build GRCh38
rs769737385 Chr.2: 201195723-201195723 on Build GRCh38
rs752039240 Chr.2: 201195862-201195862 on Build GRCh38
rs779770348 Chr.2: 201195867-201195867 on Build GRCh38
rs754547719 Chr.2: 201195871-201195871 on Build GRCh38
rs930975061 Chr.2: 201195682-201195682 on Build GRCh38
rs543872927 Chr.2: 201196075-201196075 on Build GRCh38
rs368697916 Chr.2: 201195794-201195794 on Build GRCh38
rs773188973 Chr.2: 201195764-201195764 on Build GRCh38
rs373077581 Chr.2: 201195879-201195879 on Build GRCh38
rs972443043 Chr.2: 201195880-201195880 on Build GRCh38
rs931989680 Chr.2: 201196009-201196009 on Build GRCh38
rs770997816 Chr.2: 201195884-201195884 on Build GRCh38
rs748679960 Chr.2: 201195870-201195870 on Build GRCh38
rs753282118 Chr.2: 201195841-201195841 on Build GRCh38
rs764114274 Chr.2: 201196079-201196079 on Build GRCh38
rs982619364 Chr.2: 201196043-201196043 on Build GRCh38
rs763551197 Chr.2: 201195928-201195928 on Build GRCh38
rs41363644 Chr.2: 201195678-201195678 on Build GRCh38
rs780903057 Chr.2: 201195987-201195987 on Build GRCh38
rs566510495 Chr.2: 201195701-201195701 on Build GRCh38
rs759488307 Chr.2: 201195909-201195909 on Build GRCh38
rs895371405 Chr.2: 201195889-201195889 on Build GRCh38
rs759026116 Chr.2: 201195852-201195852 on Build GRCh38
rs3731714 Chr.2: 201196097-201196097 on Build GRCh38
rs769898861 Chr.2: 201195911-201195911 on Build GRCh38
rs746876281 Chr.2: 201195882-201195882 on Build GRCh38
rs964010169 Chr.2: 201195719-201195719 on Build GRCh38
rs949571095 Chr.2: 201195702-201195702 on Build GRCh38
rs41354945 Chr.2: 201196035-201196035 on Build GRCh38
rs759671327 Chr.2: 201195839-201195839 on Build GRCh38
rs1008533963 Chr.2: 201196039-201196039 on Build GRCh38
rs200249066 Chr.2: 201195688-201195688 on Build GRCh38
rs755710677 Chr.2: 201195865-201195865 on Build GRCh38
rs948303477 Chr.2: 201196055-201196055 on Build GRCh38
rs775755140 Chr.2: 201195913-201195913 on Build GRCh38
rs942106933 Chr.2: 201196044-201196044 on Build GRCh38
rs902586245 Chr.2: 201195713-201195713 on Build GRCh38
rs970429964 Chr.2: 201196106-201196106 on Build GRCh38
rs570455304 Chr.2: 201195756-201195756 on Build GRCh38
rs1010638184 Chr.2: 201196122-201196122 on Build GRCh38
rs368707977 Chr.2: 201195763-201195763 on Build GRCh38
rs757588373 Chr.2: 201196015-201196015 on Build GRCh38
rs1053809516 Chr.2: 201195724-201195724 on Build GRCh38
rs370134219 Chr.2: 201195946-201195946 on Build GRCh38
rs772487302 Chr.2: 201195968-201195968 on Build GRCh38
rs770785873 Chr.2: 201195986-201195986 on Build GRCh38
rs893771452 Chr.2: 201195755-201195755 on Build GRCh38
rs545796839 Chr.2: 201196018-201196018 on Build GRCh38
rs574563011 Chr.2: 201196086-201196086 on Build GRCh38
rs1019844008 Chr.2: 201195797-201195797 on Build GRCh38
rs113038818 Chr.2: 201195985-201195985 on Build GRCh38
rs982734736 Chr.2: 201196114-201196114 on Build GRCh38
rs766687931 Chr.2: 201195822-201195822 on Build GRCh38
rs188421807 Chr.2: 201195790-201195790 on Build GRCh38
rs999539772 Chr.2: 201195717-201195717 on Build GRCh38
rs369596070 Chr.2: 201195977-201195977 on Build GRCh38
rs370692630 Chr.2: 201196016-201196017 on Build GRCh38
rs1047028238 Chr.2: 201195707-201195707 on Build GRCh38
rs116125703 Chr.2: 201195930-201195930 on Build GRCh38
rs200079223 Chr.2: 201195860-201195860 on Build GRCh38
rs1055342119 Chr.2: 201195691-201195691 on Build GRCh38
rs957736874 Chr.2: 201195711-201195711 on Build GRCh38
rs759715336 Chr.2: 201195819-201195822 on Build GRCh38
rs148307772 Chr.2: 201195676-201195676 on Build GRCh38
rs753005511 Chr.2: 201195881-201195881 on Build GRCh38
rs527266140 Chr.2: 201195727-201195727 on Build GRCh38
rs527532940 Chr.2: 201196107-201196107 on Build GRCh38
rs906788012 Chr.2: 201196083-201196083 on Build GRCh38
rs760890208 Chr.2: 201196059-201196059 on Build GRCh38
rs951279914 Chr.2: 201196042-201196042 on Build GRCh38
rs778773099 Chr.2: 201195874-201195874 on Build GRCh38
rs776912309 Chr.2: 201195885-201195885 on Build GRCh38
rs893562126 Chr.2: 201196001-201196001 on Build GRCh38
rs746074361 Chr.2: 201195995-201195995 on Build GRCh38
rs961213763 Chr.2: 201195864-201195864 on Build GRCh38
rs773037513 Chr.2: 201195809-201195809 on Build GRCh38
rs976718383 Chr.2: 201196032-201196035 on Build GRCh38
rs372539994 Chr.2: 201195949-201195949 on Build GRCh38
rs143882052 Chr.2: 201195947-201195947 on Build GRCh38
rs973388751 Chr.2: 201196046-201196046 on Build GRCh38
rs368583939 Chr.2: 201195948-201195948 on Build GRCh38
rs537903314 Chr.2: 201195782-201195782 on Build GRCh38
rs778362369 Chr.2: 201195963-201195963 on Build GRCh38
rs111519154 Chr.2: 201195817-201195817 on Build GRCh38
rs952354110 Chr.2: 201195887-201195887 on Build GRCh38
rs994040266 Chr.2: 201196066-201196066 on Build GRCh38

View Full Product Details