Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 201233351-201233847 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASP8
Entrez Gene ID
841
Gene Name
caspase 8
Gene Aliases
ALPS2B, CAP4, Casp-8, FLICE, MACH, MCH5
UniGene
Hs.599762
Gene Chromosome Location
Chr.2: 201233443-201287711 on Build GRCh38
Transcripts
NM_001080124.1 NM_001228.4 NM_033355.3 NM_033358.3 XM_005246887.1 XM_005246888.1 XM_005246894.3 XM_006712789.1 XM_006712793.2

SNP Detail

SNP ID Chromosome Location
rs1020567294 Chr.2: 201233563-201233563 on Build GRCh38
rs951749082 Chr.2: 201233380-201233380 on Build GRCh38
rs777229663 Chr.2: 201233417-201233417 on Build GRCh38
rs887245363 Chr.2: 201233693-201233693 on Build GRCh38
rs922873471 Chr.2: 201233643-201233643 on Build GRCh38
rs13020440 Chr.2: 201233605-201233605 on Build GRCh38
rs905320753 Chr.2: 201233783-201233783 on Build GRCh38
rs531418615 Chr.2: 201233438-201233438 on Build GRCh38
rs529000486 Chr.2: 201233776-201233776 on Build GRCh38
rs749269188 Chr.2: 201233738-201233738 on Build GRCh38
rs17860416 Chr.2: 201233408-201233408 on Build GRCh38
rs967219330 Chr.2: 201233590-201233590 on Build GRCh38
rs942307771 Chr.2: 201233618-201233618 on Build GRCh38
rs1041834688 Chr.2: 201233442-201233442 on Build GRCh38
rs527497064 Chr.2: 201233436-201233436 on Build GRCh38
rs936028219 Chr.2: 201233482-201233482 on Build GRCh38
rs975517055 Chr.2: 201233627-201233627 on Build GRCh38
rs978485171 Chr.2: 201233593-201233593 on Build GRCh38
rs889339159 Chr.2: 201233621-201233621 on Build GRCh38
rs934258526 Chr.2: 201233651-201233651 on Build GRCh38
rs552218203 Chr.2: 201233396-201233396 on Build GRCh38
rs1019511020 Chr.2: 201233644-201233644 on Build GRCh38
rs547358989 Chr.2: 201233813-201233813 on Build GRCh38
rs941549505 Chr.2: 201233731-201233731 on Build GRCh38
rs1002157939 Chr.2: 201233814-201233814 on Build GRCh38
rs34609836 Chr.2: 201233603-201233603 on Build GRCh38
rs566870779 Chr.2: 201233626-201233626 on Build GRCh38
rs866690755 Chr.2: 201233557-201233557 on Build GRCh38
rs904519772 Chr.2: 201233446-201233446 on Build GRCh38
rs960268727 Chr.2: 201233499-201233499 on Build GRCh38
rs926501611 Chr.2: 201233386-201233386 on Build GRCh38
rs992657418 Chr.2: 201233524-201233524 on Build GRCh38
rs1009169798 Chr.2: 201233630-201233631 on Build GRCh38
rs539844777 Chr.2: 201233682-201233682 on Build GRCh38
rs980369474 Chr.2: 201233749-201233749 on Build GRCh38
rs773653309 Chr.2: 201233724-201233724 on Build GRCh38
rs1005620378 Chr.2: 201233381-201233381 on Build GRCh38
rs951799652 Chr.2: 201233736-201233736 on Build GRCh38
rs969335828 Chr.2: 201233402-201233402 on Build GRCh38
rs1047214552 Chr.2: 201233667-201233667 on Build GRCh38

View Full Product Details