Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 203434499-203435018 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAPH1
Entrez Gene ID
65059
Gene Name
Ras association (RalGDS/AF-6) and pleckstrin homology domains 1
Gene Aliases
ALS2CR18, ALS2CR9, LPD, PREL-2, PREL2, RMO1, RalGDS/AF-6
UniGene
Hs.471162
Gene Chromosome Location
Chr.2: 203433682-203535335 on Build GRCh38
Transcripts
NM_213589.2 XM_005246777.3 XM_005246779.4 XM_006712695.3 XM_011511646.2 XM_011511647.1 XM_011511648.1

SNP Detail

SNP ID Chromosome Location
rs757615866 Chr.2: 203434567-203434567 on Build GRCh38
rs984256485 Chr.2: 203434625-203434625 on Build GRCh38
rs558492016 Chr.2: 203434719-203434719 on Build GRCh38
rs73060643 Chr.2: 203434611-203434611 on Build GRCh38
rs536549532 Chr.2: 203434650-203434650 on Build GRCh38
rs954395491 Chr.2: 203434697-203434697 on Build GRCh38
rs974603775 Chr.2: 203434553-203434553 on Build GRCh38
rs927986807 Chr.2: 203434880-203434880 on Build GRCh38
rs1045436830 Chr.2: 203434548-203434548 on Build GRCh38
rs984594606 Chr.2: 203434831-203434831 on Build GRCh38
rs754474420 Chr.2: 203434547-203434548 on Build GRCh38
rs376158317 Chr.2: 203434786-203434786 on Build GRCh38
rs922777041 Chr.2: 203434969-203434969 on Build GRCh38
rs1010051631 Chr.2: 203434585-203434587 on Build GRCh38
rs1050014403 Chr.2: 203434679-203434679 on Build GRCh38
rs376436125 Chr.2: 203434835-203434836 on Build GRCh38
rs998670451 Chr.2: 203434745-203434745 on Build GRCh38
rs77589546 Chr.2: 203434784-203434784 on Build GRCh38
rs1004073222 Chr.2: 203434581-203434583 on Build GRCh38
rs1012848328 Chr.2: 203434809-203434809 on Build GRCh38
rs879072049 Chr.2: 203434566-203434567 on Build GRCh38
rs908656398 Chr.2: 203434851-203434851 on Build GRCh38
rs976866378 Chr.2: 203434928-203434928 on Build GRCh38
rs7602076 Chr.2: 203434817-203434817 on Build GRCh38
rs546894098 Chr.2: 203434532-203434532 on Build GRCh38
rs915918562 Chr.2: 203434583-203434583 on Build GRCh38
rs769256811 Chr.2: 203434890-203434890 on Build GRCh38
rs1021567182 Chr.2: 203434597-203434599 on Build GRCh38
rs932925294 Chr.2: 203434656-203434656 on Build GRCh38
rs369267717 Chr.2: 203434567-203434568 on Build GRCh38
rs72926408 Chr.2: 203434717-203434717 on Build GRCh38
rs1041683320 Chr.2: 203434654-203434654 on Build GRCh38
rs930441954 Chr.2: 203434591-203434591 on Build GRCh38
rs542899128 Chr.2: 203434823-203434823 on Build GRCh38
rs199536675 Chr.2: 203434566-203434566 on Build GRCh38
rs961774725 Chr.2: 203434881-203434881 on Build GRCh38
rs964618705 Chr.2: 203434536-203434536 on Build GRCh38
rs538361716 Chr.2: 203434590-203434590 on Build GRCh38
rs906618802 Chr.2: 203434579-203434579 on Build GRCh38
rs1031075500 Chr.2: 203434582-203434583 on Build GRCh38
rs563762948 Chr.2: 203434631-203434631 on Build GRCh38
rs181994271 Chr.2: 203434780-203434780 on Build GRCh38
rs777389281 Chr.2: 203434657-203434658 on Build GRCh38
rs1042833612 Chr.2: 203434718-203434718 on Build GRCh38
rs922830768 Chr.2: 203434655-203434655 on Build GRCh38
rs891650683 Chr.2: 203434583-203434583 on Build GRCh38
rs554928727 Chr.2: 203434651-203434651 on Build GRCh38
rs573057771 Chr.2: 203434711-203434711 on Build GRCh38
rs774383329 Chr.2: 203434646-203434646 on Build GRCh38
rs888813896 Chr.2: 203434712-203434712 on Build GRCh38
rs903002546 Chr.2: 203434731-203434731 on Build GRCh38
rs2250800 Chr.2: 203434533-203434533 on Build GRCh38
rs773797051 Chr.2: 203434568-203434569 on Build GRCh38
rs959904564 Chr.2: 203434804-203434804 on Build GRCh38

View Full Product Details