Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 208427750-208428293 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTH2R
Entrez Gene ID
5746
Gene Name
parathyroid hormone 2 receptor
Gene Aliases
PTHR2
UniGene
Hs.570296
Gene Chromosome Location
Chr.2: 208359845-208494506 on Build GRCh38
Transcripts
NM_001309516.1 NM_005048.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1015677 c.110A>G p.Q37R Chr.2: 208428235-208428235 on Build GRCh38
4309234 c.132G>A p.A44A Chr.2: 208428257-208428257 on Build GRCh38
7331231 c.131C>A p.A44E Chr.2: 208428256-208428256 on Build GRCh38
1222476 c.79G>A p.D27N Chr.2: 208428204-208428204 on Build GRCh38
2714842 c.154A>T p.I52F Chr.2: 208428279-208428279 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs972698751 Chr.2: 208428048-208428048 on Build GRCh38
rs199785290 Chr.2: 208428192-208428192 on Build GRCh38
rs910610609 Chr.2: 208427860-208427860 on Build GRCh38
rs1053400973 Chr.2: 208428166-208428166 on Build GRCh38
rs570044268 Chr.2: 208428217-208428217 on Build GRCh38
rs191121150 Chr.2: 208428012-208428012 on Build GRCh38
rs965429813 Chr.2: 208428080-208428080 on Build GRCh38
rs1051828338 Chr.2: 208427792-208427792 on Build GRCh38
rs978806197 Chr.2: 208428100-208428100 on Build GRCh38
rs747625116 Chr.2: 208428157-208428158 on Build GRCh38
rs766921997 Chr.2: 208428211-208428211 on Build GRCh38
rs769437862 Chr.2: 208428192-208428193 on Build GRCh38
rs189305207 Chr.2: 208428266-208428266 on Build GRCh38
rs905519680 Chr.2: 208428239-208428239 on Build GRCh38
rs183627553 Chr.2: 208428256-208428256 on Build GRCh38
rs1053774399 Chr.2: 208427965-208427965 on Build GRCh38
rs968554073 Chr.2: 208428176-208428176 on Build GRCh38
rs1005346226 Chr.2: 208428240-208428240 on Build GRCh38
rs934804056 Chr.2: 208428151-208428151 on Build GRCh38
rs1045640388 Chr.2: 208427998-208427998 on Build GRCh38
rs1043168875 Chr.2: 208427902-208427902 on Build GRCh38
rs764574272 Chr.2: 208428089-208428089 on Build GRCh38
rs905470872 Chr.2: 208427999-208427999 on Build GRCh38
rs978480938 Chr.2: 208427928-208427928 on Build GRCh38
rs768346230 Chr.2: 208428179-208428179 on Build GRCh38
rs924286231 Chr.2: 208427941-208427941 on Build GRCh38
rs533342599 Chr.2: 208428126-208428126 on Build GRCh38
rs767388910 Chr.2: 208428232-208428232 on Build GRCh38
rs934961730 Chr.2: 208427939-208427939 on Build GRCh38
rs1048613999 Chr.2: 208428010-208428010 on Build GRCh38
rs774886297 Chr.2: 208428197-208428197 on Build GRCh38
rs910640363 Chr.2: 208428075-208428075 on Build GRCh38
rs987411402 Chr.2: 208428070-208428070 on Build GRCh38
rs145269926 Chr.2: 208428225-208428225 on Build GRCh38
rs368989589 Chr.2: 208427772-208427772 on Build GRCh38
rs1029983059 Chr.2: 208428059-208428059 on Build GRCh38
rs780203101 Chr.2: 208428243-208428243 on Build GRCh38
rs895504640 Chr.2: 208427974-208427974 on Build GRCh38
rs949566943 Chr.2: 208427992-208427993 on Build GRCh38
rs1019617698 Chr.2: 208427811-208427811 on Build GRCh38
rs1044045507 Chr.2: 208428219-208428219 on Build GRCh38
rs949716533 Chr.2: 208428193-208428193 on Build GRCh38
rs368123492 Chr.2: 208428170-208428170 on Build GRCh38
rs926180010 Chr.2: 208428102-208428102 on Build GRCh38
rs943560484 Chr.2: 208427896-208427896 on Build GRCh38
rs1016802445 Chr.2: 208428017-208428017 on Build GRCh38
rs535954982 Chr.2: 208428222-208428222 on Build GRCh38
rs996714729 Chr.2: 208428104-208428104 on Build GRCh38
rs904779472 Chr.2: 208427911-208427911 on Build GRCh38
rs954748888 Chr.2: 208428063-208428063 on Build GRCh38
rs934291149 Chr.2: 208427957-208427957 on Build GRCh38
rs866997990 Chr.2: 208428195-208428195 on Build GRCh38
rs773546849 Chr.2: 208428267-208428267 on Build GRCh38
rs200331498 Chr.2: 208427918-208427918 on Build GRCh38
rs961501619 Chr.2: 208428046-208428046 on Build GRCh38
rs965192143 Chr.2: 208427859-208427859 on Build GRCh38
rs752883890 Chr.2: 208428224-208428224 on Build GRCh38
rs905586527 Chr.2: 208428004-208428004 on Build GRCh38
rs1004135123 Chr.2: 208428045-208428045 on Build GRCh38
rs750291322 Chr.2: 208428238-208428238 on Build GRCh38
rs748862155 Chr.2: 208428171-208428171 on Build GRCh38
rs969022203 Chr.2: 208427918-208427918 on Build GRCh38
rs1056906435 Chr.2: 208427962-208427962 on Build GRCh38
rs769181476 Chr.2: 208427987-208427987 on Build GRCh38
rs921230241 Chr.2: 208427917-208427918 on Build GRCh38
rs943421531 Chr.2: 208428076-208428076 on Build GRCh38
rs376844741 Chr.2: 208428180-208428180 on Build GRCh38
rs776679308 Chr.2: 208428213-208428213 on Build GRCh38
rs555754809 Chr.2: 208428257-208428257 on Build GRCh38
rs1028562915 Chr.2: 208428139-208428139 on Build GRCh38
rs1013914427 Chr.2: 208427985-208427985 on Build GRCh38
rs756103453 Chr.2: 208428242-208428242 on Build GRCh38
rs747790135 Chr.2: 208428258-208428258 on Build GRCh38
rs772348775 Chr.2: 208428187-208428187 on Build GRCh38

View Full Product Details