Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 229477055-229477556 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNER
Entrez Gene ID
92737
Gene Name
delta/notch like EGF repeat containing
Gene Aliases
UNQ26, bet
UniGene
Hs.234074
Gene Chromosome Location
Chr.2: 229357629-229714570 on Build GRCh38
Transcripts
NM_139072.3 XM_005246950.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4957615 c.1221C>T p.C407C Chr.2: 229477180-229477180 on Build GRCh38
6090653 c.1246T>C p.C416R Chr.2: 229477155-229477155 on Build GRCh38
1732313 c.1247G>T p.C416F Chr.2: 229477154-229477154 on Build GRCh38
4001528 c.1243A>C p.T415P Chr.2: 229477158-229477158 on Build GRCh38
325190 c.1229G>T p.S410I Chr.2: 229477172-229477172 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760282334 Chr.2: 229477240-229477240 on Build GRCh38
rs1052311271 Chr.2: 229477380-229477380 on Build GRCh38
rs1032885320 Chr.2: 229477514-229477514 on Build GRCh38
rs987161014 Chr.2: 229477365-229477365 on Build GRCh38
rs779813972 Chr.2: 229477089-229477089 on Build GRCh38
rs201049418 Chr.2: 229477269-229477269 on Build GRCh38
rs902628052 Chr.2: 229477480-229477480 on Build GRCh38
rs763372650 Chr.2: 229477205-229477205 on Build GRCh38
rs780008373 Chr.2: 229477160-229477160 on Build GRCh38
rs566995350 Chr.2: 229477469-229477469 on Build GRCh38
rs57647946 Chr.2: 229477099-229477099 on Build GRCh38
rs778691728 Chr.2: 229477255-229477255 on Build GRCh38
rs763793731 Chr.2: 229477136-229477136 on Build GRCh38
rs772252322 Chr.2: 229477101-229477101 on Build GRCh38
rs776166155 Chr.2: 229477204-229477204 on Build GRCh38
rs745890355 Chr.2: 229477264-229477264 on Build GRCh38
rs752591955 Chr.2: 229477256-229477256 on Build GRCh38
rs1055721789 Chr.2: 229477430-229477430 on Build GRCh38
rs754787592 Chr.2: 229477163-229477163 on Build GRCh38
rs552813676 Chr.2: 229477503-229477503 on Build GRCh38
rs896344656 Chr.2: 229477412-229477412 on Build GRCh38
rs770982005 Chr.2: 229477173-229477173 on Build GRCh38
rs775988472 Chr.2: 229477132-229477132 on Build GRCh38
rs747062538 Chr.2: 229477106-229477106 on Build GRCh38
rs765774752 Chr.2: 229477224-229477224 on Build GRCh38
rs766728039 Chr.2: 229477322-229477322 on Build GRCh38
rs903491910 Chr.2: 229477327-229477327 on Build GRCh38
rs746508652 Chr.2: 229477124-229477127 on Build GRCh38
rs530144459 Chr.2: 229477459-229477459 on Build GRCh38
rs756003503 Chr.2: 229477159-229477159 on Build GRCh38
rs940516513 Chr.2: 229477379-229477379 on Build GRCh38
rs778018791 Chr.2: 229477104-229477104 on Build GRCh38
rs780645074 Chr.2: 229477273-229477273 on Build GRCh38
rs777126597 Chr.2: 229477110-229477110 on Build GRCh38
rs770389952 Chr.2: 229477193-229477193 on Build GRCh38
rs190600721 Chr.2: 229477307-229477307 on Build GRCh38
rs550093086 Chr.2: 229477350-229477350 on Build GRCh38
rs781115461 Chr.2: 229477179-229477179 on Build GRCh38
rs774122463 Chr.2: 229477263-229477264 on Build GRCh38
rs1032581842 Chr.2: 229477285-229477285 on Build GRCh38
rs769345026 Chr.2: 229477287-229477287 on Build GRCh38
rs538054123 Chr.2: 229477508-229477508 on Build GRCh38
rs770944920 Chr.2: 229477109-229477109 on Build GRCh38
rs907656088 Chr.2: 229477367-229477367 on Build GRCh38
rs761324481 Chr.2: 229477146-229477146 on Build GRCh38
rs757414748 Chr.2: 229477257-229477257 on Build GRCh38
rs146727100 Chr.2: 229477264-229477264 on Build GRCh38
rs769150783 Chr.2: 229477212-229477212 on Build GRCh38
rs1043397991 Chr.2: 229477310-229477310 on Build GRCh38
rs749871660 Chr.2: 229477239-229477239 on Build GRCh38
rs895108279 Chr.2: 229477442-229477442 on Build GRCh38
rs559830886 Chr.2: 229477250-229477250 on Build GRCh38
rs1034134164 Chr.2: 229477130-229477130 on Build GRCh38
rs766950356 Chr.2: 229477157-229477157 on Build GRCh38
rs747200067 Chr.2: 229477170-229477170 on Build GRCh38
rs200817648 Chr.2: 229477158-229477158 on Build GRCh38
rs1013418543 Chr.2: 229477458-229477458 on Build GRCh38
rs937215269 Chr.2: 229477407-229477407 on Build GRCh38
rs746025069 Chr.2: 229477187-229477187 on Build GRCh38
rs766255027 Chr.2: 229477248-229477248 on Build GRCh38
rs372714221 Chr.2: 229477113-229477113 on Build GRCh38
rs781514239 Chr.2: 229477260-229477260 on Build GRCh38
rs774000987 Chr.2: 229477221-229477221 on Build GRCh38
rs921561362 Chr.2: 229477346-229477346 on Build GRCh38
rs796687966 Chr.2: 229477263-229477264 on Build GRCh38
rs1024164173 Chr.2: 229477479-229477479 on Build GRCh38
rs774574999 Chr.2: 229477292-229477292 on Build GRCh38
rs777932424 Chr.2: 229477164-229477164 on Build GRCh38
rs1013609759 Chr.2: 229477446-229477446 on Build GRCh38
rs548256547 Chr.2: 229477464-229477464 on Build GRCh38
rs545596600 Chr.2: 229477279-229477279 on Build GRCh38
rs770386996 Chr.2: 229477122-229477122 on Build GRCh38
rs148375745 Chr.2: 229477504-229477504 on Build GRCh38
rs141846553 Chr.2: 229477139-229477139 on Build GRCh38
rs376092444 Chr.2: 229477249-229477249 on Build GRCh38
rs113744605 Chr.2: 229477176-229477176 on Build GRCh38
rs920207704 Chr.2: 229477141-229477141 on Build GRCh38
rs186538591 Chr.2: 229477306-229477306 on Build GRCh38
rs975026272 Chr.2: 229477313-229477313 on Build GRCh38
rs899077804 Chr.2: 229477382-229477382 on Build GRCh38
rs561811545 Chr.2: 229477387-229477387 on Build GRCh38
rs753607374 Chr.2: 229477162-229477162 on Build GRCh38
rs113052454 Chr.2: 229477496-229477496 on Build GRCh38

View Full Product Details