Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 232482999-232483509 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ECEL1
Entrez Gene ID
9427
Gene Name
endothelin converting enzyme like 1
Gene Aliases
DA5D, DINE, ECEX, XCE
UniGene
Hs.26880
Gene Chromosome Location
Chr.2: 232479827-232487859 on Build GRCh38
Transcripts
NM_001290787.1 NM_004826.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4444770 c.1499G>A p.R500Q Chr.2: 232483423-232483423 on Build GRCh38
6450228 c.1455G>T p.L485L Chr.2: 232483467-232483467 on Build GRCh38
1752435 c.1498C>T p.R500W Chr.2: 232483424-232483424 on Build GRCh38
442482 c.1579G>C p.E527Q Chr.2: 232483107-232483107 on Build GRCh38
4092395 c.1558G>A p.D520N Chr.2: 232483128-232483128 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1020391791 Chr.2: 232483269-232483269 on Build GRCh38
rs771755274 Chr.2: 232483085-232483085 on Build GRCh38
rs778658998 Chr.2: 232483481-232483481 on Build GRCh38
rs1039557294 Chr.2: 232483233-232483233 on Build GRCh38
rs374660032 Chr.2: 232483398-232483398 on Build GRCh38
rs199916648 Chr.2: 232483147-232483147 on Build GRCh38
rs374850133 Chr.2: 232483175-232483175 on Build GRCh38
rs771842952 Chr.2: 232483150-232483150 on Build GRCh38
rs951069257 Chr.2: 232483321-232483321 on Build GRCh38
rs749652211 Chr.2: 232483076-232483076 on Build GRCh38
rs965690672 Chr.2: 232483352-232483352 on Build GRCh38
rs983624433 Chr.2: 232483326-232483326 on Build GRCh38
rs34019519 Chr.2: 232483446-232483446 on Build GRCh38
rs139763270 Chr.2: 232483296-232483296 on Build GRCh38
rs542130042 Chr.2: 232483469-232483469 on Build GRCh38
rs778563534 Chr.2: 232483083-232483083 on Build GRCh38
rs753116237 Chr.2: 232483430-232483430 on Build GRCh38
rs6750085 Chr.2: 232483209-232483209 on Build GRCh38
rs144766400 Chr.2: 232483374-232483374 on Build GRCh38
rs753094153 Chr.2: 232483222-232483222 on Build GRCh38
rs767625283 Chr.2: 232483128-232483128 on Build GRCh38
rs776590182 Chr.2: 232483174-232483174 on Build GRCh38
rs768207898 Chr.2: 232483081-232483081 on Build GRCh38
rs550310740 Chr.2: 232483293-232483293 on Build GRCh38
rs1043534584 Chr.2: 232483384-232483384 on Build GRCh38
rs72482107 Chr.2: 232483410-232483410 on Build GRCh38
rs375349961 Chr.2: 232483217-232483217 on Build GRCh38
rs763859873 Chr.2: 232483445-232483445 on Build GRCh38
rs191574294 Chr.2: 232483404-232483404 on Build GRCh38
rs146818286 Chr.2: 232483443-232483443 on Build GRCh38
rs149459910 Chr.2: 232483452-232483452 on Build GRCh38
rs902560306 Chr.2: 232483229-232483229 on Build GRCh38
rs776567024 Chr.2: 232483417-232483417 on Build GRCh38
rs962969191 Chr.2: 232483299-232483299 on Build GRCh38
rs202085552 Chr.2: 232483210-232483210 on Build GRCh38
rs755331817 Chr.2: 232483145-232483145 on Build GRCh38
rs371229056 Chr.2: 232483391-232483391 on Build GRCh38
rs369526951 Chr.2: 232483140-232483140 on Build GRCh38
rs560560183 Chr.2: 232483233-232483234 on Build GRCh38
rs745860080 Chr.2: 232483396-232483396 on Build GRCh38
rs907334906 Chr.2: 232483278-232483278 on Build GRCh38
rs13002919 Chr.2: 232483056-232483056 on Build GRCh38
rs151119841 Chr.2: 232483423-232483423 on Build GRCh38
rs756676913 Chr.2: 232483132-232483132 on Build GRCh38
rs372045195 Chr.2: 232483220-232483220 on Build GRCh38
rs965140167 Chr.2: 232483357-232483357 on Build GRCh38
rs765601136 Chr.2: 232483424-232483424 on Build GRCh38
rs377348971 Chr.2: 232483202-232483202 on Build GRCh38
rs576879477 Chr.2: 232483214-232483214 on Build GRCh38
rs527721517 Chr.2: 232483258-232483258 on Build GRCh38
rs550130776 Chr.2: 232483119-232483119 on Build GRCh38
rs200937309 Chr.2: 232483218-232483218 on Build GRCh38
rs376977206 Chr.2: 232483084-232483084 on Build GRCh38
rs765888990 Chr.2: 232483383-232483383 on Build GRCh38
rs562041508 Chr.2: 232483244-232483244 on Build GRCh38
rs1026631698 Chr.2: 232483306-232483306 on Build GRCh38
rs757484998 Chr.2: 232483045-232483045 on Build GRCh38
rs1054451095 Chr.2: 232483262-232483262 on Build GRCh38
rs760757121 Chr.2: 232483092-232483092 on Build GRCh38
rs373090278 Chr.2: 232483369-232483369 on Build GRCh38
rs533271815 Chr.2: 232483286-232483286 on Build GRCh38
rs759307695 Chr.2: 232483100-232483100 on Build GRCh38
rs893151787 Chr.2: 232483263-232483263 on Build GRCh38
rs576364780 Chr.2: 232483226-232483226 on Build GRCh38
rs754160637 Chr.2: 232483144-232483144 on Build GRCh38
rs113658806 Chr.2: 232483068-232483068 on Build GRCh38
rs1010303742 Chr.2: 232483268-232483268 on Build GRCh38
rs747871273 Chr.2: 232483148-232483148 on Build GRCh38
rs745357522 Chr.2: 232483454-232483454 on Build GRCh38
rs182355168 Chr.2: 232483156-232483156 on Build GRCh38
rs542185300 Chr.2: 232483239-232483239 on Build GRCh38
rs750498508 Chr.2: 232483129-232483129 on Build GRCh38
rs775413139 Chr.2: 232483401-232483401 on Build GRCh38
rs374002082 Chr.2: 232483071-232483071 on Build GRCh38
rs745889044 Chr.2: 232483285-232483285 on Build GRCh38
rs1045044950 Chr.2: 232483427-232483427 on Build GRCh38
rs797045548 Chr.2: 232483188-232483188 on Build GRCh38
rs756091444 Chr.2: 232483467-232483467 on Build GRCh38
rs1000027670 Chr.2: 232483232-232483232 on Build GRCh38
rs772816522 Chr.2: 232483091-232483091 on Build GRCh38
rs142971707 Chr.2: 232483170-232483170 on Build GRCh38
rs374586939 Chr.2: 232483028-232483028 on Build GRCh38
rs368556568 Chr.2: 232483239-232483240 on Build GRCh38
rs776303095 Chr.2: 232483099-232483099 on Build GRCh38
rs541083158 Chr.2: 232483275-232483275 on Build GRCh38
rs746461050 Chr.2: 232483160-232483160 on Build GRCh38
rs1026241373 Chr.2: 232483319-232483319 on Build GRCh38
rs976845737 Chr.2: 232483094-232483094 on Build GRCh38
rs757285509 Chr.2: 232483060-232483060 on Build GRCh38
rs1055954547 Chr.2: 232483484-232483484 on Build GRCh38
rs564578606 Chr.2: 232483280-232483280 on Build GRCh38
rs569970209 Chr.2: 232483127-232483127 on Build GRCh38
rs375170118 Chr.2: 232483109-232483109 on Build GRCh38
rs995646527 Chr.2: 232483307-232483307 on Build GRCh38
rs780547464 Chr.2: 232483382-232483382 on Build GRCh38
rs762090965 Chr.2: 232483189-232483189 on Build GRCh38
rs2742077 Chr.2: 232483295-232483295 on Build GRCh38
rs377677167 Chr.2: 232483380-232483380 on Build GRCh38
rs760185996 Chr.2: 232483343-232483343 on Build GRCh38
rs780823763 Chr.2: 232483453-232483453 on Build GRCh38
rs549291711 Chr.2: 232483322-232483322 on Build GRCh38
rs781682011 Chr.2: 232483067-232483067 on Build GRCh38
rs960913879 Chr.2: 232483046-232483046 on Build GRCh38
rs71398722 Chr.2: 232483232-232483233 on Build GRCh38
rs935407519 Chr.2: 232483409-232483409 on Build GRCh38
rs148278283 Chr.2: 232483472-232483472 on Build GRCh38
rs971576250 Chr.2: 232483178-232483179 on Build GRCh38
rs780468559 Chr.2: 232483072-232483072 on Build GRCh38
rs770974424 Chr.2: 232483097-232483097 on Build GRCh38
rs764253018 Chr.2: 232483122-232483122 on Build GRCh38
rs749116027 Chr.2: 232483471-232483471 on Build GRCh38
rs1010356091 Chr.2: 232483117-232483117 on Build GRCh38
rs759173837 Chr.2: 232483406-232483406 on Build GRCh38
rs766518409 Chr.2: 232483412-232483412 on Build GRCh38
rs751758652 Chr.2: 232483124-232483124 on Build GRCh38
rs186824780 Chr.2: 232483353-232483353 on Build GRCh38
rs568689441 Chr.2: 232483183-232483183 on Build GRCh38

View Full Product Details