Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 241106411-241106919 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PASK
Entrez Gene ID
23178
Gene Name
PAS domain containing serine/threonine kinase
Gene Aliases
PASKIN, STK37
UniGene
Hs.397891
Gene Chromosome Location
Chr.2: 241106099-241150347 on Build GRCh38
Transcripts
NM_001252119.1 NM_001252120.1 NM_001252122.1 NM_015148.3 XM_005246991.1 XM_011510826.1 XM_011510828.1 XM_011510829.1 XM_011510830.1 XM_011510831.2 XM_011510832.2 XM_011510833.1 XM_011510834.1 XM_011510835.1 XM_017003634.1 XM_017003635.1 XM_017003636.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5032402 c.3876C>T p.A1292A Chr.2: 241106662-241106662 on Build GRCh38
4790099 c.3843G>A p.L1281L Chr.2: 241106695-241106695 on Build GRCh38
4711292 c.3916G>A p.A1306T Chr.2: 241106622-241106622 on Build GRCh38
4092961 c.3880G>A p.A1294T Chr.2: 241106658-241106658 on Build GRCh38
7087046 c.3886G>A p.E1296K Chr.2: 241106652-241106652 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751867270 Chr.2: 241106721-241106721 on Build GRCh38
rs1022532167 Chr.2: 241106820-241106820 on Build GRCh38
rs1020853036 Chr.2: 241106677-241106677 on Build GRCh38
rs892092707 Chr.2: 241106791-241106791 on Build GRCh38
rs762530395 Chr.2: 241106532-241106532 on Build GRCh38
rs771678808 Chr.2: 241106657-241106657 on Build GRCh38
rs558228496 Chr.2: 241106551-241106551 on Build GRCh38
rs769638357 Chr.2: 241106525-241106525 on Build GRCh38
rs759812741 Chr.2: 241106571-241106571 on Build GRCh38
rs779771072 Chr.2: 241106631-241106631 on Build GRCh38
rs375810546 Chr.2: 241106753-241106753 on Build GRCh38
rs771635795 Chr.2: 241106775-241106775 on Build GRCh38
rs557193827 Chr.2: 241106635-241106635 on Build GRCh38
rs749109586 Chr.2: 241106633-241106633 on Build GRCh38
rs373117938 Chr.2: 241106774-241106774 on Build GRCh38
rs367872189 Chr.2: 241106583-241106583 on Build GRCh38
rs942464454 Chr.2: 241106468-241106468 on Build GRCh38
rs756002399 Chr.2: 241106735-241106735 on Build GRCh38
rs753071648 Chr.2: 241106706-241106706 on Build GRCh38
rs142896821 Chr.2: 241106611-241106611 on Build GRCh38
rs748435091 Chr.2: 241106826-241106826 on Build GRCh38
rs55860398 Chr.2: 241106637-241106637 on Build GRCh38
rs754425460 Chr.2: 241106566-241106566 on Build GRCh38
rs148038387 Chr.2: 241106634-241106634 on Build GRCh38
rs761424648 Chr.2: 241106554-241106554 on Build GRCh38
rs775123688 Chr.2: 241106528-241106528 on Build GRCh38
rs909735371 Chr.2: 241106691-241106691 on Build GRCh38
rs966622597 Chr.2: 241106689-241106689 on Build GRCh38
rs905385314 Chr.2: 241106893-241106893 on Build GRCh38
rs1047954433 Chr.2: 241106497-241106497 on Build GRCh38
rs758553845 Chr.2: 241106580-241106583 on Build GRCh38
rs755203844 Chr.2: 241106758-241106760 on Build GRCh38
rs969517582 Chr.2: 241106585-241106585 on Build GRCh38
rs1011170535 Chr.2: 241106800-241106800 on Build GRCh38
rs369590455 Chr.2: 241106678-241106678 on Build GRCh38
rs548780816 Chr.2: 241106788-241106788 on Build GRCh38
rs763956317 Chr.2: 241106518-241106519 on Build GRCh38
rs781270108 Chr.2: 241106728-241106728 on Build GRCh38
rs900868934 Chr.2: 241106482-241106482 on Build GRCh38
rs780346610 Chr.2: 241106593-241106593 on Build GRCh38
rs751611661 Chr.2: 241106708-241106708 on Build GRCh38
rs765866099 Chr.2: 241106702-241106702 on Build GRCh38
rs368922121 Chr.2: 241106591-241106591 on Build GRCh38
rs369208565 Chr.2: 241106768-241106768 on Build GRCh38
rs534349867 Chr.2: 241106592-241106592 on Build GRCh38
rs984413329 Chr.2: 241106679-241106679 on Build GRCh38
rs114285040 Chr.2: 241106582-241106582 on Build GRCh38
rs73112118 Chr.2: 241106790-241106790 on Build GRCh38
rs750485918 Chr.2: 241106733-241106733 on Build GRCh38
rs200311696 Chr.2: 241106625-241106625 on Build GRCh38
rs747069889 Chr.2: 241106704-241106705 on Build GRCh38
rs753768742 Chr.2: 241106518-241106521 on Build GRCh38
rs753529193 Chr.2: 241106474-241106474 on Build GRCh38
rs199833369 Chr.2: 241106597-241106597 on Build GRCh38
rs950345396 Chr.2: 241106874-241106874 on Build GRCh38
rs757282946 Chr.2: 241106726-241106726 on Build GRCh38
rs545320438 Chr.2: 241106473-241106473 on Build GRCh38
rs749057604 Chr.2: 241106740-241106740 on Build GRCh38
rs766771331 Chr.2: 241106559-241106559 on Build GRCh38
rs781013176 Chr.2: 241106755-241106755 on Build GRCh38
rs371946744 Chr.2: 241106593-241106593 on Build GRCh38
rs374993877 Chr.2: 241106519-241106519 on Build GRCh38
rs867710969 Chr.2: 241106711-241106711 on Build GRCh38
rs974883743 Chr.2: 241106752-241106752 on Build GRCh38
rs886706935 Chr.2: 241106518-241106518 on Build GRCh38
rs888243722 Chr.2: 241106438-241106438 on Build GRCh38
rs761573627 Chr.2: 241106494-241106494 on Build GRCh38
rs1039403188 Chr.2: 241106480-241106480 on Build GRCh38
rs141459975 Chr.2: 241106707-241106707 on Build GRCh38
rs769732105 Chr.2: 241106626-241106626 on Build GRCh38
rs3732227 Chr.2: 241106736-241106736 on Build GRCh38
rs760062049 Chr.2: 241106698-241106698 on Build GRCh38
rs768316537 Chr.2: 241106541-241106541 on Build GRCh38

View Full Product Details