Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 189497-190000 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DEFB128
Entrez Gene ID
245939
Gene Name
defensin beta 128
Gene Aliases
DEFB-28, DEFB28, hBD-28
UniGene
Hs.516818
Gene Chromosome Location
Chr.20: 187853-189681 on Build GRCh38
Transcripts
NM_001037732.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
723564 c.34T>A p.F12I Chr.20: 189590-189590 on Build GRCh38
6642202 c.49G>A p.D17N Chr.20: 189575-189575 on Build GRCh38
5590500 c.3G>A p.M1I Chr.20: 189621-189621 on Build GRCh38
1726686 c.8T>G p.L3R Chr.20: 189616-189616 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1009926431 Chr.20: 189700-189700 on Build GRCh38
rs186231332 Chr.20: 189609-189609 on Build GRCh38
rs925966684 Chr.20: 189872-189872 on Build GRCh38
rs756571397 Chr.20: 189602-189602 on Build GRCh38
rs181351730 Chr.20: 189871-189871 on Build GRCh38
rs757520751 Chr.20: 189806-189806 on Build GRCh38
rs529895775 Chr.20: 189723-189723 on Build GRCh38
rs767465243 Chr.20: 189539-189539 on Build GRCh38
rs764369969 Chr.20: 189531-189531 on Build GRCh38
rs1051347760 Chr.20: 189689-189689 on Build GRCh38
rs548579362 Chr.20: 189760-189760 on Build GRCh38
rs777661205 Chr.20: 189694-189694 on Build GRCh38
rs545721948 Chr.20: 189565-189565 on Build GRCh38
rs142647569 Chr.20: 189584-189584 on Build GRCh38
rs746611198 Chr.20: 189578-189578 on Build GRCh38
rs866653422 Chr.20: 189523-189523 on Build GRCh38
rs191061572 Chr.20: 189941-189941 on Build GRCh38
rs898266767 Chr.20: 189776-189776 on Build GRCh38
rs574485955 Chr.20: 189936-189936 on Build GRCh38
rs545684923 Chr.20: 189967-189967 on Build GRCh38
rs763753297 Chr.20: 189675-189675 on Build GRCh38
rs754917044 Chr.20: 189556-189556 on Build GRCh38
rs150510703 Chr.20: 189781-189781 on Build GRCh38
rs758252897 Chr.20: 189566-189566 on Build GRCh38
rs185821314 Chr.20: 189897-189897 on Build GRCh38
rs774674636 Chr.20: 189641-189641 on Build GRCh38
rs796382079 Chr.20: 189834-189835 on Build GRCh38
rs983527945 Chr.20: 189844-189844 on Build GRCh38
rs771170192 Chr.20: 189724-189724 on Build GRCh38
rs936151395 Chr.20: 189617-189617 on Build GRCh38
rs772096778 Chr.20: 189838-189838 on Build GRCh38
rs993893529 Chr.20: 189962-189962 on Build GRCh38
rs1021372661 Chr.20: 189705-189705 on Build GRCh38
rs369242723 Chr.20: 189533-189533 on Build GRCh38
rs138365912 Chr.20: 189904-189904 on Build GRCh38
rs546518270 Chr.20: 189727-189727 on Build GRCh38
rs531767077 Chr.20: 189704-189704 on Build GRCh38
rs752658728 Chr.20: 189669-189669 on Build GRCh38
rs753552363 Chr.20: 189780-189780 on Build GRCh38
rs761901494 Chr.20: 189651-189651 on Build GRCh38
rs777616956 Chr.20: 189577-189577 on Build GRCh38
rs989308030 Chr.20: 189892-189892 on Build GRCh38
rs756820058 Chr.20: 189581-189581 on Build GRCh38
rs74340988 Chr.20: 189557-189557 on Build GRCh38
rs768991463 Chr.20: 189637-189637 on Build GRCh38
rs1006595469 Chr.20: 189695-189695 on Build GRCh38
rs145944118 Chr.20: 189622-189622 on Build GRCh38
rs762005415 Chr.20: 189536-189536 on Build GRCh38
rs960628911 Chr.20: 189522-189522 on Build GRCh38
rs202123340 Chr.20: 189623-189623 on Build GRCh38
rs957446221 Chr.20: 189886-189886 on Build GRCh38
rs764107542 Chr.20: 189684-189684 on Build GRCh38
rs373187977 Chr.20: 189555-189555 on Build GRCh38
rs535293780 Chr.20: 189901-189901 on Build GRCh38
rs911505239 Chr.20: 189920-189920 on Build GRCh38
rs970020206 Chr.20: 189817-189817 on Build GRCh38
rs746397816 Chr.20: 189593-189593 on Build GRCh38
rs577637569 Chr.20: 189896-189897 on Build GRCh38
rs1047290530 Chr.20: 189970-189970 on Build GRCh38
rs917209085 Chr.20: 189541-189541 on Build GRCh38
rs994341023 Chr.20: 189779-189779 on Build GRCh38
rs773109710 Chr.20: 189659-189659 on Build GRCh38
rs749537653 Chr.20: 189680-189680 on Build GRCh38
rs745912565 Chr.20: 189815-189815 on Build GRCh38
rs1469782 Chr.20: 189948-189948 on Build GRCh38
rs753846010 Chr.20: 189552-189552 on Build GRCh38
rs891970601 Chr.20: 189632-189632 on Build GRCh38
rs148019784 Chr.20: 189716-189716 on Build GRCh38
rs776936746 Chr.20: 189667-189667 on Build GRCh38
rs765321223 Chr.20: 189661-189661 on Build GRCh38
rs141677294 Chr.20: 189728-189728 on Build GRCh38
rs758308034 Chr.20: 189671-189671 on Build GRCh38
rs183645273 Chr.20: 189944-189944 on Build GRCh38
rs918071546 Chr.20: 189740-189741 on Build GRCh38

View Full Product Details