Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 67707514-67708023 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNG12
Entrez Gene ID
55970
Gene Name
G protein subunit gamma 12
Gene Aliases
-
UniGene
Hs.431101
Gene Chromosome Location
Chr.1: 67701466-67833779 on Build GRCh38
Transcripts
NM_018841.5 XM_017001809.1 XM_017001810.1 XM_017001811.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3360991 c.29A>G p.N10S Chr.1: 67707658-67707658 on Build GRCh38
4009670 c.35C>T p.A12V Chr.1: 67707652-67707652 on Build GRCh38
7131650 c.20G>T p.S7I Chr.1: 67707667-67707667 on Build GRCh38
4777551 c.17C>A p.A6E Chr.1: 67707670-67707670 on Build GRCh38
4778183 c.20G>C p.S7T Chr.1: 67707667-67707667 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs891911183 Chr.1: 67707959-67707959 on Build GRCh38
rs763401953 Chr.1: 67707656-67707656 on Build GRCh38
rs148601799 Chr.1: 67707648-67707648 on Build GRCh38
rs575518588 Chr.1: 67707640-67707640 on Build GRCh38
rs775977018 Chr.1: 67707655-67707655 on Build GRCh38
rs1009058760 Chr.1: 67707937-67707937 on Build GRCh38
rs777709090 Chr.1: 67707721-67707721 on Build GRCh38
rs603912 Chr.1: 67707853-67707853 on Build GRCh38
rs183019249 Chr.1: 67707970-67707970 on Build GRCh38
rs909770679 Chr.1: 67707830-67707830 on Build GRCh38
rs985508744 Chr.1: 67707729-67707729 on Build GRCh38
rs146714036 Chr.1: 67707670-67707670 on Build GRCh38
rs777401241 Chr.1: 67707567-67707568 on Build GRCh38
rs2295942 Chr.1: 67707581-67707581 on Build GRCh38
rs745687696 Chr.1: 67707730-67707731 on Build GRCh38
rs761792170 Chr.1: 67707957-67707957 on Build GRCh38
rs377152082 Chr.1: 67707696-67707696 on Build GRCh38
rs1033018155 Chr.1: 67707966-67707966 on Build GRCh38
rs900608613 Chr.1: 67707950-67707950 on Build GRCh38
rs1026499451 Chr.1: 67707582-67707582 on Build GRCh38
rs938721232 Chr.1: 67707954-67707954 on Build GRCh38
rs930911766 Chr.1: 67707864-67707864 on Build GRCh38
rs754761560 Chr.1: 67707559-67707559 on Build GRCh38
rs376430298 Chr.1: 67707968-67707968 on Build GRCh38
rs536592431 Chr.1: 67707863-67707863 on Build GRCh38
rs190440273 Chr.1: 67707909-67707909 on Build GRCh38
rs1008944006 Chr.1: 67707588-67707588 on Build GRCh38
rs1021768042 Chr.1: 67707987-67707987 on Build GRCh38
rs968956579 Chr.1: 67707723-67707723 on Build GRCh38
rs937728308 Chr.1: 67707761-67707761 on Build GRCh38
rs958000590 Chr.1: 67707662-67707662 on Build GRCh38
rs756909558 Chr.1: 67707722-67707723 on Build GRCh38
rs776177860 Chr.1: 67707635-67707635 on Build GRCh38
rs976518596 Chr.1: 67707758-67707758 on Build GRCh38
rs767597583 Chr.1: 67707666-67707666 on Build GRCh38
rs568794650 Chr.1: 67707535-67707535 on Build GRCh38
rs201538319 Chr.1: 67707580-67707580 on Build GRCh38
rs918377163 Chr.1: 67707780-67707780 on Build GRCh38
rs557977842 Chr.1: 67707657-67707657 on Build GRCh38
rs778480911 Chr.1: 67707723-67707723 on Build GRCh38
rs1019052244 Chr.1: 67707944-67707944 on Build GRCh38
rs766497630 Chr.1: 67707692-67707692 on Build GRCh38
rs983981772 Chr.1: 67707799-67707799 on Build GRCh38
rs189511777 Chr.1: 67707551-67707551 on Build GRCh38
rs186431887 Chr.1: 67707850-67707850 on Build GRCh38
rs768405915 Chr.1: 67707817-67707817 on Build GRCh38
rs141636525 Chr.1: 67707687-67707687 on Build GRCh38
rs61754629 Chr.1: 67707651-67707651 on Build GRCh38
rs181022053 Chr.1: 67707730-67707730 on Build GRCh38
rs17850020 Chr.1: 67707602-67707602 on Build GRCh38
rs777619534 Chr.1: 67707591-67707591 on Build GRCh38
rs531876641 Chr.1: 67707906-67707906 on Build GRCh38
rs199827530 Chr.1: 67707567-67707567 on Build GRCh38
rs578162334 Chr.1: 67707674-67707674 on Build GRCh38
rs752059593 Chr.1: 67707658-67707658 on Build GRCh38
rs565120217 Chr.1: 67707960-67707960 on Build GRCh38
rs1044414633 Chr.1: 67707571-67707571 on Build GRCh38
rs964762627 Chr.1: 67707645-67707645 on Build GRCh38
rs748743389 Chr.1: 67707582-67707582 on Build GRCh38
rs376652206 Chr.1: 67707568-67707568 on Build GRCh38
rs1036233206 Chr.1: 67707585-67707585 on Build GRCh38
rs762008495 Chr.1: 67707661-67707661 on Build GRCh38
rs746535321 Chr.1: 67707614-67707614 on Build GRCh38
rs560303172 Chr.1: 67707751-67707751 on Build GRCh38
rs929775574 Chr.1: 67707791-67707791 on Build GRCh38
rs755408086 Chr.1: 67707546-67707546 on Build GRCh38
rs757152715 Chr.1: 67707733-67707733 on Build GRCh38
rs770377785 Chr.1: 67707618-67707618 on Build GRCh38

View Full Product Details