Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 68148048-68148544 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNG12-AS1
Entrez Gene ID
100289178
Gene Name
GNG12 antisense RNA 1
Gene Aliases
-
UniGene
Hs.677603
Gene Chromosome Location
Chr.1: 67832288-68202987 on Build GRCh38
Transcripts
NR_040077.1 NM_001002292.3 NM_001193334.1 NM_024911.6 XM_011542191.2 XM_011542192.2 XM_017002390.1

Gene Symbol
WLS
Entrez Gene ID
79971
Gene Name
wntless Wnt ligand secretion mediator
Gene Aliases
C1orf139, EVI, GPR177, MRP, mig-14
UniGene
Hs.647659
Gene Chromosome Location
Chr.1: 68098459-68233000 on Build GRCh38
Transcripts
NR_040077.1 NM_001002292.3 NM_001193334.1 NM_024911.6 XM_011542191.2 XM_011542192.2 XM_017002390.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6344499 c.1109C>A p.T370N Chr.1: 68148161-68148161 on Build GRCh38
4619075 c.1085C>T p.T362M Chr.1: 68148185-68148185 on Build GRCh38
4619074 c.1079C>T p.T360M Chr.1: 68148185-68148185 on Build GRCh38
5792439 c.1069G>T p.V357L Chr.1: 68148195-68148195 on Build GRCh38
5792440 c.1075G>T p.V359L Chr.1: 68148195-68148195 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs553438738 Chr.1: 68148516-68148516 on Build GRCh38
rs962474278 Chr.1: 68148412-68148412 on Build GRCh38
rs200839533 Chr.1: 68148185-68148185 on Build GRCh38
rs1001829703 Chr.1: 68148083-68148083 on Build GRCh38
rs912017348 Chr.1: 68148301-68148301 on Build GRCh38
rs377649019 Chr.1: 68148161-68148161 on Build GRCh38
rs759360066 Chr.1: 68148206-68148206 on Build GRCh38
rs758331534 Chr.1: 68148130-68148130 on Build GRCh38
rs755807405 Chr.1: 68148514-68148514 on Build GRCh38
rs769709973 Chr.1: 68148127-68148127 on Build GRCh38
rs765062207 Chr.1: 68148220-68148220 on Build GRCh38
rs770242439 Chr.1: 68148156-68148156 on Build GRCh38
rs992504131 Chr.1: 68148445-68148445 on Build GRCh38
rs775339419 Chr.1: 68148222-68148222 on Build GRCh38
rs777203536 Chr.1: 68148135-68148135 on Build GRCh38
rs746256483 Chr.1: 68148152-68148152 on Build GRCh38
rs767918428 Chr.1: 68148455-68148455 on Build GRCh38
rs903299472 Chr.1: 68148213-68148213 on Build GRCh38
rs950142058 Chr.1: 68148153-68148153 on Build GRCh38
rs767723226 Chr.1: 68148097-68148097 on Build GRCh38
rs551825485 Chr.1: 68148346-68148346 on Build GRCh38
rs988475650 Chr.1: 68148365-68148365 on Build GRCh38
rs769953132 Chr.1: 68148111-68148111 on Build GRCh38
rs1015279100 Chr.1: 68148398-68148398 on Build GRCh38
rs6670417 Chr.1: 68148085-68148085 on Build GRCh38
rs2566789 Chr.1: 68148330-68148330 on Build GRCh38
rs769194665 Chr.1: 68148167-68148167 on Build GRCh38
rs539858795 Chr.1: 68148489-68148489 on Build GRCh38
rs936075665 Chr.1: 68148492-68148492 on Build GRCh38
rs76273218 Chr.1: 68148472-68148472 on Build GRCh38
rs973702992 Chr.1: 68148418-68148418 on Build GRCh38
rs1029805567 Chr.1: 68148359-68148359 on Build GRCh38
rs762860449 Chr.1: 68148239-68148239 on Build GRCh38
rs959280989 Chr.1: 68148444-68148444 on Build GRCh38
rs773394076 Chr.1: 68148124-68148128 on Build GRCh38
rs531983505 Chr.1: 68148344-68148344 on Build GRCh38
rs780933286 Chr.1: 68148140-68148140 on Build GRCh38
rs570833056 Chr.1: 68148483-68148483 on Build GRCh38
rs537712753 Chr.1: 68148467-68148467 on Build GRCh38
rs987605965 Chr.1: 68148300-68148300 on Build GRCh38
rs774363072 Chr.1: 68148175-68148175 on Build GRCh38
rs753638193 Chr.1: 68148099-68148099 on Build GRCh38
rs568605319 Chr.1: 68148466-68148466 on Build GRCh38
rs955741656 Chr.1: 68148362-68148362 on Build GRCh38
rs756860259 Chr.1: 68148139-68148139 on Build GRCh38
rs138033241 Chr.1: 68148136-68148136 on Build GRCh38
rs751043409 Chr.1: 68148246-68148246 on Build GRCh38
rs563741495 Chr.1: 68148269-68148269 on Build GRCh38
rs774498776 Chr.1: 68148364-68148364 on Build GRCh38
rs773376949 Chr.1: 68148203-68148203 on Build GRCh38
rs975422103 Chr.1: 68148328-68148328 on Build GRCh38
rs528203523 Chr.1: 68148371-68148371 on Build GRCh38
rs750139640 Chr.1: 68148513-68148513 on Build GRCh38
rs965007534 Chr.1: 68148306-68148306 on Build GRCh38
rs973732545 Chr.1: 68148081-68148081 on Build GRCh38
rs749809422 Chr.1: 68148164-68148164 on Build GRCh38
rs912990432 Chr.1: 68148366-68148366 on Build GRCh38
rs112690441 Chr.1: 68148111-68148111 on Build GRCh38
rs2303949 Chr.1: 68148250-68148250 on Build GRCh38
rs200433684 Chr.1: 68148517-68148517 on Build GRCh38
rs937959854 Chr.1: 68148115-68148115 on Build GRCh38
rs748259304 Chr.1: 68148093-68148093 on Build GRCh38
rs573830869 Chr.1: 68148184-68148184 on Build GRCh38
rs373490737 Chr.1: 68148241-68148241 on Build GRCh38
rs767307033 Chr.1: 68148251-68148251 on Build GRCh38
rs759272195 Chr.1: 68148120-68148120 on Build GRCh38
rs905669698 Chr.1: 68148438-68148438 on Build GRCh38
rs559036028 Chr.1: 68148358-68148358 on Build GRCh38
rs900046504 Chr.1: 68148319-68148319 on Build GRCh38
rs773698381 Chr.1: 68148479-68148479 on Build GRCh38
rs891529 Chr.1: 68148439-68148439 on Build GRCh38
rs574673896 Chr.1: 68148193-68148193 on Build GRCh38
rs866873411 Chr.1: 68148464-68148464 on Build GRCh38
rs917731772 Chr.1: 68148460-68148460 on Build GRCh38
rs749276343 Chr.1: 68148294-68148294 on Build GRCh38

View Full Product Details