Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 71067069-71067580 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZRANB2
Entrez Gene ID
9406
Gene Name
zinc finger RANBP2-type containing 2
Gene Aliases
ZIS, ZIS1, ZIS2, ZNF265
UniGene
Hs.194718
Gene Chromosome Location
Chr.1: 71063291-71081289 on Build GRCh38
Transcripts
NM_005455.4 NM_203350.2 NR_038420.1

Gene Symbol
ZRANB2-AS1
Entrez Gene ID
100132618
Gene Name
ZRANB2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.720513
Gene Chromosome Location
Chr.1: 71046506-71067184 on Build GRCh38
Transcripts
NM_005455.4 NM_203350.2 NR_038420.1

SNP Detail

SNP ID Chromosome Location
rs186002934 Chr.1: 71067203-71067203 on Build GRCh38
rs1042942940 Chr.1: 71067238-71067238 on Build GRCh38
rs1053059441 Chr.1: 71067437-71067437 on Build GRCh38
rs575851798 Chr.1: 71067359-71067359 on Build GRCh38
rs1055646489 Chr.1: 71067517-71067517 on Build GRCh38
rs888906274 Chr.1: 71067197-71067197 on Build GRCh38
rs1031407203 Chr.1: 71067258-71067258 on Build GRCh38
rs1025061656 Chr.1: 71067292-71067292 on Build GRCh38
rs914302879 Chr.1: 71067107-71067107 on Build GRCh38
rs1026572975 Chr.1: 71067553-71067553 on Build GRCh38
rs956743453 Chr.1: 71067276-71067276 on Build GRCh38
rs963581511 Chr.1: 71067326-71067326 on Build GRCh38
rs569948662 Chr.1: 71067320-71067320 on Build GRCh38
rs549828417 Chr.1: 71067443-71067443 on Build GRCh38
rs868119587 Chr.1: 71067389-71067389 on Build GRCh38
rs987590626 Chr.1: 71067374-71067374 on Build GRCh38
rs980897512 Chr.1: 71067300-71067300 on Build GRCh38
rs764893667 Chr.1: 71067277-71067277 on Build GRCh38
rs1012178752 Chr.1: 71067253-71067253 on Build GRCh38
rs938481213 Chr.1: 71067495-71067495 on Build GRCh38
rs976283214 Chr.1: 71067342-71067342 on Build GRCh38
rs933053549 Chr.1: 71067177-71067177 on Build GRCh38
rs1002270722 Chr.1: 71067496-71067496 on Build GRCh38
rs918955611 Chr.1: 71067176-71067176 on Build GRCh38
rs1041518589 Chr.1: 71067113-71067113 on Build GRCh38
rs190859238 Chr.1: 71067520-71067520 on Build GRCh38
rs71500632 Chr.1: 71067350-71067350 on Build GRCh38
rs533826033 Chr.1: 71067110-71067110 on Build GRCh38
rs549106878 Chr.1: 71067462-71067462 on Build GRCh38
rs931216189 Chr.1: 71067539-71067539 on Build GRCh38
rs1048332910 Chr.1: 71067550-71067550 on Build GRCh38
rs1014999487 Chr.1: 71067313-71067313 on Build GRCh38
rs544354996 Chr.1: 71067449-71067449 on Build GRCh38
rs998795303 Chr.1: 71067255-71067255 on Build GRCh38
rs535642587 Chr.1: 71067392-71067392 on Build GRCh38
rs904386057 Chr.1: 71067407-71067407 on Build GRCh38
rs1027592662 Chr.1: 71067274-71067274 on Build GRCh38
rs1016426792 Chr.1: 71067301-71067301 on Build GRCh38
rs914230636 Chr.1: 71067405-71067405 on Build GRCh38
rs560742912 Chr.1: 71067241-71067241 on Build GRCh38
rs934544520 Chr.1: 71067430-71067430 on Build GRCh38
rs1051537069 Chr.1: 71067183-71067183 on Build GRCh38
rs562338409 Chr.1: 71067467-71067470 on Build GRCh38
rs966569183 Chr.1: 71067316-71067316 on Build GRCh38
rs1016376093 Chr.1: 71067505-71067505 on Build GRCh38
rs564853308 Chr.1: 71067549-71067549 on Build GRCh38
rs933021106 Chr.1: 71067351-71067351 on Build GRCh38
rs945763068 Chr.1: 71067112-71067112 on Build GRCh38
rs990426525 Chr.1: 71067398-71067398 on Build GRCh38
rs938567660 Chr.1: 71067143-71067143 on Build GRCh38
rs1004914804 Chr.1: 71067281-71067281 on Build GRCh38
rs887808125 Chr.1: 71067280-71067280 on Build GRCh38
rs542363305 Chr.1: 71067234-71067234 on Build GRCh38
rs972373957 Chr.1: 71067328-71067328 on Build GRCh38
rs969495588 Chr.1: 71067299-71067299 on Build GRCh38

View Full Product Details