Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 4856238-4856746 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC23A2
Entrez Gene ID
9962
Gene Name
solute carrier family 23 member 2
Gene Aliases
NBTL1, SLC23A1, SVCT2, YSPL2, hSVCT2
UniGene
Hs.516866
Gene Chromosome Location
Chr.20: 4852356-5010293 on Build GRCh38
Transcripts
NM_005116.5 NM_203327.1 XM_011529414.2 XM_011529415.2 XM_011529416.2 XM_011529417.2 XM_017028171.1 XM_017028172.1 XM_017028173.1 XM_017028174.1 XM_017028175.1 XM_017028176.1 XM_017028177.1 XM_017028178.1 XM_017028179.1 XM_017028180.1

SNP Detail

SNP ID Chromosome Location
rs1020443464 Chr.20: 4856420-4856420 on Build GRCh38
rs900111048 Chr.20: 4856590-4856590 on Build GRCh38
rs958912940 Chr.20: 4856704-4856704 on Build GRCh38
rs563439781 Chr.20: 4856588-4856588 on Build GRCh38
rs6037992 Chr.20: 4856675-4856675 on Build GRCh38
rs560840935 Chr.20: 4856589-4856589 on Build GRCh38
rs1014734720 Chr.20: 4856636-4856636 on Build GRCh38
rs930869745 Chr.20: 4856268-4856268 on Build GRCh38
rs765571667 Chr.20: 4856551-4856551 on Build GRCh38
rs965813060 Chr.20: 4856266-4856266 on Build GRCh38
rs112295813 Chr.20: 4856462-4856462 on Build GRCh38
rs773626224 Chr.20: 4856419-4856419 on Build GRCh38
rs16990314 Chr.20: 4856513-4856513 on Build GRCh38
rs1041835384 Chr.20: 4856583-4856583 on Build GRCh38
rs904749731 Chr.20: 4856436-4856436 on Build GRCh38
rs190449853 Chr.20: 4856355-4856355 on Build GRCh38
rs553982279 Chr.20: 4856412-4856412 on Build GRCh38
rs575964499 Chr.20: 4856301-4856301 on Build GRCh38
rs370156206 Chr.20: 4856562-4856562 on Build GRCh38
rs974490173 Chr.20: 4856516-4856516 on Build GRCh38
rs529930794 Chr.20: 4856550-4856550 on Build GRCh38
rs548424703 Chr.20: 4856560-4856560 on Build GRCh38
rs1030447929 Chr.20: 4856607-4856607 on Build GRCh38
rs541210880 Chr.20: 4856541-4856541 on Build GRCh38
rs73597400 Chr.20: 4856686-4856686 on Build GRCh38
rs770519623 Chr.20: 4856465-4856465 on Build GRCh38
rs772603571 Chr.20: 4856451-4856451 on Build GRCh38
rs954445152 Chr.20: 4856613-4856613 on Build GRCh38
rs963191764 Chr.20: 4856667-4856667 on Build GRCh38
rs191677713 Chr.20: 4856501-4856501 on Build GRCh38
rs954648991 Chr.20: 4856468-4856468 on Build GRCh38
rs571457144 Chr.20: 4856274-4856274 on Build GRCh38
rs73893848 Chr.20: 4856416-4856416 on Build GRCh38
rs186778700 Chr.20: 4856418-4856418 on Build GRCh38
rs748023678 Chr.20: 4856389-4856389 on Build GRCh38
rs144294752 Chr.20: 4856480-4856480 on Build GRCh38
rs903828044 Chr.20: 4856695-4856696 on Build GRCh38
rs114992000 Chr.20: 4856477-4856477 on Build GRCh38
rs185825841 Chr.20: 4856267-4856267 on Build GRCh38
rs995847276 Chr.20: 4856600-4856600 on Build GRCh38
rs541843451 Chr.20: 4856369-4856369 on Build GRCh38
rs936649732 Chr.20: 4856713-4856713 on Build GRCh38
rs974837164 Chr.20: 4856463-4856463 on Build GRCh38
rs973339291 Chr.20: 4856500-4856500 on Build GRCh38
rs759614452 Chr.20: 4856629-4856629 on Build GRCh38
rs1000407382 Chr.20: 4856438-4856438 on Build GRCh38
rs941890931 Chr.20: 4856489-4856489 on Build GRCh38
rs564677071 Chr.20: 4856585-4856585 on Build GRCh38
rs559615782 Chr.20: 4856542-4856542 on Build GRCh38
rs16990312 Chr.20: 4856417-4856417 on Build GRCh38
rs1057450598 Chr.20: 4856552-4856552 on Build GRCh38
rs1037673579 Chr.20: 4856299-4856299 on Build GRCh38
rs767367632 Chr.20: 4856605-4856605 on Build GRCh38

View Full Product Details