Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 5545149-5545692 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPCPD1
Entrez Gene ID
56261
Gene Name
glycerophosphocholine phosphodiesterase 1
Gene Aliases
EDI3, GDE5, GDPD6, PREI4
UniGene
Hs.636359
Gene Chromosome Location
Chr.20: 5544434-5611026 on Build GRCh38
Transcripts
NM_019593.3 XM_005260758.4 XM_005260761.1 XM_006723596.3

SNP Detail

SNP ID Chromosome Location
rs986002511 Chr.20: 5545577-5545577 on Build GRCh38
rs938540483 Chr.20: 5545378-5545378 on Build GRCh38
rs1055725081 Chr.20: 5545474-5545474 on Build GRCh38
rs543289151 Chr.20: 5545652-5545652 on Build GRCh38
rs1010927176 Chr.20: 5545496-5545496 on Build GRCh38
rs144195496 Chr.20: 5545641-5545641 on Build GRCh38
rs146202378 Chr.20: 5545266-5545266 on Build GRCh38
rs972933332 Chr.20: 5545505-5545505 on Build GRCh38
rs578042758 Chr.20: 5545440-5545440 on Build GRCh38
rs6107650 Chr.20: 5545467-5545467 on Build GRCh38
rs575337488 Chr.20: 5545561-5545561 on Build GRCh38
rs879415576 Chr.20: 5545376-5545376 on Build GRCh38
rs577924440 Chr.20: 5545180-5545180 on Build GRCh38
rs759822730 Chr.20: 5545297-5545297 on Build GRCh38
rs1006883133 Chr.20: 5545427-5545427 on Build GRCh38
rs984978019 Chr.20: 5545558-5545558 on Build GRCh38
rs1007344951 Chr.20: 5545649-5545649 on Build GRCh38
rs369105457 Chr.20: 5545642-5545642 on Build GRCh38
rs1053399 Chr.20: 5545340-5545340 on Build GRCh38
rs766813361 Chr.20: 5545215-5545215 on Build GRCh38
rs929751190 Chr.20: 5545636-5545636 on Build GRCh38
rs929698650 Chr.20: 5545433-5545433 on Build GRCh38
rs1011686041 Chr.20: 5545413-5545413 on Build GRCh38
rs116790932 Chr.20: 5545528-5545528 on Build GRCh38
rs893016031 Chr.20: 5545494-5545496 on Build GRCh38
rs979796164 Chr.20: 5545564-5545564 on Build GRCh38
rs115424798 Chr.20: 5545650-5545650 on Build GRCh38
rs948430513 Chr.20: 5545562-5545562 on Build GRCh38
rs1020323353 Chr.20: 5545439-5545439 on Build GRCh38
rs1052948388 Chr.20: 5545385-5545385 on Build GRCh38
rs1001452078 Chr.20: 5545207-5545207 on Build GRCh38
rs893078880 Chr.20: 5545656-5545656 on Build GRCh38
rs984535142 Chr.20: 5545281-5545281 on Build GRCh38
rs989854879 Chr.20: 5545255-5545255 on Build GRCh38
rs1044741211 Chr.20: 5545341-5545341 on Build GRCh38
rs911496086 Chr.20: 5545578-5545578 on Build GRCh38
rs138033782 Chr.20: 5545420-5545420 on Build GRCh38
rs978256441 Chr.20: 5545293-5545293 on Build GRCh38
rs540476862 Chr.20: 5545614-5545614 on Build GRCh38
rs571754724 Chr.20: 5545632-5545632 on Build GRCh38
rs774860427 Chr.20: 5545285-5545285 on Build GRCh38
rs751156944 Chr.20: 5545357-5545357 on Build GRCh38
rs543980500 Chr.20: 5545630-5545630 on Build GRCh38
rs188387214 Chr.20: 5545574-5545574 on Build GRCh38
rs941048934 Chr.20: 5545384-5545384 on Build GRCh38
rs1045626292 Chr.20: 5545448-5545448 on Build GRCh38
rs756622757 Chr.20: 5545494-5545494 on Build GRCh38
rs934273562 Chr.20: 5545299-5545299 on Build GRCh38
rs6085202 Chr.20: 5545386-5545386 on Build GRCh38
rs1001736517 Chr.20: 5545470-5545470 on Build GRCh38
rs191336893 Chr.20: 5545590-5545590 on Build GRCh38
rs889812488 Chr.20: 5545424-5545424 on Build GRCh38
rs901161085 Chr.20: 5545654-5545654 on Build GRCh38
rs115150317 Chr.20: 5545599-5545599 on Build GRCh38
rs999560804 Chr.20: 5545555-5545555 on Build GRCh38
rs1053001456 Chr.20: 5545596-5545596 on Build GRCh38
rs575967859 Chr.20: 5545354-5545354 on Build GRCh38
rs959404489 Chr.20: 5545244-5545244 on Build GRCh38
rs1034137220 Chr.20: 5545229-5545229 on Build GRCh38
rs183994276 Chr.20: 5545508-5545508 on Build GRCh38

View Full Product Details