Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 5956995-5957504 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MCM8
Entrez Gene ID
84515
Gene Name
minichromosome maintenance 8 homologous recombination repair factor
Gene Aliases
C20orf154, POF10, dJ967N21.5
UniGene
Hs.597484
Gene Chromosome Location
Chr.20: 5950652-6000941 on Build GRCh38
Transcripts
NM_001281520.1 NM_001281521.1 NM_001281522.1 NM_032485.5 NM_182802.2 XM_011529387.2 XM_017028105.1 XM_017028106.1 XM_017028107.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5766698 c.494C>G p.T165S Chr.20: 5957133-5957133 on Build GRCh38
1615793 c.538G>A p.G180R Chr.20: 5957177-5957177 on Build GRCh38
7125380 c.550G>A p.D184N Chr.20: 5957189-5957189 on Build GRCh38
6451603 c.544T>A p.S182T Chr.20: 5957183-5957183 on Build GRCh38
7125379 c.520G>C p.E174Q Chr.20: 5957159-5957159 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs542232374 Chr.20: 5957248-5957248 on Build GRCh38
rs774482954 Chr.20: 5957152-5957152 on Build GRCh38
rs9917453 Chr.20: 5957295-5957295 on Build GRCh38
rs374488426 Chr.20: 5957332-5957332 on Build GRCh38
rs745857924 Chr.20: 5957226-5957226 on Build GRCh38
rs760183819 Chr.20: 5957185-5957187 on Build GRCh38
rs1038517614 Chr.20: 5957334-5957334 on Build GRCh38
rs201792638 Chr.20: 5957259-5957259 on Build GRCh38
rs146121619 Chr.20: 5957339-5957339 on Build GRCh38
rs775233073 Chr.20: 5957243-5957243 on Build GRCh38
rs200285727 Chr.20: 5957155-5957155 on Build GRCh38
rs41282142 Chr.20: 5957073-5957073 on Build GRCh38
rs761731992 Chr.20: 5957154-5957154 on Build GRCh38
rs977938807 Chr.20: 5957297-5957297 on Build GRCh38
rs747711524 Chr.20: 5957188-5957188 on Build GRCh38
rs773458929 Chr.20: 5957274-5957274 on Build GRCh38
rs370358224 Chr.20: 5957098-5957098 on Build GRCh38
rs528984354 Chr.20: 5957168-5957168 on Build GRCh38
rs542998430 Chr.20: 5957317-5957317 on Build GRCh38
rs746630122 Chr.20: 5957213-5957213 on Build GRCh38
rs755508302 Chr.20: 5957180-5957180 on Build GRCh38
rs759495991 Chr.20: 5957141-5957141 on Build GRCh38
rs777525798 Chr.20: 5957408-5957408 on Build GRCh38
rs1022778844 Chr.20: 5957419-5957419 on Build GRCh38
rs766001359 Chr.20: 5957220-5957220 on Build GRCh38
rs772096815 Chr.20: 5957271-5957271 on Build GRCh38
rs966821765 Chr.20: 5957431-5957431 on Build GRCh38
rs1012491279 Chr.20: 5957045-5957045 on Build GRCh38
rs58310169 Chr.20: 5957364-5957364 on Build GRCh38
rs368612206 Chr.20: 5957055-5957055 on Build GRCh38
rs548189532 Chr.20: 5957448-5957448 on Build GRCh38
rs991473813 Chr.20: 5957462-5957462 on Build GRCh38
rs993711707 Chr.20: 5957151-5957151 on Build GRCh38
rs777600551 Chr.20: 5957202-5957202 on Build GRCh38
rs962243203 Chr.20: 5957438-5957438 on Build GRCh38
rs930631353 Chr.20: 5957298-5957298 on Build GRCh38
rs55942815 Chr.20: 5957052-5957052 on Build GRCh38
rs543117977 Chr.20: 5957033-5957033 on Build GRCh38
rs191319311 Chr.20: 5957023-5957023 on Build GRCh38
rs999301760 Chr.20: 5957059-5957059 on Build GRCh38
rs374828473 Chr.20: 5957158-5957158 on Build GRCh38
rs1010935283 Chr.20: 5957413-5957413 on Build GRCh38
rs754483613 Chr.20: 5957105-5957105 on Build GRCh38
rs933621611 Chr.20: 5957040-5957040 on Build GRCh38
rs749264359 Chr.20: 5957380-5957380 on Build GRCh38
rs73600734 Chr.20: 5957395-5957395 on Build GRCh38
rs1010522218 Chr.20: 5957360-5957360 on Build GRCh38
rs776751706 Chr.20: 5957129-5957129 on Build GRCh38
rs972101361 Chr.20: 5957440-5957440 on Build GRCh38
rs150686080 Chr.20: 5957224-5957224 on Build GRCh38
rs16991591 Chr.20: 5957187-5957187 on Build GRCh38
rs771764854 Chr.20: 5957112-5957112 on Build GRCh38
rs985357582 Chr.20: 5957468-5957468 on Build GRCh38
rs769773862 Chr.20: 5957229-5957229 on Build GRCh38
rs1024842328 Chr.20: 5957049-5957049 on Build GRCh38
rs543575026 Chr.20: 5957338-5957338 on Build GRCh38
rs546960115 Chr.20: 5957062-5957062 on Build GRCh38
rs778461469 Chr.20: 5957106-5957106 on Build GRCh38
rs553913098 Chr.20: 5957302-5957302 on Build GRCh38
rs188517491 Chr.20: 5957324-5957324 on Build GRCh38
rs181247569 Chr.20: 5957356-5957356 on Build GRCh38
rs770810627 Chr.20: 5957126-5957126 on Build GRCh38
rs894939457 Chr.20: 5957041-5957041 on Build GRCh38
rs756497370 Chr.20: 5957169-5957169 on Build GRCh38
rs772945701 Chr.20: 5957117-5957117 on Build GRCh38
rs766695443 Chr.20: 5957171-5957171 on Build GRCh38
rs148807471 Chr.20: 5957354-5957354 on Build GRCh38
rs770900805 Chr.20: 5957219-5957219 on Build GRCh38
rs185070048 Chr.20: 5957450-5957450 on Build GRCh38
rs371608635 Chr.20: 5957149-5957149 on Build GRCh38
rs1019499825 Chr.20: 5957412-5957412 on Build GRCh38
rs911172674 Chr.20: 5957480-5957480 on Build GRCh38
rs756391577 Chr.20: 5957337-5957337 on Build GRCh38
rs114773856 Chr.20: 5957296-5957296 on Build GRCh38
rs895055684 Chr.20: 5957309-5957309 on Build GRCh38
rs747825147 Chr.20: 5957109-5957109 on Build GRCh38
rs530748997 Chr.20: 5957461-5957461 on Build GRCh38
rs142983627 Chr.20: 5957286-5957286 on Build GRCh38
rs746822710 Chr.20: 5957124-5957124 on Build GRCh38
rs564595798 Chr.20: 5957039-5957039 on Build GRCh38
rs572395668 Chr.20: 5957305-5957305 on Build GRCh38
rs754388420 Chr.20: 5957179-5957179 on Build GRCh38
rs1038164519 Chr.20: 5957057-5957057 on Build GRCh38
rs757801718 Chr.20: 5957193-5957193 on Build GRCh38

View Full Product Details