Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 20369375-20369868 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INSM1
Entrez Gene ID
3642
Gene Name
INSM transcriptional repressor 1
Gene Aliases
IA-1, IA1
UniGene
Hs.89584
Gene Chromosome Location
Chr.20: 20368121-20370949 on Build GRCh38
Transcripts
NM_002196.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3544570 c.1212C>T p.A404A Chr.20: 20369479-20369479 on Build GRCh38
7298240 c.1109A>G p.H370R Chr.20: 20369376-20369376 on Build GRCh38
3544571 c.1407C>T p.F469F Chr.20: 20369674-20369674 on Build GRCh38
5967559 c.1237delG p.D415fs*20 Chr.20: 20369504-20369504 on Build GRCh38
7078971 c.1459C>T p.H487Y Chr.20: 20369726-20369726 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs932248291 Chr.20: 20369450-20369450 on Build GRCh38
rs531505955 Chr.20: 20369662-20369662 on Build GRCh38
rs748646966 Chr.20: 20369598-20369598 on Build GRCh38
rs780014369 Chr.20: 20369398-20369398 on Build GRCh38
rs758441285 Chr.20: 20369733-20369733 on Build GRCh38
rs765263425 Chr.20: 20369565-20369565 on Build GRCh38
rs745868622 Chr.20: 20369508-20369508 on Build GRCh38
rs762121750 Chr.20: 20369463-20369463 on Build GRCh38
rs974349556 Chr.20: 20369437-20369437 on Build GRCh38
rs749422329 Chr.20: 20369577-20369577 on Build GRCh38
rs746077325 Chr.20: 20369526-20369526 on Build GRCh38
rs373203712 Chr.20: 20369566-20369566 on Build GRCh38
rs970478715 Chr.20: 20369641-20369641 on Build GRCh38
rs1010989309 Chr.20: 20369715-20369715 on Build GRCh38
rs771301377 Chr.20: 20369410-20369410 on Build GRCh38
rs13038580 Chr.20: 20369701-20369701 on Build GRCh38
rs747841267 Chr.20: 20369408-20369408 on Build GRCh38
rs538411838 Chr.20: 20369431-20369431 on Build GRCh38
rs748852352 Chr.20: 20369776-20369776 on Build GRCh38
rs781528288 Chr.20: 20369424-20369424 on Build GRCh38
rs778667570 Chr.20: 20369504-20369504 on Build GRCh38
rs907734322 Chr.20: 20369614-20369614 on Build GRCh38
rs763081977 Chr.20: 20369814-20369814 on Build GRCh38
rs752571863 Chr.20: 20369506-20369506 on Build GRCh38
rs1001196034 Chr.20: 20369578-20369578 on Build GRCh38
rs1037353749 Chr.20: 20369429-20369429 on Build GRCh38
rs775638830 Chr.20: 20369618-20369618 on Build GRCh38
rs778297710 Chr.20: 20369847-20369847 on Build GRCh38
rs920088967 Chr.20: 20369470-20369470 on Build GRCh38
rs769233282 Chr.20: 20369543-20369543 on Build GRCh38
rs771181787 Chr.20: 20369650-20369650 on Build GRCh38
rs780192370 Chr.20: 20369530-20369530 on Build GRCh38
rs1053675785 Chr.20: 20369585-20369585 on Build GRCh38
rs757721985 Chr.20: 20369820-20369820 on Build GRCh38
rs532755899 Chr.20: 20369809-20369809 on Build GRCh38
rs747327681 Chr.20: 20369606-20369606 on Build GRCh38
rs779530790 Chr.20: 20369596-20369596 on Build GRCh38
rs774797225 Chr.20: 20369453-20369453 on Build GRCh38
rs564197812 Chr.20: 20369630-20369630 on Build GRCh38
rs944093350 Chr.20: 20369521-20369521 on Build GRCh38
rs138934668 Chr.20: 20369635-20369635 on Build GRCh38
rs774995604 Chr.20: 20369690-20369690 on Build GRCh38
rs755151592 Chr.20: 20369633-20369633 on Build GRCh38
rs574942900 Chr.20: 20369568-20369568 on Build GRCh38
rs775159800 Chr.20: 20369664-20369664 on Build GRCh38
rs778774414 Chr.20: 20369403-20369403 on Build GRCh38
rs758929654 Chr.20: 20369564-20369564 on Build GRCh38
rs185120151 Chr.20: 20369569-20369569 on Build GRCh38
rs760326605 Chr.20: 20369554-20369554 on Build GRCh38
rs768859492 Chr.20: 20369754-20369757 on Build GRCh38
rs778669302 Chr.20: 20369467-20369467 on Build GRCh38
rs551376661 Chr.20: 20369838-20369838 on Build GRCh38
rs561434691 Chr.20: 20369723-20369723 on Build GRCh38
rs756379486 Chr.20: 20369572-20369572 on Build GRCh38
rs768749964 Chr.20: 20369676-20369676 on Build GRCh38
rs954372486 Chr.20: 20369706-20369706 on Build GRCh38
rs763518733 Chr.20: 20369449-20369449 on Build GRCh38
rs753223102 Chr.20: 20369492-20369492 on Build GRCh38
rs773581454 Chr.20: 20369605-20369605 on Build GRCh38
rs768356201 Chr.20: 20369839-20369839 on Build GRCh38
rs774689301 Chr.20: 20369550-20369550 on Build GRCh38
rs112131258 Chr.20: 20369476-20369476 on Build GRCh38
rs764643353 Chr.20: 20369611-20369611 on Build GRCh38
rs780094822 Chr.20: 20369576-20369576 on Build GRCh38
rs761658396 Chr.20: 20369794-20369794 on Build GRCh38
rs751593238 Chr.20: 20369818-20369818 on Build GRCh38
rs1045559588 Chr.20: 20369717-20369717 on Build GRCh38
rs765907616 Chr.20: 20369485-20369485 on Build GRCh38
rs921689507 Chr.20: 20369824-20369824 on Build GRCh38
rs779818524 Chr.20: 20369772-20369772 on Build GRCh38
rs759690264 Chr.20: 20369712-20369712 on Build GRCh38
rs953050469 Chr.20: 20369828-20369828 on Build GRCh38
rs765395519 Chr.20: 20369721-20369721 on Build GRCh38
rs202019561 Chr.20: 20369590-20369590 on Build GRCh38
rs370307027 Chr.20: 20369552-20369552 on Build GRCh38
rs764159487 Chr.20: 20369816-20369816 on Build GRCh38
rs767197930 Chr.20: 20369473-20369473 on Build GRCh38
rs745505871 Chr.20: 20369658-20369658 on Build GRCh38
rs778213243 Chr.20: 20369637-20369637 on Build GRCh38
rs770648671 Chr.20: 20369608-20369608 on Build GRCh38
rs772905117 Chr.20: 20369804-20369804 on Build GRCh38
rs762213940 Chr.20: 20369551-20369551 on Build GRCh38
rs753992828 Chr.20: 20369631-20369631 on Build GRCh38
rs752895152 Chr.20: 20369813-20369813 on Build GRCh38
rs774405537 Chr.20: 20369695-20369695 on Build GRCh38
rs771985771 Chr.20: 20369800-20369800 on Build GRCh38
rs768267922 Chr.20: 20369782-20369782 on Build GRCh38
rs142730946 Chr.20: 20369711-20369711 on Build GRCh38
rs775922193 Chr.20: 20369443-20369443 on Build GRCh38
rs150719975 Chr.20: 20369788-20369788 on Build GRCh38
rs749274733 Chr.20: 20369532-20369532 on Build GRCh38
rs201280847 Chr.20: 20369771-20369771 on Build GRCh38
rs200889143 Chr.20: 20369629-20369629 on Build GRCh38
rs61733922 Chr.20: 20369743-20369743 on Build GRCh38
rs746191549 Chr.20: 20369758-20369758 on Build GRCh38
rs774491068 Chr.20: 20369829-20369830 on Build GRCh38
rs752807301 Chr.20: 20369729-20369729 on Build GRCh38
rs370866542 Chr.20: 20369757-20369757 on Build GRCh38
rs759139506 Chr.20: 20369610-20369610 on Build GRCh38
rs754959370 Chr.20: 20369500-20369500 on Build GRCh38
rs756520428 Chr.20: 20369831-20369831 on Build GRCh38
rs987077866 Chr.20: 20369834-20369834 on Build GRCh38
rs750791305 Chr.20: 20369825-20369825 on Build GRCh38
rs750055006 Chr.20: 20369475-20369475 on Build GRCh38
rs754798154 Chr.20: 20369402-20369402 on Build GRCh38
rs374207914 Chr.20: 20369787-20369787 on Build GRCh38
rs143026654 Chr.20: 20369697-20369697 on Build GRCh38
rs781693483 Chr.20: 20369822-20369822 on Build GRCh38
rs377333005 Chr.20: 20369399-20369399 on Build GRCh38
rs765891839 Chr.20: 20369556-20369556 on Build GRCh38
rs572384565 Chr.20: 20369626-20369626 on Build GRCh38
rs556795808 Chr.20: 20369563-20369563 on Build GRCh38
rs1051936496 Chr.20: 20369649-20369649 on Build GRCh38
rs528952402 Chr.20: 20369780-20369780 on Build GRCh38
rs963034856 Chr.20: 20369660-20369660 on Build GRCh38
rs745989349 Chr.20: 20369425-20369425 on Build GRCh38
rs758240556 Chr.20: 20369507-20369507 on Build GRCh38
rs768984339 Chr.20: 20369451-20369451 on Build GRCh38
rs770072780 Chr.20: 20369528-20369528 on Build GRCh38
rs751338296 Chr.20: 20369571-20369571 on Build GRCh38
rs762509361 Chr.20: 20369675-20369675 on Build GRCh38
rs778302937 Chr.20: 20369735-20369735 on Build GRCh38
rs1052446890 Chr.20: 20369478-20369478 on Build GRCh38
rs776407227 Chr.20: 20369653-20369653 on Build GRCh38
rs758929075 Chr.20: 20369636-20369636 on Build GRCh38
rs761725201 Chr.20: 20369696-20369696 on Build GRCh38
rs762979020 Chr.20: 20369533-20369550 on Build GRCh38
rs752000823 Chr.20: 20369741-20369741 on Build GRCh38
rs1023628563 Chr.20: 20369514-20369514 on Build GRCh38
rs1004580925 Chr.20: 20369648-20369648 on Build GRCh38
rs775425291 Chr.20: 20369837-20369837 on Build GRCh38
rs772498911 Chr.20: 20369602-20369602 on Build GRCh38
rs755589160 Chr.20: 20369396-20369396 on Build GRCh38
rs200467355 Chr.20: 20369805-20369805 on Build GRCh38

View Full Product Details