Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 21396265-21396769 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NKX2-4
Entrez Gene ID
644524
Gene Name
NK2 homeobox 4
Gene Aliases
NKX2.4, NKX2D
UniGene
Hs.456662
Gene Chromosome Location
Chr.20: 21395367-21397409 on Build GRCh38
Transcripts
NM_033176.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5479820 c.472G>A p.V158M Chr.20: 21396504-21396504 on Build GRCh38
4097070 c.555A>G p.A185A Chr.20: 21396421-21396421 on Build GRCh38
6726622 c.609G>T p.E203D Chr.20: 21396367-21396367 on Build GRCh38
7292625 c.627G>A p.K209K Chr.20: 21396349-21396349 on Build GRCh38
6726625 c.689C>A p.P230H Chr.20: 21396287-21396287 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767040814 Chr.20: 21396318-21396318 on Build GRCh38
rs774228726 Chr.20: 21396369-21396369 on Build GRCh38
rs982777448 Chr.20: 21396729-21396729 on Build GRCh38
rs941608741 Chr.20: 21396741-21396741 on Build GRCh38
rs761739260 Chr.20: 21396376-21396376 on Build GRCh38
rs910157925 Chr.20: 21396730-21396730 on Build GRCh38
rs371741137 Chr.20: 21396343-21396343 on Build GRCh38
rs759268126 Chr.20: 21396474-21396474 on Build GRCh38
rs910125301 Chr.20: 21396574-21396574 on Build GRCh38
rs951444834 Chr.20: 21396728-21396728 on Build GRCh38
rs1009503627 Chr.20: 21396355-21396355 on Build GRCh38
rs563976636 Chr.20: 21396423-21396423 on Build GRCh38
rs752135229 Chr.20: 21396336-21396336 on Build GRCh38
rs921447872 Chr.20: 21396750-21396750 on Build GRCh38
rs771512316 Chr.20: 21396388-21396388 on Build GRCh38
rs893038353 Chr.20: 21396676-21396676 on Build GRCh38
rs1032934149 Chr.20: 21396442-21396442 on Build GRCh38
rs750913793 Chr.20: 21396494-21396494 on Build GRCh38
rs756192019 Chr.20: 21396438-21396438 on Build GRCh38
rs768039831 Chr.20: 21396487-21396487 on Build GRCh38
rs780210342 Chr.20: 21396357-21396357 on Build GRCh38
rs199747643 Chr.20: 21396445-21396445 on Build GRCh38
rs779172479 Chr.20: 21396552-21396552 on Build GRCh38
rs899972996 Chr.20: 21396579-21396579 on Build GRCh38
rs747994584 Chr.20: 21396454-21396454 on Build GRCh38
rs562318143 Chr.20: 21396507-21396507 on Build GRCh38
rs764499372 Chr.20: 21396477-21396477 on Build GRCh38
rs766005689 Chr.20: 21396328-21396328 on Build GRCh38
rs902859213 Chr.20: 21396642-21396642 on Build GRCh38
rs1000412367 Chr.20: 21396419-21396419 on Build GRCh38
rs983037795 Chr.20: 21396573-21396573 on Build GRCh38
rs970565836 Chr.20: 21396600-21396600 on Build GRCh38
rs761260962 Chr.20: 21396307-21396307 on Build GRCh38
rs750515308 Chr.20: 21396436-21396436 on Build GRCh38
rs753099139 Chr.20: 21396331-21396331 on Build GRCh38
rs768459805 Chr.20: 21396364-21396364 on Build GRCh38
rs781429920 Chr.20: 21396347-21396347 on Build GRCh38
rs770046656 Chr.20: 21396352-21396352 on Build GRCh38
rs757856518 Chr.20: 21396430-21396430 on Build GRCh38
rs548068586 Chr.20: 21396653-21396653 on Build GRCh38
rs760166948 Chr.20: 21396391-21396391 on Build GRCh38
rs755178877 Chr.20: 21396550-21396550 on Build GRCh38
rs527585095 Chr.20: 21396708-21396708 on Build GRCh38
rs971613677 Chr.20: 21396695-21396695 on Build GRCh38
rs765334447 Chr.20: 21396598-21396598 on Build GRCh38
rs941546870 Chr.20: 21396576-21396581 on Build GRCh38
rs531435005 Chr.20: 21396632-21396632 on Build GRCh38
rs528209481 Chr.20: 21396482-21396482 on Build GRCh38
rs1039875551 Chr.20: 21396577-21396577 on Build GRCh38
rs982574809 Chr.20: 21396518-21396518 on Build GRCh38
rs1022765099 Chr.20: 21396691-21396691 on Build GRCh38
rs1012733378 Chr.20: 21396684-21396685 on Build GRCh38
rs757373462 Chr.20: 21396344-21396344 on Build GRCh38
rs755804118 Chr.20: 21396326-21396326 on Build GRCh38
rs753662642 Chr.20: 21396383-21396383 on Build GRCh38
rs753998000 Chr.20: 21396511-21396511 on Build GRCh38
rs934121253 Chr.20: 21396751-21396751 on Build GRCh38
rs535861369 Chr.20: 21396439-21396439 on Build GRCh38
rs998788152 Chr.20: 21396463-21396463 on Build GRCh38
rs375642949 Chr.20: 21396319-21396319 on Build GRCh38
rs759442145 Chr.20: 21396509-21396509 on Build GRCh38
rs773060972 Chr.20: 21396458-21396458 on Build GRCh38
rs1051249551 Chr.20: 21396633-21396633 on Build GRCh38
rs879444172 Chr.20: 21396581-21396581 on Build GRCh38
rs1032930757 Chr.20: 21396658-21396658 on Build GRCh38
rs568059393 Chr.20: 21396703-21396703 on Build GRCh38
rs916687741 Chr.20: 21396504-21396504 on Build GRCh38
rs756175522 Chr.20: 21396496-21396496 on Build GRCh38
rs539897092 Chr.20: 21396380-21396380 on Build GRCh38
rs774318763 Chr.20: 21396303-21396303 on Build GRCh38
rs755976718 Chr.20: 21396421-21396422 on Build GRCh38
rs371025178 Chr.20: 21396299-21396299 on Build GRCh38
rs763362678 Chr.20: 21396402-21396402 on Build GRCh38
rs746024545 Chr.20: 21396350-21396350 on Build GRCh38
rs929927073 Chr.20: 21396558-21396558 on Build GRCh38
rs776307948 Chr.20: 21396469-21396469 on Build GRCh38
rs998919957 Chr.20: 21396654-21396654 on Build GRCh38
rs975640431 Chr.20: 21396743-21396743 on Build GRCh38
rs1047321331 Chr.20: 21396395-21396395 on Build GRCh38
rs958996815 Chr.20: 21396443-21396443 on Build GRCh38
rs936040345 Chr.20: 21396707-21396707 on Build GRCh38
rs772765676 Chr.20: 21396390-21396390 on Build GRCh38
rs868252659 Chr.20: 21396526-21396526 on Build GRCh38
rs770372296 Chr.20: 21396462-21396462 on Build GRCh38
rs888617221 Chr.20: 21396411-21396411 on Build GRCh38
rs760208755 Chr.20: 21396460-21396460 on Build GRCh38
rs967308951 Chr.20: 21396297-21396297 on Build GRCh38
rs1046373530 Chr.20: 21396358-21396358 on Build GRCh38
rs560099970 Chr.20: 21396401-21396401 on Build GRCh38
rs370549869 Chr.20: 21396413-21396413 on Build GRCh38
rs745626916 Chr.20: 21396422-21396424 on Build GRCh38
rs545902466 Chr.20: 21396471-21396471 on Build GRCh38
rs933775431 Chr.20: 21396603-21396603 on Build GRCh38
rs752982670 Chr.20: 21396667-21396667 on Build GRCh38
rs751963549 Chr.20: 21396478-21396478 on Build GRCh38
rs367598502 Chr.20: 21396332-21396332 on Build GRCh38
rs1020428813 Chr.20: 21396370-21396370 on Build GRCh38

View Full Product Details