Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 36612424-36612939 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C20orf24
Entrez Gene ID
55969
Gene Name
chromosome 20 open reading frame 24
Gene Aliases
PNAS-11, RIP5
UniGene
Hs.584985
Gene Chromosome Location
Chr.20: 36605734-36612557 on Build GRCh38
Transcripts
NM_001199534.1 NM_018840.4 NM_199483.2 NR_026562.3 NM_032214.3 NM_175077.2 NM_001199535.1

Gene Symbol
SLA2
Entrez Gene ID
84174
Gene Name
Src like adaptor 2
Gene Aliases
C20orf156, MARS, SLAP-2, SLAP2
UniGene
Hs.713578
Gene Chromosome Location
Chr.20: 36612519-36646216 on Build GRCh38
Transcripts
NM_001199534.1 NM_018840.4 NM_199483.2 NR_026562.3 NM_032214.3 NM_175077.2 NM_001199535.1

Gene Symbol
TGIF2-C20orf24
Entrez Gene ID
100527943
Gene Name
TGIF2-C20orf24 readthrough
Gene Aliases
-
UniGene
Hs.584985
Gene Chromosome Location
Chr.20: 36574554-36612557 on Build GRCh38
Transcripts
NM_001199534.1 NM_018840.4 NM_199483.2 NR_026562.3 NM_032214.3 NM_175077.2 NM_001199535.1

SNP Detail

SNP ID Chromosome Location
rs981840256 Chr.20: 36612458-36612458 on Build GRCh38
rs1013488219 Chr.20: 36612755-36612755 on Build GRCh38
rs11474169 Chr.20: 36612853-36612854 on Build GRCh38
rs185853679 Chr.20: 36612677-36612677 on Build GRCh38
rs766609435 Chr.20: 36612464-36612464 on Build GRCh38
rs777018235 Chr.20: 36612812-36612812 on Build GRCh38
rs540364406 Chr.20: 36612704-36612704 on Build GRCh38
rs1048315711 Chr.20: 36612852-36612852 on Build GRCh38
rs997986588 Chr.20: 36612613-36612613 on Build GRCh38
rs894981768 Chr.20: 36612691-36612692 on Build GRCh38
rs544579547 Chr.20: 36612822-36612822 on Build GRCh38
rs899574267 Chr.20: 36612612-36612612 on Build GRCh38
rs1036629718 Chr.20: 36612841-36612842 on Build GRCh38
rs995451760 Chr.20: 36612571-36612571 on Build GRCh38
rs1046291394 Chr.20: 36612874-36612874 on Build GRCh38
rs1031020773 Chr.20: 36612717-36612718 on Build GRCh38
rs897066451 Chr.20: 36612564-36612564 on Build GRCh38
rs924703863 Chr.20: 36612776-36612776 on Build GRCh38
rs893746487 Chr.20: 36612540-36612540 on Build GRCh38
rs1039942983 Chr.20: 36612836-36612836 on Build GRCh38
rs906586863 Chr.20: 36612905-36612905 on Build GRCh38
rs970824537 Chr.20: 36612702-36612702 on Build GRCh38
rs146777412 Chr.20: 36612849-36612850 on Build GRCh38
rs931183874 Chr.20: 36612849-36612850 on Build GRCh38
rs367865670 Chr.20: 36612678-36612678 on Build GRCh38
rs1049500036 Chr.20: 36612484-36612484 on Build GRCh38
rs7870 Chr.20: 36612525-36612525 on Build GRCh38
rs776941087 Chr.20: 36612548-36612548 on Build GRCh38
rs1039570243 Chr.20: 36612553-36612553 on Build GRCh38
rs1017115209 Chr.20: 36612726-36612726 on Build GRCh38
rs956002379 Chr.20: 36612791-36612791 on Build GRCh38
rs971885051 Chr.20: 36612770-36612770 on Build GRCh38
rs990173238 Chr.20: 36612800-36612800 on Build GRCh38
rs376471373 Chr.20: 36612451-36612451 on Build GRCh38
rs946691703 Chr.20: 36612817-36612817 on Build GRCh38
rs759897290 Chr.20: 36612581-36612581 on Build GRCh38
rs962955008 Chr.20: 36612754-36612754 on Build GRCh38
rs191538269 Chr.20: 36612763-36612763 on Build GRCh38
rs11643 Chr.20: 36612500-36612500 on Build GRCh38
rs533434475 Chr.20: 36612898-36612898 on Build GRCh38
rs912295521 Chr.20: 36612798-36612798 on Build GRCh38
rs999512976 Chr.20: 36612709-36612709 on Build GRCh38
rs981515787 Chr.20: 36612772-36612772 on Build GRCh38
rs1012713128 Chr.20: 36612692-36612695 on Build GRCh38
rs370482514 Chr.20: 36612775-36612775 on Build GRCh38
rs933824117 Chr.20: 36612875-36612875 on Build GRCh38
rs572240742 Chr.20: 36612697-36612697 on Build GRCh38
rs555946900 Chr.20: 36612722-36612722 on Build GRCh38
rs11540276 Chr.20: 36612449-36612449 on Build GRCh38
rs895090303 Chr.20: 36612854-36612855 on Build GRCh38
rs1036681930 Chr.20: 36612535-36612535 on Build GRCh38
rs182921495 Chr.20: 36612873-36612873 on Build GRCh38
rs765673786 Chr.20: 36612841-36612841 on Build GRCh38
rs958508020 Chr.20: 36612744-36612744 on Build GRCh38

View Full Product Details