Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 38318016-38318526 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BPI
Entrez Gene ID
671
Gene Name
bactericidal/permeability-increasing protein
Gene Aliases
BPIFD1, rBPI
UniGene
Hs.529019
Gene Chromosome Location
Chr.20: 38304150-38337503 on Build GRCh38
Transcripts
NM_001725.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1026637 c.632A>C p.N211T Chr.20: 38318432-38318432 on Build GRCh38
4291871 c.658T>G p.Y220D Chr.20: 38318458-38318458 on Build GRCh38
149171 c.646G>A p.E216K Chr.20: 38318446-38318446 on Build GRCh38
1246192 c.635C>T p.S212F Chr.20: 38318435-38318435 on Build GRCh38
723762 c.641C>T p.S214F Chr.20: 38318441-38318441 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765514981 Chr.20: 38318456-38318456 on Build GRCh38
rs993719783 Chr.20: 38318152-38318152 on Build GRCh38
rs545033858 Chr.20: 38318185-38318185 on Build GRCh38
rs952169653 Chr.20: 38318183-38318183 on Build GRCh38
rs371706866 Chr.20: 38318392-38318392 on Build GRCh38
rs943718957 Chr.20: 38318344-38318344 on Build GRCh38
rs1049362026 Chr.20: 38318064-38318064 on Build GRCh38
rs780110388 Chr.20: 38318172-38318172 on Build GRCh38
rs375120673 Chr.20: 38318458-38318458 on Build GRCh38
rs750867799 Chr.20: 38318366-38318369 on Build GRCh38
rs372552649 Chr.20: 38318435-38318435 on Build GRCh38
rs1007878464 Chr.20: 38318129-38318129 on Build GRCh38
rs985283723 Chr.20: 38318500-38318500 on Build GRCh38
rs912300915 Chr.20: 38318329-38318329 on Build GRCh38
rs932436716 Chr.20: 38318283-38318283 on Build GRCh38
rs913862129 Chr.20: 38318236-38318236 on Build GRCh38
rs767576684 Chr.20: 38318390-38318390 on Build GRCh38
rs1019235941 Chr.20: 38318134-38318134 on Build GRCh38
rs577566493 Chr.20: 38318124-38318124 on Build GRCh38
rs528824628 Chr.20: 38318267-38318267 on Build GRCh38
rs140498581 Chr.20: 38318244-38318244 on Build GRCh38
rs775397439 Chr.20: 38318439-38318439 on Build GRCh38
rs770298931 Chr.20: 38318493-38318493 on Build GRCh38
rs1033656904 Chr.20: 38318199-38318199 on Build GRCh38
rs774090383 Chr.20: 38318434-38318434 on Build GRCh38
rs758585190 Chr.20: 38318466-38318466 on Build GRCh38
rs747360374 Chr.20: 38318476-38318476 on Build GRCh38
rs960725289 Chr.20: 38318208-38318208 on Build GRCh38
rs4358188 Chr.20: 38318446-38318446 on Build GRCh38
rs747553107 Chr.20: 38318428-38318428 on Build GRCh38
rs373059376 Chr.20: 38318347-38318347 on Build GRCh38
rs753842705 Chr.20: 38318406-38318406 on Build GRCh38
rs757099741 Chr.20: 38318176-38318176 on Build GRCh38
rs767382698 Chr.20: 38318438-38318438 on Build GRCh38
rs144221701 Chr.20: 38318418-38318418 on Build GRCh38
rs777950714 Chr.20: 38318399-38318399 on Build GRCh38
rs950774415 Chr.20: 38318325-38318325 on Build GRCh38
rs1049240222 Chr.20: 38318388-38318388 on Build GRCh38
rs780977947 Chr.20: 38318277-38318277 on Build GRCh38
rs757222022 Chr.20: 38318455-38318455 on Build GRCh38
rs57987577 Chr.20: 38318133-38318133 on Build GRCh38
rs757561111 Chr.20: 38318413-38318413 on Build GRCh38
rs887917136 Chr.20: 38318410-38318410 on Build GRCh38
rs903605437 Chr.20: 38318041-38318041 on Build GRCh38
rs111327200 Chr.20: 38318473-38318473 on Build GRCh38
rs954042070 Chr.20: 38318486-38318486 on Build GRCh38
rs5741802 Chr.20: 38318353-38318353 on Build GRCh38
rs377613758 Chr.20: 38318109-38318121 on Build GRCh38
rs369264578 Chr.20: 38318430-38318430 on Build GRCh38
rs992114980 Chr.20: 38318214-38318214 on Build GRCh38
rs774275266 Chr.20: 38318312-38318312 on Build GRCh38
rs953956725 Chr.20: 38318271-38318271 on Build GRCh38
rs919250149 Chr.20: 38318324-38318324 on Build GRCh38
rs765934187 Chr.20: 38318375-38318375 on Build GRCh38
rs748844520 Chr.20: 38318487-38318487 on Build GRCh38
rs770622158 Chr.20: 38318432-38318432 on Build GRCh38
rs150340828 Chr.20: 38318483-38318483 on Build GRCh38
rs367567438 Chr.20: 38318380-38318380 on Build GRCh38
rs932359473 Chr.20: 38318083-38318083 on Build GRCh38
rs537140348 Chr.20: 38318288-38318288 on Build GRCh38
rs948916093 Chr.20: 38318080-38318080 on Build GRCh38
rs145662699 Chr.20: 38318301-38318301 on Build GRCh38
rs529472050 Chr.20: 38318365-38318365 on Build GRCh38
rs938425272 Chr.20: 38318401-38318401 on Build GRCh38
rs772564423 Chr.20: 38318363-38318363 on Build GRCh38
rs1049436677 Chr.20: 38318122-38318122 on Build GRCh38
rs1044553062 Chr.20: 38318120-38318121 on Build GRCh38
rs909355491 Chr.20: 38318065-38318065 on Build GRCh38
rs981233451 Chr.20: 38318190-38318190 on Build GRCh38
rs780568663 Chr.20: 38318426-38318426 on Build GRCh38
rs151033582 Chr.20: 38318429-38318429 on Build GRCh38
rs754020896 Chr.20: 38318454-38318454 on Build GRCh38
rs751466835 Chr.20: 38318379-38318379 on Build GRCh38
rs746774852 Chr.20: 38318147-38318147 on Build GRCh38
rs151215961 Chr.20: 38318445-38318445 on Build GRCh38
rs909435010 Chr.20: 38318376-38318376 on Build GRCh38
rs772370916 Chr.20: 38318419-38318419 on Build GRCh38
rs376281638 Chr.20: 38318398-38318398 on Build GRCh38

View Full Product Details