Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 84181896-84182430 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRKACB
Entrez Gene ID
5567
Gene Name
protein kinase cAMP-activated catalytic subunit beta
Gene Aliases
PKA C-beta, PKACB
UniGene
Hs.487325
Gene Chromosome Location
Chr.1: 84077975-84238498 on Build GRCh38
Transcripts
NM_001242857.2 NM_001242858.2 NM_001242859.2 NM_001242860.2 NM_001242861.2 NM_001242862.2 NM_001300915.1 NM_001300916.1 NM_001300917.1 NM_002731.3 NM_182948.3 NM_207578.2 XM_005271015.3 XM_005271016.1 XM_005271017.3 XM_005271018.4 XM_005271019.1 XM_005271020.1 XM_006710758.1 XM_017001706.1 XM_017001707.1 XM_017001708.1 XM_017001709.1 XM_017001710.1 XM_017001711.1 XM_017001712.1 XM_017001713.1 XM_017001714.1 XM_017001715.1 XM_017001716.1 XM_017001717.1 XM_017001718.1 XM_017001719.1 XM_017001720.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2243532 c.172G>T p.G58* Chr.1: 84182242-84182242 on Build GRCh38
2243536 c.173G>T p.G58V Chr.1: 84182243-84182243 on Build GRCh38
6451174 c.166A>C p.T56P Chr.1: 84182236-84182236 on Build GRCh38
912543 c.341A>C p.E114A Chr.1: 84182291-84182291 on Build GRCh38
1602708 c.186A>G p.K62K Chr.1: 84182277-84182277 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750679516 Chr.1: 84182188-84182188 on Build GRCh38
rs755081600 Chr.1: 84182233-84182233 on Build GRCh38
rs767786398 Chr.1: 84182341-84182341 on Build GRCh38
rs746920926 Chr.1: 84182255-84182255 on Build GRCh38
rs960377698 Chr.1: 84182301-84182301 on Build GRCh38
rs376111738 Chr.1: 84182324-84182324 on Build GRCh38
rs1004795830 Chr.1: 84182048-84182048 on Build GRCh38
rs370005044 Chr.1: 84182208-84182208 on Build GRCh38
rs796108271 Chr.1: 84182386-84182386 on Build GRCh38
rs999565681 Chr.1: 84181959-84181959 on Build GRCh38
rs766750091 Chr.1: 84182363-84182363 on Build GRCh38
rs769500443 Chr.1: 84182297-84182297 on Build GRCh38
rs748843138 Chr.1: 84182328-84182328 on Build GRCh38
rs202240979 Chr.1: 84182169-84182169 on Build GRCh38
rs769302578 Chr.1: 84182160-84182160 on Build GRCh38
rs773772175 Chr.1: 84182352-84182352 on Build GRCh38
rs762106128 Chr.1: 84182184-84182184 on Build GRCh38
rs752082954 Chr.1: 84182173-84182173 on Build GRCh38
rs774750660 Chr.1: 84182334-84182334 on Build GRCh38
rs770632163 Chr.1: 84182266-84182266 on Build GRCh38
rs956517305 Chr.1: 84182241-84182241 on Build GRCh38
rs373856613 Chr.1: 84182056-84182056 on Build GRCh38
rs754495455 Chr.1: 84181929-84181929 on Build GRCh38
rs759118703 Chr.1: 84182154-84182154 on Build GRCh38
rs1023061545 Chr.1: 84182142-84182142 on Build GRCh38
rs566538238 Chr.1: 84182164-84182164 on Build GRCh38
rs151323322 Chr.1: 84182250-84182250 on Build GRCh38
rs185944690 Chr.1: 84182361-84182361 on Build GRCh38
rs780979027 Chr.1: 84182292-84182292 on Build GRCh38
rs762315361 Chr.1: 84182339-84182339 on Build GRCh38
rs752082511 Chr.1: 84182381-84182381 on Build GRCh38
rs994472895 Chr.1: 84182029-84182029 on Build GRCh38
rs370687949 Chr.1: 84182185-84182185 on Build GRCh38
rs11552595 Chr.1: 84182256-84182256 on Build GRCh38
rs903961953 Chr.1: 84181958-84181958 on Build GRCh38
rs375692509 Chr.1: 84182088-84182088 on Build GRCh38
rs1044325853 Chr.1: 84182368-84182368 on Build GRCh38
rs916847700 Chr.1: 84182360-84182360 on Build GRCh38
rs896336937 Chr.1: 84182105-84182105 on Build GRCh38
rs891134594 Chr.1: 84182039-84182039 on Build GRCh38
rs775091017 Chr.1: 84182319-84182319 on Build GRCh38
rs371980693 Chr.1: 84182046-84182046 on Build GRCh38
rs181366824 Chr.1: 84182090-84182090 on Build GRCh38
rs569145388 Chr.1: 84182369-84182369 on Build GRCh38
rs754040223 Chr.1: 84182226-84182226 on Build GRCh38
rs535896034 Chr.1: 84182161-84182161 on Build GRCh38
rs374245973 Chr.1: 84181974-84181974 on Build GRCh38
rs745646679 Chr.1: 84182295-84182295 on Build GRCh38
rs1012055954 Chr.1: 84182125-84182125 on Build GRCh38
rs935467846 Chr.1: 84182406-84182406 on Build GRCh38
rs148797911 Chr.1: 84182209-84182209 on Build GRCh38
rs768162523 Chr.1: 84182329-84182329 on Build GRCh38
rs1052419344 Chr.1: 84182006-84182006 on Build GRCh38
rs996435196 Chr.1: 84182114-84182114 on Build GRCh38
rs775036025 Chr.1: 84182163-84182163 on Build GRCh38

View Full Product Details