Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 84554821-84555318 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTBS
Entrez Gene ID
1486
Gene Name
chitobiase
Gene Aliases
CTB
UniGene
Hs.513557
Gene Chromosome Location
Chr.1: 84553121-84574480 on Build GRCh38
Transcripts
NM_004388.2 NM_001310156.1 XM_011542514.2

Gene Symbol
SPATA1
Entrez Gene ID
100505741
Gene Name
spermatogenesis associated 1
Gene Aliases
SP-2, SPAP1
UniGene
Hs.584956
Gene Chromosome Location
Chr.1: 84506301-84556495 on Build GRCh38
Transcripts
NM_004388.2 NM_001310156.1 XM_011542514.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5899814 c.1114G>A p.E372K Chr.1: 84555043-84555043 on Build GRCh38
912584 c.962C>A p.P321H Chr.1: 84555195-84555195 on Build GRCh38
1249364 c.1157G>A p.*386* Chr.1: 84555000-84555000 on Build GRCh38
6637184 c.1039C>T p.R347C Chr.1: 84555118-84555118 on Build GRCh38
4601634 c.976C>T p.H326Y Chr.1: 84555181-84555181 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs11801891 Chr.1: 84554954-84554954 on Build GRCh38
rs1034087870 Chr.1: 84555125-84555125 on Build GRCh38
rs754652720 Chr.1: 84554991-84554991 on Build GRCh38
rs566173961 Chr.1: 84554861-84554861 on Build GRCh38
rs139699803 Chr.1: 84555095-84555095 on Build GRCh38
rs147594119 Chr.1: 84555112-84555112 on Build GRCh38
rs138201181 Chr.1: 84555000-84555000 on Build GRCh38
rs779324470 Chr.1: 84555232-84555232 on Build GRCh38
rs138861244 Chr.1: 84555086-84555086 on Build GRCh38
rs760513098 Chr.1: 84554981-84554985 on Build GRCh38
rs765136882 Chr.1: 84555144-84555144 on Build GRCh38
rs561994963 Chr.1: 84555204-84555204 on Build GRCh38
rs990096420 Chr.1: 84554892-84554892 on Build GRCh38
rs757321200 Chr.1: 84555206-84555206 on Build GRCh38
rs757973408 Chr.1: 84555078-84555078 on Build GRCh38
rs113788476 Chr.1: 84555087-84555087 on Build GRCh38
rs763056916 Chr.1: 84554992-84554992 on Build GRCh38
rs529294378 Chr.1: 84555108-84555108 on Build GRCh38
rs181215666 Chr.1: 84555249-84555249 on Build GRCh38
rs889980771 Chr.1: 84554981-84554981 on Build GRCh38
rs775151391 Chr.1: 84554950-84554950 on Build GRCh38
rs756813307 Chr.1: 84555171-84555171 on Build GRCh38
rs780468138 Chr.1: 84555223-84555223 on Build GRCh38
rs751093487 Chr.1: 84555168-84555168 on Build GRCh38
rs751010887 Chr.1: 84555189-84555189 on Build GRCh38
rs752461357 Chr.1: 84555071-84555071 on Build GRCh38
rs945962764 Chr.1: 84554929-84554929 on Build GRCh38
rs1043493 Chr.1: 84554966-84554966 on Build GRCh38
rs914381269 Chr.1: 84555286-84555286 on Build GRCh38
rs763997358 Chr.1: 84555020-84555021 on Build GRCh38
rs34160527 Chr.1: 84555012-84555012 on Build GRCh38
rs766985516 Chr.1: 84555175-84555175 on Build GRCh38
rs1025735721 Chr.1: 84555110-84555110 on Build GRCh38
rs774511903 Chr.1: 84555009-84555009 on Build GRCh38
rs772343723 Chr.1: 84555235-84555235 on Build GRCh38
rs1047754724 Chr.1: 84554917-84554917 on Build GRCh38
rs775088103 Chr.1: 84555145-84555145 on Build GRCh38
rs776668169 Chr.1: 84555130-84555130 on Build GRCh38
rs111674297 Chr.1: 84555015-84555015 on Build GRCh38
rs372730923 Chr.1: 84555276-84555278 on Build GRCh38
rs939303876 Chr.1: 84555273-84555275 on Build GRCh38
rs937152325 Chr.1: 84554873-84554873 on Build GRCh38
rs183916139 Chr.1: 84554947-84554947 on Build GRCh38
rs753705550 Chr.1: 84555048-84555048 on Build GRCh38
rs759385732 Chr.1: 84555135-84555135 on Build GRCh38
rs755271633 Chr.1: 84555229-84555229 on Build GRCh38
rs910725200 Chr.1: 84554881-84554881 on Build GRCh38
rs745831379 Chr.1: 84555240-84555240 on Build GRCh38
rs764908593 Chr.1: 84555052-84555052 on Build GRCh38
rs756570083 Chr.1: 84555211-84555211 on Build GRCh38
rs775834132 Chr.1: 84554971-84554971 on Build GRCh38
rs748114267 Chr.1: 84554936-84554936 on Build GRCh38
rs748349116 Chr.1: 84555233-84555233 on Build GRCh38
rs564541760 Chr.1: 84555121-84555121 on Build GRCh38
rs149668513 Chr.1: 84555120-84555120 on Build GRCh38
rs780281748 Chr.1: 84555117-84555117 on Build GRCh38
rs1037932172 Chr.1: 84554886-84554886 on Build GRCh38
rs766374860 Chr.1: 84555045-84555045 on Build GRCh38
rs141347801 Chr.1: 84555096-84555096 on Build GRCh38
rs966691223 Chr.1: 84555066-84555066 on Build GRCh38
rs772253736 Chr.1: 84555124-84555124 on Build GRCh38
rs193181499 Chr.1: 84554901-84554901 on Build GRCh38
rs199627584 Chr.1: 84555236-84555236 on Build GRCh38
rs943518172 Chr.1: 84554885-84554885 on Build GRCh38
rs746203845 Chr.1: 84554958-84554958 on Build GRCh38
rs763697037 Chr.1: 84555159-84555159 on Build GRCh38
rs768929698 Chr.1: 84555118-84555118 on Build GRCh38
rs753793114 Chr.1: 84555031-84555031 on Build GRCh38
rs762671358 Chr.1: 84555157-84555157 on Build GRCh38
rs756490636 Chr.1: 84555225-84555225 on Build GRCh38
rs534701919 Chr.1: 84554900-84554900 on Build GRCh38

View Full Product Details