Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 62464427-62464938 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GATA5
Entrez Gene ID
140628
Gene Name
GATA binding protein 5
Gene Aliases
GATAS, bB379O24.1
UniGene
Hs.352250
Gene Chromosome Location
Chr.20: 62463497-62475970 on Build GRCh38
Transcripts
NM_080473.4 XM_006723699.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6369556 c.1114C>T p.P372S Chr.20: 62464916-62464916 on Build GRCh38
1751544 c.1093G>C p.E365Q Chr.20: 62464937-62464937 on Build GRCh38
6380292 c.1099G>T p.E367* Chr.20: 62464931-62464931 on Build GRCh38
6558971 c.1140T>A p.A380A Chr.20: 62464890-62464890 on Build GRCh38
1740651 c.1115C>G p.P372R Chr.20: 62464915-62464915 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs565076349 Chr.20: 62464604-62464604 on Build GRCh38
rs376834986 Chr.20: 62464458-62464458 on Build GRCh38
rs1042753855 Chr.20: 62464581-62464581 on Build GRCh38
rs976067539 Chr.20: 62464484-62464484 on Build GRCh38
rs980332454 Chr.20: 62464719-62464719 on Build GRCh38
rs782631347 Chr.20: 62464856-62464856 on Build GRCh38
rs781885102 Chr.20: 62464820-62464820 on Build GRCh38
rs781950321 Chr.20: 62464877-62464877 on Build GRCh38
rs782208988 Chr.20: 62464912-62464912 on Build GRCh38
rs556479202 Chr.20: 62464549-62464549 on Build GRCh38
rs997556291 Chr.20: 62464611-62464611 on Build GRCh38
rs969932573 Chr.20: 62464683-62464683 on Build GRCh38
rs201124352 Chr.20: 62464810-62464810 on Build GRCh38
rs782799636 Chr.20: 62464789-62464789 on Build GRCh38
rs571597195 Chr.20: 62464759-62464759 on Build GRCh38
rs919363634 Chr.20: 62464485-62464485 on Build GRCh38
rs891587605 Chr.20: 62464572-62464572 on Build GRCh38
rs1052703368 Chr.20: 62464569-62464569 on Build GRCh38
rs6061550 Chr.20: 62464902-62464902 on Build GRCh38
rs544499909 Chr.20: 62464540-62464540 on Build GRCh38
rs1011273184 Chr.20: 62464573-62464573 on Build GRCh38
rs201116843 Chr.20: 62464903-62464903 on Build GRCh38
rs550087773 Chr.20: 62464729-62464729 on Build GRCh38
rs112617105 Chr.20: 62464865-62464865 on Build GRCh38
rs201914738 Chr.20: 62464786-62464786 on Build GRCh38
rs148724629 Chr.20: 62464881-62464881 on Build GRCh38
rs781791817 Chr.20: 62464899-62464899 on Build GRCh38
rs782345116 Chr.20: 62464855-62464855 on Build GRCh38
rs782452663 Chr.20: 62464900-62464900 on Build GRCh38
rs112955419 Chr.20: 62464546-62464546 on Build GRCh38
rs782541275 Chr.20: 62464876-62464877 on Build GRCh38
rs200199777 Chr.20: 62464851-62464851 on Build GRCh38
rs928516308 Chr.20: 62464557-62464557 on Build GRCh38
rs782198035 Chr.20: 62464852-62464852 on Build GRCh38
rs782493007 Chr.20: 62464669-62464669 on Build GRCh38
rs111825703 Chr.20: 62464720-62464720 on Build GRCh38
rs950774790 Chr.20: 62464508-62464508 on Build GRCh38
rs183849000 Chr.20: 62464463-62464463 on Build GRCh38
rs1026309521 Chr.20: 62464614-62464614 on Build GRCh38
rs782818698 Chr.20: 62464890-62464890 on Build GRCh38
rs782451619 Chr.20: 62464809-62464809 on Build GRCh38
rs782759156 Chr.20: 62464857-62464857 on Build GRCh38
rs145205240 Chr.20: 62464871-62464871 on Build GRCh38
rs527750293 Chr.20: 62464593-62464593 on Build GRCh38
rs555888990 Chr.20: 62464493-62464493 on Build GRCh38
rs781786523 Chr.20: 62464802-62464802 on Build GRCh38
rs948517365 Chr.20: 62464524-62464524 on Build GRCh38
rs782641854 Chr.20: 62464817-62464817 on Build GRCh38
rs1044340339 Chr.20: 62464539-62464539 on Build GRCh38
rs782511379 Chr.20: 62464815-62464815 on Build GRCh38
rs782550994 Chr.20: 62464823-62464823 on Build GRCh38
rs145330430 Chr.20: 62464870-62464870 on Build GRCh38
rs938597256 Chr.20: 62464754-62464754 on Build GRCh38
rs943573933 Chr.20: 62464859-62464859 on Build GRCh38
rs73149261 Chr.20: 62464606-62464606 on Build GRCh38
rs112195861 Chr.20: 62464564-62464564 on Build GRCh38
rs145474602 Chr.20: 62464476-62464476 on Build GRCh38
rs947015402 Chr.20: 62464840-62464840 on Build GRCh38
rs950660661 Chr.20: 62464650-62464650 on Build GRCh38
rs560305163 Chr.20: 62464586-62464586 on Build GRCh38
rs531578994 Chr.20: 62464626-62464626 on Build GRCh38
rs1016735947 Chr.20: 62464663-62464663 on Build GRCh38
rs782645578 Chr.20: 62464906-62464906 on Build GRCh38
rs782144120 Chr.20: 62464886-62464886 on Build GRCh38
rs782487182 Chr.20: 62464581-62464581 on Build GRCh38
rs781867987 Chr.20: 62464654-62464654 on Build GRCh38
rs573898029 Chr.20: 62464509-62464509 on Build GRCh38
rs782020634 Chr.20: 62464861-62464861 on Build GRCh38
rs966677113 Chr.20: 62464610-62464610 on Build GRCh38
rs151232305 Chr.20: 62464908-62464908 on Build GRCh38
rs148845867 Chr.20: 62464558-62464558 on Build GRCh38
rs909051373 Chr.20: 62464522-62464522 on Build GRCh38

View Full Product Details