Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 41243291-41243790 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BACE2
Entrez Gene ID
25825
Gene Name
beta-site APP-cleaving enzyme 2
Gene Aliases
AEPLC, ALP56, ASP1, ASP21, BAE2, CDA13, CEAP1, DRAP
UniGene
Hs.43047
Gene Chromosome Location
Chr.21: 41167801-41282534 on Build GRCh38
Transcripts
NM_012105.4 NM_138991.2 NM_138992.2 XM_017028314.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1471425 c.775T>A p.Y259N Chr.21: 41243403-41243403 on Build GRCh38
5587268 c.823C>A p.Q275K Chr.21: 41243451-41243451 on Build GRCh38
3740304 c.766C>T p.P256S Chr.21: 41243394-41243394 on Build GRCh38
1030840 c.750C>A p.V250V Chr.21: 41243378-41243378 on Build GRCh38
3550921 c.763G>A p.E255K Chr.21: 41243391-41243391 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs970090841 Chr.21: 41243607-41243607 on Build GRCh38
rs756984326 Chr.21: 41243634-41243634 on Build GRCh38
rs777818763 Chr.21: 41243446-41243446 on Build GRCh38
rs185479259 Chr.21: 41243520-41243520 on Build GRCh38
rs760226647 Chr.21: 41243529-41243529 on Build GRCh38
rs748155448 Chr.21: 41243541-41243550 on Build GRCh38
rs191388562 Chr.21: 41243522-41243522 on Build GRCh38
rs756102271 Chr.21: 41243435-41243435 on Build GRCh38
rs765795534 Chr.21: 41243351-41243351 on Build GRCh38
rs879828973 Chr.21: 41243582-41243582 on Build GRCh38
rs920718829 Chr.21: 41243614-41243614 on Build GRCh38
rs749152955 Chr.21: 41243464-41243464 on Build GRCh38
rs551807339 Chr.21: 41243620-41243620 on Build GRCh38
rs745593301 Chr.21: 41243505-41243505 on Build GRCh38
rs960698330 Chr.21: 41243551-41243551 on Build GRCh38
rs772962045 Chr.21: 41243612-41243612 on Build GRCh38
rs886727106 Chr.21: 41243347-41243347 on Build GRCh38
rs900690961 Chr.21: 41243701-41243701 on Build GRCh38
rs201436207 Chr.21: 41243550-41243550 on Build GRCh38
rs78770161 Chr.21: 41243727-41243727 on Build GRCh38
rs530813869 Chr.21: 41243404-41243404 on Build GRCh38
rs767249577 Chr.21: 41243436-41243436 on Build GRCh38
rs773834467 Chr.21: 41243545-41243545 on Build GRCh38
rs566764846 Chr.21: 41243580-41243580 on Build GRCh38
rs983510944 Chr.21: 41243659-41243659 on Build GRCh38
rs140187448 Chr.21: 41243626-41243626 on Build GRCh38
rs75038777 Chr.21: 41243732-41243732 on Build GRCh38
rs758756856 Chr.21: 41243392-41243392 on Build GRCh38
rs369361949 Chr.21: 41243373-41243373 on Build GRCh38
rs187781767 Chr.21: 41243583-41243583 on Build GRCh38
rs763414581 Chr.21: 41243371-41243371 on Build GRCh38
rs903638045 Chr.21: 41243719-41243719 on Build GRCh38
rs182763933 Chr.21: 41243566-41243566 on Build GRCh38
rs1043484986 Chr.21: 41243710-41243710 on Build GRCh38
rs781499365 Chr.21: 41243426-41243426 on Build GRCh38
rs763731426 Chr.21: 41243555-41243555 on Build GRCh38
rs755265850 Chr.21: 41243411-41243411 on Build GRCh38
rs768019666 Chr.21: 41243538-41243538 on Build GRCh38
rs73222274 Chr.21: 41243667-41243667 on Build GRCh38
rs910395703 Chr.21: 41243714-41243714 on Build GRCh38
rs896049040 Chr.21: 41243410-41243410 on Build GRCh38
rs878977136 Chr.21: 41243407-41243407 on Build GRCh38
rs751978125 Chr.21: 41243556-41243556 on Build GRCh38
rs778583641 Chr.21: 41243503-41243503 on Build GRCh38
rs944948480 Chr.21: 41243690-41243690 on Build GRCh38
rs1040536615 Chr.21: 41243692-41243692 on Build GRCh38
rs904537893 Chr.21: 41243350-41243350 on Build GRCh38
rs932162775 Chr.21: 41243703-41243703 on Build GRCh38
rs371362113 Chr.21: 41243430-41243430 on Build GRCh38
rs778816118 Chr.21: 41243533-41243534 on Build GRCh38
rs568809796 Chr.21: 41243639-41243639 on Build GRCh38
rs770828662 Chr.21: 41243499-41243499 on Build GRCh38
rs2252576 Chr.21: 41243366-41243366 on Build GRCh38

View Full Product Details