Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 44455245-44455779 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC3
Entrez Gene ID
81543
Gene Name
leucine rich repeat containing 3
Gene Aliases
C21orf102
UniGene
Hs.326579
Gene Chromosome Location
Chr.21: 44455486-44458856 on Build GRCh38
Transcripts
NM_030891.4 NR_047476.1

Gene Symbol
LRRC3-AS1
Entrez Gene ID
100861510
Gene Name
LRRC3 antisense RNA 1 (head to head)
Gene Aliases
-
UniGene
Hs.505254
Gene Chromosome Location
Chr.21: 44450986-44455284 on Build GRCh38
Transcripts
NM_030891.4 NR_047476.1

SNP Detail

SNP ID Chromosome Location
rs970437215 Chr.21: 44455693-44455693 on Build GRCh38
rs900633502 Chr.21: 44455478-44455478 on Build GRCh38
rs185293803 Chr.21: 44455420-44455420 on Build GRCh38
rs1034662877 Chr.21: 44455515-44455515 on Build GRCh38
rs202096965 Chr.21: 44455556-44455556 on Build GRCh38
rs936086706 Chr.21: 44455426-44455426 on Build GRCh38
rs1023429273 Chr.21: 44455284-44455294 on Build GRCh38
rs940640408 Chr.21: 44455655-44455655 on Build GRCh38
rs896277287 Chr.21: 44455700-44455700 on Build GRCh38
rs974781171 Chr.21: 44455600-44455600 on Build GRCh38
rs1035760423 Chr.21: 44455709-44455709 on Build GRCh38
rs973921251 Chr.21: 44455558-44455558 on Build GRCh38
rs553172852 Chr.21: 44455471-44455471 on Build GRCh38
rs946212393 Chr.21: 44455390-44455390 on Build GRCh38
rs956137897 Chr.21: 44455721-44455721 on Build GRCh38
rs572240748 Chr.21: 44455476-44455476 on Build GRCh38
rs190269439 Chr.21: 44455719-44455719 on Build GRCh38
rs1008532881 Chr.21: 44455714-44455714 on Build GRCh38
rs567630942 Chr.21: 44455370-44455370 on Build GRCh38
rs967652891 Chr.21: 44455726-44455726 on Build GRCh38
rs895027115 Chr.21: 44455280-44455280 on Build GRCh38
rs879747256 Chr.21: 44455710-44455710 on Build GRCh38
rs371664800 Chr.21: 44455286-44455286 on Build GRCh38
rs1044319309 Chr.21: 44455490-44455490 on Build GRCh38
rs935061193 Chr.21: 44455681-44455681 on Build GRCh38
rs994621305 Chr.21: 44455525-44455525 on Build GRCh38
rs953046277 Chr.21: 44455560-44455560 on Build GRCh38
rs1019163490 Chr.21: 44455587-44455587 on Build GRCh38
rs1011077785 Chr.21: 44455480-44455480 on Build GRCh38
rs902693305 Chr.21: 44455651-44455651 on Build GRCh38
rs575933140 Chr.21: 44455647-44455647 on Build GRCh38
rs376277198 Chr.21: 44455589-44455589 on Build GRCh38
rs983958165 Chr.21: 44455575-44455575 on Build GRCh38
rs1023285532 Chr.21: 44455711-44455711 on Build GRCh38
rs921635344 Chr.21: 44455344-44455344 on Build GRCh38
rs1056141937 Chr.21: 44455692-44455692 on Build GRCh38
rs1012614197 Chr.21: 44455317-44455317 on Build GRCh38
rs970617783 Chr.21: 44455297-44455297 on Build GRCh38
rs11701359 Chr.21: 44455312-44455312 on Build GRCh38
rs977577928 Chr.21: 44455591-44455591 on Build GRCh38
rs1012077578 Chr.21: 44455544-44455544 on Build GRCh38
rs13047477 Chr.21: 44455529-44455529 on Build GRCh38
rs866176815 Chr.21: 44455401-44455401 on Build GRCh38
rs960367742 Chr.21: 44455523-44455523 on Build GRCh38
rs1055720567 Chr.21: 44455434-44455434 on Build GRCh38
rs969109957 Chr.21: 44455346-44455347 on Build GRCh38
rs111322158 Chr.21: 44455483-44455483 on Build GRCh38
rs1045722820 Chr.21: 44455635-44455635 on Build GRCh38
rs999700332 Chr.21: 44455503-44455503 on Build GRCh38
rs558077772 Chr.21: 44455615-44455615 on Build GRCh38
rs1043876423 Chr.21: 44455631-44455631 on Build GRCh38
rs1036189080 Chr.21: 44455666-44455666 on Build GRCh38
rs1028722217 Chr.21: 44455707-44455707 on Build GRCh38
rs899020253 Chr.21: 44455686-44455686 on Build GRCh38
rs926153733 Chr.21: 44455592-44455592 on Build GRCh38
rs973022845 Chr.21: 44455336-44455336 on Build GRCh38
rs963473565 Chr.21: 44455316-44455316 on Build GRCh38
rs537695037 Chr.21: 44455311-44455311 on Build GRCh38
rs1004692534 Chr.21: 44455498-44455498 on Build GRCh38
rs963154897 Chr.21: 44455522-44455523 on Build GRCh38
rs987749782 Chr.21: 44455384-44455384 on Build GRCh38
rs1002895850 Chr.21: 44455697-44455697 on Build GRCh38
rs918441482 Chr.21: 44455563-44455563 on Build GRCh38
rs911861980 Chr.21: 44455581-44455581 on Build GRCh38
rs915896055 Chr.21: 44455633-44455633 on Build GRCh38
rs950809555 Chr.21: 44455564-44455564 on Build GRCh38
rs944914061 Chr.21: 44455269-44455269 on Build GRCh38
rs113733312 Chr.21: 44455501-44455501 on Build GRCh38
rs926026176 Chr.21: 44455418-44455418 on Build GRCh38
rs946981785 Chr.21: 44455627-44455627 on Build GRCh38
rs117139334 Chr.21: 44455385-44455385 on Build GRCh38
rs1055730723 Chr.21: 44455275-44455275 on Build GRCh38
rs1023503417 Chr.21: 44455550-44455550 on Build GRCh38

View Full Product Details