Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 24515121-24515627 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UPB1
Entrez Gene ID
51733
Gene Name
beta-ureidopropionase 1
Gene Aliases
BUP1
UniGene
Hs.731656
Gene Chromosome Location
Chr.22: 24495060-24528681 on Build GRCh38
Transcripts
NM_016327.2 XM_011530222.2 XM_011530223.2 XM_011530224.2 XM_011530225.2 XM_017028825.1 XM_017028826.1 XM_017028827.1 XM_017028828.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4164903 c.687G>A p.A229A Chr.22: 24515266-24515266 on Build GRCh38
1270000 c.734_735insCAG p.S246_I247insS Chr.22: 24515313-24515314 on Build GRCh38
6344729 c.636G>T p.M212I Chr.22: 24515215-24515215 on Build GRCh38
3552655 c.777G>A p.T259T Chr.22: 24515356-24515356 on Build GRCh38
3552654 c.680G>A p.R227K Chr.22: 24515259-24515259 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs988786819 Chr.22: 24515466-24515466 on Build GRCh38
rs765854590 Chr.22: 24515327-24515328 on Build GRCh38
rs942919552 Chr.22: 24515576-24515576 on Build GRCh38
rs536911464 Chr.22: 24515433-24515433 on Build GRCh38
rs138473981 Chr.22: 24515265-24515265 on Build GRCh38
rs750044777 Chr.22: 24515365-24515365 on Build GRCh38
rs763543766 Chr.22: 24515160-24515160 on Build GRCh38
rs765997452 Chr.22: 24515344-24515344 on Build GRCh38
rs757422743 Chr.22: 24515296-24515296 on Build GRCh38
rs776152017 Chr.22: 24515156-24515156 on Build GRCh38
rs539480751 Chr.22: 24515506-24515506 on Build GRCh38
rs756444659 Chr.22: 24515387-24515387 on Build GRCh38
rs746122883 Chr.22: 24515299-24515299 on Build GRCh38
rs746496215 Chr.22: 24515422-24515422 on Build GRCh38
rs113976771 Chr.22: 24515603-24515603 on Build GRCh38
rs754923905 Chr.22: 24515286-24515286 on Build GRCh38
rs938061328 Chr.22: 24515368-24515368 on Build GRCh38
rs748687705 Chr.22: 24515404-24515404 on Build GRCh38
rs752682016 Chr.22: 24515361-24515361 on Build GRCh38
rs770091431 Chr.22: 24515302-24515302 on Build GRCh38
rs147133523 Chr.22: 24515255-24515255 on Build GRCh38
rs755892229 Chr.22: 24515217-24515217 on Build GRCh38
rs764880194 Chr.22: 24515355-24515355 on Build GRCh38
rs1028191336 Chr.22: 24515301-24515301 on Build GRCh38
rs74883165 Chr.22: 24515281-24515281 on Build GRCh38
rs773774550 Chr.22: 24515263-24515263 on Build GRCh38
rs374480199 Chr.22: 24515326-24515326 on Build GRCh38
rs368688414 Chr.22: 24515336-24515336 on Build GRCh38
rs753638837 Chr.22: 24515349-24515349 on Build GRCh38
rs570247589 Chr.22: 24515225-24515225 on Build GRCh38
rs150690829 Chr.22: 24515311-24515311 on Build GRCh38
rs143521220 Chr.22: 24515237-24515237 on Build GRCh38
rs778901070 Chr.22: 24515288-24515288 on Build GRCh38
rs558108661 Chr.22: 24515516-24515516 on Build GRCh38
rs779583239 Chr.22: 24515402-24515403 on Build GRCh38
rs751032342 Chr.22: 24515180-24515180 on Build GRCh38
rs369523102 Chr.22: 24515412-24515412 on Build GRCh38
rs374257101 Chr.22: 24515278-24515278 on Build GRCh38
rs370930876 Chr.22: 24515266-24515266 on Build GRCh38
rs779082431 Chr.22: 24515401-24515401 on Build GRCh38
rs756486950 Chr.22: 24515204-24515204 on Build GRCh38
rs913444862 Chr.22: 24515275-24515275 on Build GRCh38
rs199794499 Chr.22: 24515393-24515393 on Build GRCh38
rs913247230 Chr.22: 24515492-24515492 on Build GRCh38
rs779769195 Chr.22: 24515218-24515218 on Build GRCh38
rs182805014 Chr.22: 24515562-24515562 on Build GRCh38
rs147994426 Chr.22: 24515248-24515248 on Build GRCh38
rs550836033 Chr.22: 24515536-24515536 on Build GRCh38
rs149812800 Chr.22: 24515370-24515370 on Build GRCh38
rs753400028 Chr.22: 24515372-24515373 on Build GRCh38
rs781352126 Chr.22: 24515297-24515297 on Build GRCh38
rs139932368 Chr.22: 24515324-24515324 on Build GRCh38
rs370757106 Chr.22: 24515293-24515293 on Build GRCh38
rs1049986104 Chr.22: 24515507-24515507 on Build GRCh38
rs759971779 Chr.22: 24515151-24515151 on Build GRCh38
rs769111199 Chr.22: 24515313-24515313 on Build GRCh38
rs780614364 Chr.22: 24515308-24515308 on Build GRCh38
rs756909197 Chr.22: 24515213-24515213 on Build GRCh38
rs769184232 Chr.22: 24515262-24515262 on Build GRCh38
rs1033063297 Chr.22: 24515431-24515431 on Build GRCh38
rs748313940 Chr.22: 24515519-24515519 on Build GRCh38
rs144135211 Chr.22: 24515285-24515285 on Build GRCh38
rs765900681 Chr.22: 24515155-24515155 on Build GRCh38
rs919496708 Chr.22: 24515339-24515339 on Build GRCh38
rs149294607 Chr.22: 24515236-24515236 on Build GRCh38
rs763449450 Chr.22: 24515157-24515157 on Build GRCh38
rs755500357 Chr.22: 24515392-24515392 on Build GRCh38
rs143438140 Chr.22: 24515254-24515254 on Build GRCh38
rs746026530 Chr.22: 24515247-24515247 on Build GRCh38
rs376130197 Chr.22: 24515292-24515292 on Build GRCh38
rs575607732 Chr.22: 24515395-24515395 on Build GRCh38
rs780319133 Chr.22: 24515392-24515392 on Build GRCh38
rs371352339 Chr.22: 24515356-24515356 on Build GRCh38
rs760086834 Chr.22: 24515222-24515223 on Build GRCh38
rs955592903 Chr.22: 24515509-24515509 on Build GRCh38
rs772522964 Chr.22: 24515409-24515409 on Build GRCh38
rs540720327 Chr.22: 24515573-24515573 on Build GRCh38
rs760346924 Chr.22: 24515342-24515342 on Build GRCh38
rs1047389346 Chr.22: 24515333-24515333 on Build GRCh38
rs898820305 Chr.22: 24515323-24515323 on Build GRCh38
rs942219538 Chr.22: 24515183-24515183 on Build GRCh38
rs774909683 Chr.22: 24515322-24515322 on Build GRCh38
rs199744697 Chr.22: 24515234-24515234 on Build GRCh38
rs953925362 Chr.22: 24515305-24515305 on Build GRCh38
rs749190814 Chr.22: 24515220-24515220 on Build GRCh38
rs957687510 Chr.22: 24515442-24515442 on Build GRCh38
rs201435289 Chr.22: 24515230-24515230 on Build GRCh38
rs200508190 Chr.22: 24515350-24515350 on Build GRCh38
rs766196011 Chr.22: 24515282-24515282 on Build GRCh38
rs781442372 Chr.22: 24515373-24515373 on Build GRCh38
rs750286662 Chr.22: 24515214-24515214 on Build GRCh38
rs200633007 Chr.22: 24515400-24515400 on Build GRCh38
rs374432230 Chr.22: 24515375-24515375 on Build GRCh38

View Full Product Details