Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 24571981-24572484 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNRPD3
Entrez Gene ID
6634
Gene Name
small nuclear ribonucleoprotein D3 polypeptide
Gene Aliases
SMD3, Sm-D3
UniGene
Hs.356549
Gene Chromosome Location
Chr.22: 24555650-24574964 on Build GRCh38
Transcripts
NM_001278656.1 NM_004175.4 NR_103819.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3842316 c.327G>A p.L109L Chr.22: 24572025-24572025 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs868508310 Chr.22: 24572324-24572324 on Build GRCh38
rs761870566 Chr.22: 24572007-24572007 on Build GRCh38
rs917394205 Chr.22: 24572453-24572453 on Build GRCh38
rs1017121544 Chr.22: 24572404-24572404 on Build GRCh38
rs765355854 Chr.22: 24572074-24572074 on Build GRCh38
rs997815266 Chr.22: 24572435-24572435 on Build GRCh38
rs752998013 Chr.22: 24572085-24572085 on Build GRCh38
rs965886265 Chr.22: 24572333-24572333 on Build GRCh38
rs1056442263 Chr.22: 24572261-24572261 on Build GRCh38
rs550760175 Chr.22: 24572062-24572062 on Build GRCh38
rs905822866 Chr.22: 24572119-24572119 on Build GRCh38
rs976364391 Chr.22: 24572027-24572027 on Build GRCh38
rs561047283 Chr.22: 24572135-24572139 on Build GRCh38
rs955728119 Chr.22: 24572023-24572023 on Build GRCh38
rs757650281 Chr.22: 24572389-24572389 on Build GRCh38
rs948342622 Chr.22: 24572334-24572334 on Build GRCh38
rs972974135 Chr.22: 24572448-24572448 on Build GRCh38
rs1037183323 Chr.22: 24572067-24572068 on Build GRCh38
rs763810550 Chr.22: 24572057-24572057 on Build GRCh38
rs750305886 Chr.22: 24572016-24572016 on Build GRCh38
rs537442662 Chr.22: 24572357-24572357 on Build GRCh38
rs549976679 Chr.22: 24572442-24572442 on Build GRCh38
rs999679610 Chr.22: 24572378-24572378 on Build GRCh38
rs767348858 Chr.22: 24572015-24572015 on Build GRCh38
rs918659029 Chr.22: 24572201-24572201 on Build GRCh38
rs1053297713 Chr.22: 24572392-24572392 on Build GRCh38
rs566269358 Chr.22: 24572172-24572172 on Build GRCh38
rs191761698 Chr.22: 24572198-24572198 on Build GRCh38
rs188465154 Chr.22: 24572087-24572087 on Build GRCh38
rs891517300 Chr.22: 24572396-24572396 on Build GRCh38
rs759697742 Chr.22: 24572072-24572072 on Build GRCh38
rs184005320 Chr.22: 24572341-24572341 on Build GRCh38
rs917320793 Chr.22: 24572081-24572081 on Build GRCh38
rs941197735 Chr.22: 24572185-24572185 on Build GRCh38
rs1018759988 Chr.22: 24572321-24572321 on Build GRCh38
rs548563175 Chr.22: 24572279-24572279 on Build GRCh38
rs916834190 Chr.22: 24572303-24572303 on Build GRCh38
rs985315301 Chr.22: 24572125-24572125 on Build GRCh38
rs867672691 Chr.22: 24572082-24572082 on Build GRCh38
rs939354279 Chr.22: 24572239-24572239 on Build GRCh38
rs940189012 Chr.22: 24572049-24572049 on Build GRCh38
rs951791876 Chr.22: 24572379-24572379 on Build GRCh38
rs865894165 Chr.22: 24572121-24572121 on Build GRCh38
rs984510482 Chr.22: 24572381-24572381 on Build GRCh38
rs909746609 Chr.22: 24572166-24572166 on Build GRCh38
rs1029547775 Chr.22: 24572053-24572053 on Build GRCh38
rs774868985 Chr.22: 24572050-24572051 on Build GRCh38
rs1044506400 Chr.22: 24572097-24572097 on Build GRCh38

View Full Product Details