Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 91968060-91968565 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRDT
Entrez Gene ID
676
Gene Name
bromodomain testis associated
Gene Aliases
BRD6, CT9
UniGene
Hs.482520
Gene Chromosome Location
Chr.1: 91949371-92014428 on Build GRCh38
Transcripts
NM_001242805.2 NM_001242806.2 NM_001242807.2 NM_001242808.2 NM_001242810.2 NM_001726.4 NM_207189.3 XM_006710853.3 XM_006710854.3 XM_006710855.3 XM_006710856.3 XM_006710857.3 XM_011542032.2 XM_011542033.2 XM_011542034.2 XM_011542035.2 XM_011542036.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7357730 c.395A>T p.Q132L Chr.1: 91968210-91968210 on Build GRCh38
7104176 c.409G>C p.E137Q Chr.1: 91968224-91968224 on Build GRCh38
5881810 c.418G>T p.V140L Chr.1: 91968233-91968233 on Build GRCh38
4917901 c.346G>T p.V116F Chr.1: 91968161-91968161 on Build GRCh38
1688024 c.347T>C p.V116A Chr.1: 91968162-91968162 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs576541262 Chr.1: 91968318-91968319 on Build GRCh38
rs759854716 Chr.1: 91968294-91968295 on Build GRCh38
rs995446450 Chr.1: 91968265-91968265 on Build GRCh38
rs371270716 Chr.1: 91968164-91968164 on Build GRCh38
rs769495449 Chr.1: 91968457-91968457 on Build GRCh38
rs758923042 Chr.1: 91968311-91968311 on Build GRCh38
rs201895618 Chr.1: 91968178-91968178 on Build GRCh38
rs747588891 Chr.1: 91968298-91968298 on Build GRCh38
rs753397891 Chr.1: 91968146-91968146 on Build GRCh38
rs576896085 Chr.1: 91968424-91968424 on Build GRCh38
rs998939280 Chr.1: 91968193-91968193 on Build GRCh38
rs755274471 Chr.1: 91968232-91968232 on Build GRCh38
rs761447494 Chr.1: 91968307-91968310 on Build GRCh38
rs772653292 Chr.1: 91968129-91968129 on Build GRCh38
rs903168722 Chr.1: 91968441-91968441 on Build GRCh38
rs534616259 Chr.1: 91968465-91968465 on Build GRCh38
rs377545364 Chr.1: 91968309-91968309 on Build GRCh38
rs1043023794 Chr.1: 91968112-91968112 on Build GRCh38
rs912973873 Chr.1: 91968357-91968357 on Build GRCh38
rs774817366 Chr.1: 91968122-91968122 on Build GRCh38
rs183793740 Chr.1: 91968213-91968213 on Build GRCh38
rs934617375 Chr.1: 91968462-91968462 on Build GRCh38
rs772271165 Chr.1: 91968283-91968283 on Build GRCh38
rs759276811 Chr.1: 91968150-91968150 on Build GRCh38
rs897241055 Chr.1: 91968522-91968522 on Build GRCh38
rs890255289 Chr.1: 91968173-91968173 on Build GRCh38
rs921189298 Chr.1: 91968345-91968345 on Build GRCh38
rs565221248 Chr.1: 91968107-91968107 on Build GRCh38
rs1008746553 Chr.1: 91968202-91968202 on Build GRCh38
rs577256865 Chr.1: 91968336-91968336 on Build GRCh38
rs758089934 Chr.1: 91968168-91968168 on Build GRCh38
rs1037098594 Chr.1: 91968511-91968511 on Build GRCh38
rs748339704 Chr.1: 91968377-91968377 on Build GRCh38
rs771536534 Chr.1: 91968305-91968305 on Build GRCh38
rs778949542 Chr.1: 91968235-91968235 on Build GRCh38
rs749569003 Chr.1: 91968109-91968109 on Build GRCh38
rs947060730 Chr.1: 91968414-91968414 on Build GRCh38
rs762058958 Chr.1: 91968127-91968127 on Build GRCh38
rs940520285 Chr.1: 91968510-91968510 on Build GRCh38
rs765401772 Chr.1: 91968320-91968321 on Build GRCh38
rs921564798 Chr.1: 91968449-91968449 on Build GRCh38
rs374669808 Chr.1: 91968172-91968172 on Build GRCh38
rs970278891 Chr.1: 91968290-91968290 on Build GRCh38
rs974178334 Chr.1: 91968293-91968293 on Build GRCh38
rs1036273780 Chr.1: 91968324-91968324 on Build GRCh38
rs768018267 Chr.1: 91968297-91968297 on Build GRCh38
rs111809141 Chr.1: 91968210-91968210 on Build GRCh38
rs777502998 Chr.1: 91968489-91968489 on Build GRCh38
rs766048850 Chr.1: 91968141-91968141 on Build GRCh38
rs576861050 Chr.1: 91968428-91968428 on Build GRCh38
rs114091851 Chr.1: 91968501-91968501 on Build GRCh38
rs191694183 Chr.1: 91968137-91968137 on Build GRCh38
rs753146789 Chr.1: 91968296-91968297 on Build GRCh38
rs975179816 Chr.1: 91968342-91968342 on Build GRCh38
rs758795755 Chr.1: 91968319-91968320 on Build GRCh38
rs139011380 Chr.1: 91968205-91968205 on Build GRCh38
rs978421418 Chr.1: 91968417-91968417 on Build GRCh38
rs558658040 Chr.1: 91968330-91968330 on Build GRCh38
rs963628905 Chr.1: 91968280-91968280 on Build GRCh38
rs146865063 Chr.1: 91968113-91968113 on Build GRCh38
rs1023639532 Chr.1: 91968212-91968212 on Build GRCh38
rs931893560 Chr.1: 91968470-91968470 on Build GRCh38
rs995498526 Chr.1: 91968540-91968540 on Build GRCh38
rs537230850 Chr.1: 91968296-91968296 on Build GRCh38
rs764775938 Chr.1: 91968151-91968151 on Build GRCh38
rs773702673 Chr.1: 91968291-91968291 on Build GRCh38
rs988981232 Chr.1: 91968352-91968352 on Build GRCh38
rs777691110 Chr.1: 91968171-91968171 on Build GRCh38
rs796973729 Chr.1: 91968318-91968319 on Build GRCh38
rs754364560 Chr.1: 91968155-91968156 on Build GRCh38
rs748572833 Chr.1: 91968236-91968236 on Build GRCh38
rs368145053 Chr.1: 91968132-91968132 on Build GRCh38
rs201156875 Chr.1: 91968318-91968318 on Build GRCh38
rs55649816 Chr.1: 91968203-91968203 on Build GRCh38
rs749293297 Chr.1: 91968231-91968231 on Build GRCh38

View Full Product Details