Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 30266633-30267140 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OSM
Entrez Gene ID
5008
Gene Name
oncostatin M
Gene Aliases
-
UniGene
Hs.248156
Gene Chromosome Location
Chr.22: 30262828-30266843 on Build GRCh38
Transcripts
NM_020530.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5421209 c.33C>T p.L11L Chr.22: 30266767-30266767 on Build GRCh38
5151244 c.10C>T p.L4L Chr.22: 30266790-30266790 on Build GRCh38
5834113 c.22_23delAG p.R8fs*61 Chr.22: 30266777-30266778 on Build GRCh38
7003468 c.27G>T p.T9T Chr.22: 30266773-30266773 on Build GRCh38
5017923 c.7delG p.V3fs*26 Chr.22: 30266793-30266793 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs937521271 Chr.22: 30266707-30266707 on Build GRCh38
rs762087973 Chr.22: 30266796-30266796 on Build GRCh38
rs1033392412 Chr.22: 30267053-30267053 on Build GRCh38
rs1048204917 Chr.22: 30267005-30267005 on Build GRCh38
rs201346538 Chr.22: 30266817-30266817 on Build GRCh38
rs910463433 Chr.22: 30267075-30267075 on Build GRCh38
rs374457471 Chr.22: 30266843-30266843 on Build GRCh38
rs924611685 Chr.22: 30266876-30266876 on Build GRCh38
rs962310170 Chr.22: 30266659-30266659 on Build GRCh38
rs754219364 Chr.22: 30266737-30266737 on Build GRCh38
rs767656484 Chr.22: 30266730-30266730 on Build GRCh38
rs766992089 Chr.22: 30266820-30266820 on Build GRCh38
rs200546565 Chr.22: 30266829-30266829 on Build GRCh38
rs906166913 Chr.22: 30267023-30267023 on Build GRCh38
rs543193005 Chr.22: 30266825-30266825 on Build GRCh38
rs13057223 Chr.22: 30266950-30266950 on Build GRCh38
rs537157034 Chr.22: 30267046-30267046 on Build GRCh38
rs113090322 Chr.22: 30266948-30266948 on Build GRCh38
rs150830767 Chr.22: 30266983-30266983 on Build GRCh38
rs764363028 Chr.22: 30266720-30266720 on Build GRCh38
rs201600911 Chr.22: 30266716-30266716 on Build GRCh38
rs777351171 Chr.22: 30266815-30266815 on Build GRCh38
rs200002012 Chr.22: 30266811-30266811 on Build GRCh38
rs971435857 Chr.22: 30267052-30267052 on Build GRCh38
rs529882318 Chr.22: 30266999-30266999 on Build GRCh38
rs374916412 Chr.22: 30266744-30266744 on Build GRCh38
rs199877492 Chr.22: 30266886-30266886 on Build GRCh38
rs200578717 Chr.22: 30266734-30266734 on Build GRCh38
rs199577355 Chr.22: 30266731-30266731 on Build GRCh38
rs5753056 Chr.22: 30266784-30266784 on Build GRCh38
rs746986431 Chr.22: 30266766-30266766 on Build GRCh38
rs5763919 Chr.22: 30266774-30266774 on Build GRCh38
rs747208758 Chr.22: 30266656-30266656 on Build GRCh38
rs777221420 Chr.22: 30266757-30266757 on Build GRCh38
rs991557623 Chr.22: 30266709-30266709 on Build GRCh38
rs991655576 Chr.22: 30266838-30266838 on Build GRCh38
rs745670219 Chr.22: 30266777-30266777 on Build GRCh38
rs935810739 Chr.22: 30267066-30267066 on Build GRCh38
rs1025806674 Chr.22: 30266974-30266974 on Build GRCh38
rs201319597 Chr.22: 30266847-30266847 on Build GRCh38
rs187758119 Chr.22: 30266828-30266828 on Build GRCh38
rs558644325 Chr.22: 30267114-30267114 on Build GRCh38
rs576072075 Chr.22: 30266708-30266708 on Build GRCh38
rs943950838 Chr.22: 30266748-30266748 on Build GRCh38
rs991497107 Chr.22: 30267040-30267040 on Build GRCh38
rs959207206 Chr.22: 30267067-30267067 on Build GRCh38
rs758011487 Chr.22: 30266826-30266826 on Build GRCh38
rs982126620 Chr.22: 30266679-30266679 on Build GRCh38
rs779681018 Chr.22: 30266848-30266848 on Build GRCh38
rs1013204591 Chr.22: 30267088-30267088 on Build GRCh38
rs200134196 Chr.22: 30266745-30266745 on Build GRCh38
rs547421527 Chr.22: 30266933-30266933 on Build GRCh38
rs201836267 Chr.22: 30266735-30266735 on Build GRCh38
rs943321086 Chr.22: 30267108-30267108 on Build GRCh38
rs561872808 Chr.22: 30267007-30267007 on Build GRCh38
rs1040933499 Chr.22: 30266906-30266906 on Build GRCh38
rs554408336 Chr.22: 30266704-30266704 on Build GRCh38
rs13058518 Chr.22: 30266949-30266949 on Build GRCh38
rs1006154646 Chr.22: 30267006-30267006 on Build GRCh38
rs373813803 Chr.22: 30266773-30266773 on Build GRCh38
rs1041733556 Chr.22: 30266741-30266741 on Build GRCh38
rs770741657 Chr.22: 30266715-30266715 on Build GRCh38
rs1003897797 Chr.22: 30267015-30267015 on Build GRCh38
rs947494300 Chr.22: 30266729-30266729 on Build GRCh38
rs770831672 Chr.22: 30266770-30266770 on Build GRCh38
rs112031212 Chr.22: 30266824-30266824 on Build GRCh38
rs774361182 Chr.22: 30266721-30266721 on Build GRCh38
rs761956425 Chr.22: 30266725-30266725 on Build GRCh38
rs774656058 Chr.22: 30266785-30266785 on Build GRCh38
rs532326135 Chr.22: 30266932-30266932 on Build GRCh38
rs769407747 Chr.22: 30266779-30266779 on Build GRCh38
rs199606297 Chr.22: 30266855-30266855 on Build GRCh38
rs769713692 Chr.22: 30266988-30266988 on Build GRCh38
rs201208436 Chr.22: 30266872-30266872 on Build GRCh38
rs914779977 Chr.22: 30266839-30266839 on Build GRCh38
rs202168238 Chr.22: 30266822-30266822 on Build GRCh38
rs767746513 Chr.22: 30266809-30266809 on Build GRCh38
rs200933685 Chr.22: 30266751-30266751 on Build GRCh38
rs527502359 Chr.22: 30267018-30267018 on Build GRCh38
rs201106496 Chr.22: 30266818-30266818 on Build GRCh38
rs200232846 Chr.22: 30266823-30266823 on Build GRCh38
rs757253126 Chr.22: 30266834-30266834 on Build GRCh38
rs747702309 Chr.22: 30266750-30266750 on Build GRCh38
rs200276463 Chr.22: 30266865-30266865 on Build GRCh38
rs1054349685 Chr.22: 30266852-30266852 on Build GRCh38
rs944049950 Chr.22: 30266901-30266901 on Build GRCh38

View Full Product Details