Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 32483954-32484498 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO7
Entrez Gene ID
25793
Gene Name
F-box protein 7
Gene Aliases
FBX, FBX07, FBX7, PARK15, PKPS
UniGene
Hs.5912
Gene Chromosome Location
Chr.22: 32474682-32498831 on Build GRCh38
Transcripts
NM_001033024.1 NM_001257990.1 NM_012179.3 XM_011530106.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7103571 c.545C>T p.S182L Chr.22: 32484024-32484024 on Build GRCh38
4103711 c.542A>C p.H181P Chr.22: 32484021-32484021 on Build GRCh38
1714311 c.356C>T p.A119V Chr.22: 32484072-32484072 on Build GRCh38
5772107 c.403C>T p.P135S Chr.22: 32484119-32484119 on Build GRCh38
7078928 c.510C>T p.L170L Chr.22: 32483989-32483989 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs964713096 Chr.22: 32484356-32484356 on Build GRCh38
rs376455464 Chr.22: 32484000-32484000 on Build GRCh38
rs1056027917 Chr.22: 32484232-32484232 on Build GRCh38
rs771557092 Chr.22: 32484008-32484008 on Build GRCh38
rs891035065 Chr.22: 32484311-32484311 on Build GRCh38
rs561706353 Chr.22: 32484438-32484438 on Build GRCh38
rs536454057 Chr.22: 32484289-32484289 on Build GRCh38
rs541812199 Chr.22: 32484425-32484425 on Build GRCh38
rs774173729 Chr.22: 32484168-32484168 on Build GRCh38
rs573247314 Chr.22: 32484407-32484407 on Build GRCh38
rs947913701 Chr.22: 32484223-32484223 on Build GRCh38
rs556798436 Chr.22: 32484360-32484360 on Build GRCh38
rs748979295 Chr.22: 32484082-32484082 on Build GRCh38
rs748634926 Chr.22: 32483984-32483984 on Build GRCh38
rs370890171 Chr.22: 32484127-32484127 on Build GRCh38
rs374263493 Chr.22: 32484043-32484043 on Build GRCh38
rs113757678 Chr.22: 32484250-32484250 on Build GRCh38
rs73884927 Chr.22: 32484200-32484200 on Build GRCh38
rs781083417 Chr.22: 32484184-32484184 on Build GRCh38
rs1031645841 Chr.22: 32484365-32484365 on Build GRCh38
rs144041512 Chr.22: 32484143-32484143 on Build GRCh38
rs41311141 Chr.22: 32484019-32484019 on Build GRCh38
rs778770873 Chr.22: 32484053-32484054 on Build GRCh38
rs577331850 Chr.22: 32484386-32484386 on Build GRCh38
rs956153738 Chr.22: 32484021-32484021 on Build GRCh38
rs371294823 Chr.22: 32484014-32484014 on Build GRCh38
rs564422823 Chr.22: 32484268-32484268 on Build GRCh38
rs947767980 Chr.22: 32484245-32484246 on Build GRCh38
rs894833764 Chr.22: 32484238-32484238 on Build GRCh38
rs777298117 Chr.22: 32484137-32484137 on Build GRCh38
rs749987502 Chr.22: 32484074-32484074 on Build GRCh38
rs1021814487 Chr.22: 32484050-32484050 on Build GRCh38
rs746336005 Chr.22: 32484141-32484141 on Build GRCh38
rs891858829 Chr.22: 32484375-32484375 on Build GRCh38
rs369968316 Chr.22: 32484394-32484394 on Build GRCh38
rs186474609 Chr.22: 32484197-32484197 on Build GRCh38
rs147299941 Chr.22: 32484406-32484406 on Build GRCh38
rs775586796 Chr.22: 32484261-32484261 on Build GRCh38
rs182333581 Chr.22: 32484357-32484357 on Build GRCh38
rs747792857 Chr.22: 32484126-32484128 on Build GRCh38
rs758036175 Chr.22: 32484176-32484176 on Build GRCh38
rs762807197 Chr.22: 32484321-32484321 on Build GRCh38
rs200064513 Chr.22: 32483987-32483987 on Build GRCh38
rs778265071 Chr.22: 32484098-32484098 on Build GRCh38
rs1016700343 Chr.22: 32484059-32484059 on Build GRCh38
rs766883090 Chr.22: 32484126-32484127 on Build GRCh38
rs894986377 Chr.22: 32484221-32484221 on Build GRCh38
rs61730783 Chr.22: 32484080-32484080 on Build GRCh38
rs752994693 Chr.22: 32484039-32484039 on Build GRCh38
rs954232038 Chr.22: 32484445-32484445 on Build GRCh38
rs201788989 Chr.22: 32484153-32484153 on Build GRCh38
rs190085466 Chr.22: 32484207-32484207 on Build GRCh38
rs968560924 Chr.22: 32484455-32484455 on Build GRCh38
rs978271473 Chr.22: 32484473-32484473 on Build GRCh38
rs939627860 Chr.22: 32484219-32484219 on Build GRCh38
rs754569362 Chr.22: 32484083-32484083 on Build GRCh38
rs763963522 Chr.22: 32484057-32484057 on Build GRCh38
rs779737534 Chr.22: 32483976-32483976 on Build GRCh38
rs972418747 Chr.22: 32484192-32484192 on Build GRCh38
rs987453630 Chr.22: 32484095-32484095 on Build GRCh38
rs368665145 Chr.22: 32484069-32484069 on Build GRCh38
rs548204763 Chr.22: 32484066-32484066 on Build GRCh38
rs186493739 Chr.22: 32484391-32484391 on Build GRCh38
rs990695251 Chr.22: 32484041-32484041 on Build GRCh38
rs1025923884 Chr.22: 32484296-32484296 on Build GRCh38
rs1007511723 Chr.22: 32484463-32484463 on Build GRCh38
rs776886014 Chr.22: 32484170-32484170 on Build GRCh38
rs755646509 Chr.22: 32484077-32484077 on Build GRCh38
rs772581980 Chr.22: 32484001-32484001 on Build GRCh38
rs775374545 Chr.22: 32484149-32484149 on Build GRCh38
rs574361432 Chr.22: 32484343-32484343 on Build GRCh38
rs866788907 Chr.22: 32484083-32484084 on Build GRCh38
rs773386419 Chr.22: 32483996-32483996 on Build GRCh38
rs994014483 Chr.22: 32484280-32484280 on Build GRCh38
rs114187411 Chr.22: 32484156-32484156 on Build GRCh38
rs765181065 Chr.22: 32484031-32484031 on Build GRCh38
rs748847363 Chr.22: 32483985-32483985 on Build GRCh38
rs747343769 Chr.22: 32484104-32484104 on Build GRCh38

View Full Product Details