Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 35393578-35394086 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HMOX1
Entrez Gene ID
3162
Gene Name
heme oxygenase 1
Gene Aliases
HMOX1D, HO-1, HSP32, bK286B10
UniGene
Hs.517581
Gene Chromosome Location
Chr.22: 35381067-35394214 on Build GRCh38
Transcripts
NM_002133.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7371350 c.860C>G p.A287G Chr.22: 35393591-35393591 on Build GRCh38
2939477 c.856T>C p.Y286H Chr.22: 35393587-35393587 on Build GRCh38
7371349 c.857A>C p.Y286S Chr.22: 35393588-35393588 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs753850950 Chr.22: 35393679-35393679 on Build GRCh38
rs944731148 Chr.22: 35393779-35393779 on Build GRCh38
rs995042284 Chr.22: 35393633-35393633 on Build GRCh38
rs911889128 Chr.22: 35393770-35393770 on Build GRCh38
rs769322032 Chr.22: 35393934-35393934 on Build GRCh38
rs367987395 Chr.22: 35393610-35393610 on Build GRCh38
rs901692220 Chr.22: 35393775-35393775 on Build GRCh38
rs773615991 Chr.22: 35393612-35393612 on Build GRCh38
rs906855760 Chr.22: 35393923-35393923 on Build GRCh38
rs967971070 Chr.22: 35393814-35393815 on Build GRCh38
rs539113031 Chr.22: 35393673-35393673 on Build GRCh38
rs550546401 Chr.22: 35393651-35393651 on Build GRCh38
rs930769568 Chr.22: 35393836-35393836 on Build GRCh38
rs577322782 Chr.22: 35393946-35393946 on Build GRCh38
rs978729173 Chr.22: 35393819-35393819 on Build GRCh38
rs1011790497 Chr.22: 35393965-35393965 on Build GRCh38
rs990017315 Chr.22: 35393702-35393702 on Build GRCh38
rs112611068 Chr.22: 35393909-35393909 on Build GRCh38
rs957403922 Chr.22: 35393696-35393696 on Build GRCh38
rs186402981 Chr.22: 35394044-35394044 on Build GRCh38
rs566260020 Chr.22: 35393753-35393753 on Build GRCh38
rs1048795422 Chr.22: 35393699-35393699 on Build GRCh38
rs749455128 Chr.22: 35393691-35393691 on Build GRCh38
rs371438796 Chr.22: 35393831-35393831 on Build GRCh38
rs771027320 Chr.22: 35394051-35394051 on Build GRCh38
rs1036389777 Chr.22: 35393780-35393780 on Build GRCh38
rs1019162710 Chr.22: 35393749-35393749 on Build GRCh38
rs11555832 Chr.22: 35393876-35393876 on Build GRCh38
rs894731419 Chr.22: 35393646-35393646 on Build GRCh38
rs181746701 Chr.22: 35393978-35393978 on Build GRCh38
rs149739243 Chr.22: 35393662-35393662 on Build GRCh38
rs559481884 Chr.22: 35393669-35393669 on Build GRCh38
rs147862586 Chr.22: 35393747-35393747 on Build GRCh38
rs894671137 Chr.22: 35393851-35393851 on Build GRCh38
rs912505747 Chr.22: 35394018-35394018 on Build GRCh38
rs760436375 Chr.22: 35393649-35393649 on Build GRCh38
rs757798115 Chr.22: 35393730-35393730 on Build GRCh38
rs774924065 Chr.22: 35393600-35393600 on Build GRCh38
rs557349117 Chr.22: 35393942-35393942 on Build GRCh38
rs906139651 Chr.22: 35393690-35393690 on Build GRCh38
rs930923547 Chr.22: 35393973-35393973 on Build GRCh38
rs559862155 Chr.22: 35394008-35394008 on Build GRCh38
rs17880056 Chr.22: 35393842-35393842 on Build GRCh38
rs201438906 Chr.22: 35393637-35393637 on Build GRCh38
rs772833576 Chr.22: 35393987-35393987 on Build GRCh38
rs776640266 Chr.22: 35393638-35393638 on Build GRCh38
rs866643988 Chr.22: 35393636-35393636 on Build GRCh38
rs977151022 Chr.22: 35393689-35393689 on Build GRCh38
rs572339648 Chr.22: 35393963-35393963 on Build GRCh38
rs748685954 Chr.22: 35394041-35394041 on Build GRCh38
rs957145319 Chr.22: 35393960-35393960 on Build GRCh38
rs535207353 Chr.22: 35393788-35393788 on Build GRCh38
rs370298007 Chr.22: 35393941-35393941 on Build GRCh38
rs779325381 Chr.22: 35393733-35393733 on Build GRCh38
rs954051818 Chr.22: 35393993-35393993 on Build GRCh38
rs748669503 Chr.22: 35393602-35393602 on Build GRCh38
rs111787422 Chr.22: 35393731-35393731 on Build GRCh38
rs989347198 Chr.22: 35393660-35393660 on Build GRCh38
rs759671776 Chr.22: 35393649-35393649 on Build GRCh38
rs758713757 Chr.22: 35393820-35393820 on Build GRCh38
rs986660564 Chr.22: 35393999-35393999 on Build GRCh38
rs886147364 Chr.22: 35393742-35393742 on Build GRCh38
rs530752316 Chr.22: 35393623-35393623 on Build GRCh38
rs907546802 Chr.22: 35394000-35394001 on Build GRCh38

View Full Product Details