Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 38068952-38069457 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PICK1
Entrez Gene ID
9463
Gene Name
protein interacting with PRKCA 1
Gene Aliases
PICK, PRKCABP
UniGene
Hs.180871
Gene Chromosome Location
Chr.22: 38056311-38075704 on Build GRCh38
Transcripts
NM_001039583.1 NM_001039584.1 NM_012407.3 XM_006724379.2 XM_006724380.2 XM_011530517.2 XM_017029090.1 XM_017029091.1 XM_017029092.1 XM_017029093.1 XM_017029094.1 XM_017029095.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4492486 c.398C>T p.T133I Chr.22: 38069081-38069081 on Build GRCh38
6256543 c.361G>T p.V121F Chr.22: 38069044-38069044 on Build GRCh38
444981 c.377T>G p.V126G Chr.22: 38069060-38069060 on Build GRCh38
5749697 c.376G>A p.V126M Chr.22: 38069059-38069059 on Build GRCh38
6162288 c.367C>T p.H123Y Chr.22: 38069050-38069050 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781088929 Chr.22: 38069147-38069147 on Build GRCh38
rs867650759 Chr.22: 38069138-38069138 on Build GRCh38
rs566149092 Chr.22: 38069181-38069181 on Build GRCh38
rs946885382 Chr.22: 38069256-38069256 on Build GRCh38
rs776641391 Chr.22: 38069065-38069065 on Build GRCh38
rs556131700 Chr.22: 38069362-38069362 on Build GRCh38
rs894001486 Chr.22: 38069249-38069249 on Build GRCh38
rs1042056583 Chr.22: 38069398-38069398 on Build GRCh38
rs922892034 Chr.22: 38069169-38069169 on Build GRCh38
rs755404092 Chr.22: 38069101-38069101 on Build GRCh38
rs759023839 Chr.22: 38069198-38069198 on Build GRCh38
rs368517908 Chr.22: 38069132-38069132 on Build GRCh38
rs924217481 Chr.22: 38069417-38069417 on Build GRCh38
rs768374064 Chr.22: 38069167-38069167 on Build GRCh38
rs747168009 Chr.22: 38069135-38069135 on Build GRCh38
rs538134619 Chr.22: 38069311-38069311 on Build GRCh38
rs544496738 Chr.22: 38069220-38069220 on Build GRCh38
rs1003563350 Chr.22: 38069149-38069149 on Build GRCh38
rs773477637 Chr.22: 38069060-38069060 on Build GRCh38
rs775410954 Chr.22: 38069155-38069155 on Build GRCh38
rs983925051 Chr.22: 38069397-38069397 on Build GRCh38
rs981290209 Chr.22: 38069028-38069028 on Build GRCh38
rs1011013513 Chr.22: 38069054-38069054 on Build GRCh38
rs757946960 Chr.22: 38068993-38068993 on Build GRCh38
rs73419888 Chr.22: 38069027-38069027 on Build GRCh38
rs201374426 Chr.22: 38069109-38069109 on Build GRCh38
rs572434626 Chr.22: 38069022-38069022 on Build GRCh38
rs370162829 Chr.22: 38069153-38069153 on Build GRCh38
rs756670812 Chr.22: 38069011-38069011 on Build GRCh38
rs369175718 Chr.22: 38069298-38069298 on Build GRCh38
rs922942948 Chr.22: 38069375-38069375 on Build GRCh38
rs752984261 Chr.22: 38069110-38069110 on Build GRCh38
rs374902648 Chr.22: 38069009-38069009 on Build GRCh38
rs563658210 Chr.22: 38069020-38069020 on Build GRCh38
rs751138380 Chr.22: 38069086-38069086 on Build GRCh38
rs1042632188 Chr.22: 38069212-38069212 on Build GRCh38
rs991730575 Chr.22: 38069183-38069183 on Build GRCh38
rs961146463 Chr.22: 38069431-38069431 on Build GRCh38
rs374575390 Chr.22: 38069020-38069020 on Build GRCh38
rs767917749 Chr.22: 38069021-38069022 on Build GRCh38
rs1042472718 Chr.22: 38069276-38069276 on Build GRCh38
rs756623836 Chr.22: 38069380-38069380 on Build GRCh38
rs1015265905 Chr.22: 38069428-38069428 on Build GRCh38
rs202090294 Chr.22: 38069026-38069026 on Build GRCh38
rs529753389 Chr.22: 38069104-38069104 on Build GRCh38
rs1002763539 Chr.22: 38069254-38069256 on Build GRCh38
rs766956739 Chr.22: 38069089-38069089 on Build GRCh38
rs775116537 Chr.22: 38069073-38069073 on Build GRCh38
rs147840806 Chr.22: 38069082-38069082 on Build GRCh38
rs771152445 Chr.22: 38069142-38069142 on Build GRCh38
rs570689009 Chr.22: 38069228-38069228 on Build GRCh38
rs1050330683 Chr.22: 38069246-38069246 on Build GRCh38
rs779380103 Chr.22: 38069105-38069105 on Build GRCh38
rs751057062 Chr.22: 38069100-38069100 on Build GRCh38
rs12485234 Chr.22: 38069406-38069406 on Build GRCh38
rs373730309 Chr.22: 38069157-38069157 on Build GRCh38
rs1053994338 Chr.22: 38069429-38069429 on Build GRCh38
rs551737263 Chr.22: 38069221-38069221 on Build GRCh38
rs759339017 Chr.22: 38069067-38069067 on Build GRCh38
rs546541833 Chr.22: 38069025-38069025 on Build GRCh38
rs750906297 Chr.22: 38069116-38069116 on Build GRCh38
rs754392890 Chr.22: 38069091-38069091 on Build GRCh38
rs3026690 Chr.22: 38069367-38069367 on Build GRCh38
rs1003510942 Chr.22: 38069299-38069299 on Build GRCh38
rs761319828 Chr.22: 38069088-38069088 on Build GRCh38
rs550145275 Chr.22: 38069061-38069061 on Build GRCh38
rs771064347 Chr.22: 38069063-38069063 on Build GRCh38
rs936910309 Chr.22: 38069419-38069419 on Build GRCh38
rs908320409 Chr.22: 38069222-38069222 on Build GRCh38
rs959991015 Chr.22: 38069180-38069180 on Build GRCh38
rs745856405 Chr.22: 38069152-38069152 on Build GRCh38
rs377307272 Chr.22: 38069007-38069007 on Build GRCh38
rs769660774 Chr.22: 38069068-38069068 on Build GRCh38
rs1035566080 Chr.22: 38069170-38069170 on Build GRCh38
rs758755158 Chr.22: 38069121-38069121 on Build GRCh38
rs778058061 Chr.22: 38069430-38069430 on Build GRCh38
rs939651537 Chr.22: 38069223-38069223 on Build GRCh38
rs746381585 Chr.22: 38069245-38069245 on Build GRCh38
rs143350563 Chr.22: 38069389-38069389 on Build GRCh38
rs1023317325 Chr.22: 38069374-38069374 on Build GRCh38
rs763721808 Chr.22: 38069083-38069083 on Build GRCh38
rs922850646 Chr.22: 38069231-38069231 on Build GRCh38
rs1022821963 Chr.22: 38069192-38069192 on Build GRCh38
rs895704522 Chr.22: 38069317-38069317 on Build GRCh38
rs773758489 Chr.22: 38069005-38069006 on Build GRCh38
rs950626273 Chr.22: 38069297-38069297 on Build GRCh38

View Full Product Details