Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 40680871-40681367 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MCHR1
Entrez Gene ID
2847
Gene Name
melanin concentrating hormone receptor 1
Gene Aliases
GPR24, MCH-1R, MCH1R, SLC-1, SLC1
UniGene
Hs.248122
Gene Chromosome Location
Chr.22: 40679178-40682814 on Build GRCh38
Transcripts
NM_005297.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4701137 c.335T>C p.M112T Chr.22: 40680994-40680994 on Build GRCh38
3554850 c.408G>A p.K136K Chr.22: 40681067-40681067 on Build GRCh38
3554851 c.540T>A p.F180L Chr.22: 40681199-40681199 on Build GRCh38
4701138 c.600C>A p.T200T Chr.22: 40681259-40681259 on Build GRCh38
1034480 c.578C>T p.A193V Chr.22: 40681237-40681237 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769413588 Chr.22: 40680958-40680958 on Build GRCh38
rs775793339 Chr.22: 40681045-40681045 on Build GRCh38
rs200659421 Chr.22: 40681161-40681161 on Build GRCh38
rs752822536 Chr.22: 40680976-40680976 on Build GRCh38
rs571399123 Chr.22: 40681012-40681012 on Build GRCh38
rs764306337 Chr.22: 40680982-40680982 on Build GRCh38
rs199847547 Chr.22: 40681007-40681007 on Build GRCh38
rs201438193 Chr.22: 40681329-40681329 on Build GRCh38
rs915365004 Chr.22: 40681237-40681237 on Build GRCh38
rs201808043 Chr.22: 40681321-40681321 on Build GRCh38
rs1007338393 Chr.22: 40681128-40681128 on Build GRCh38
rs369664918 Chr.22: 40681053-40681053 on Build GRCh38
rs777541906 Chr.22: 40681024-40681024 on Build GRCh38
rs201495946 Chr.22: 40680927-40680927 on Build GRCh38
rs201299757 Chr.22: 40681252-40681252 on Build GRCh38
rs770829840 Chr.22: 40681031-40681031 on Build GRCh38
rs375716449 Chr.22: 40681187-40681187 on Build GRCh38
rs202164457 Chr.22: 40681047-40681047 on Build GRCh38
rs754405878 Chr.22: 40681008-40681008 on Build GRCh38
rs979600517 Chr.22: 40680962-40680962 on Build GRCh38
rs752123648 Chr.22: 40681124-40681124 on Build GRCh38
rs201202695 Chr.22: 40680965-40680965 on Build GRCh38
rs746908607 Chr.22: 40681308-40681308 on Build GRCh38
rs374850218 Chr.22: 40680973-40680973 on Build GRCh38
rs745882964 Chr.22: 40681178-40681178 on Build GRCh38
rs768333904 Chr.22: 40680931-40680931 on Build GRCh38
rs80015181 Chr.22: 40681269-40681269 on Build GRCh38
rs45623433 Chr.22: 40680978-40680978 on Build GRCh38
rs773420850 Chr.22: 40681243-40681243 on Build GRCh38
rs765877994 Chr.22: 40681057-40681057 on Build GRCh38
rs200416119 Chr.22: 40681069-40681069 on Build GRCh38
rs748371706 Chr.22: 40681232-40681232 on Build GRCh38
rs987667577 Chr.22: 40681148-40681148 on Build GRCh38
rs758174382 Chr.22: 40681297-40681297 on Build GRCh38
rs180949089 Chr.22: 40681309-40681309 on Build GRCh38
rs199740916 Chr.22: 40681278-40681278 on Build GRCh38
rs763196557 Chr.22: 40681088-40681090 on Build GRCh38
rs200119224 Chr.22: 40680935-40680935 on Build GRCh38
rs896993823 Chr.22: 40681077-40681077 on Build GRCh38
rs749745254 Chr.22: 40680922-40680922 on Build GRCh38
rs752035154 Chr.22: 40680928-40680928 on Build GRCh38
rs200627010 Chr.22: 40680960-40680960 on Build GRCh38
rs200956049 Chr.22: 40681034-40681034 on Build GRCh38
rs200579028 Chr.22: 40680926-40680926 on Build GRCh38
rs778664994 Chr.22: 40681176-40681176 on Build GRCh38
rs201285391 Chr.22: 40680977-40680977 on Build GRCh38
rs200876813 Chr.22: 40681218-40681218 on Build GRCh38
rs184206916 Chr.22: 40681020-40681020 on Build GRCh38
rs761007989 Chr.22: 40681099-40681099 on Build GRCh38
rs200222615 Chr.22: 40681271-40681271 on Build GRCh38
rs748006521 Chr.22: 40680941-40680941 on Build GRCh38
rs143942609 Chr.22: 40681062-40681062 on Build GRCh38
rs778826311 Chr.22: 40681081-40681081 on Build GRCh38
rs202107852 Chr.22: 40681035-40681035 on Build GRCh38
rs201390387 Chr.22: 40681296-40681296 on Build GRCh38
rs200435236 Chr.22: 40681039-40681039 on Build GRCh38
rs200897081 Chr.22: 40681100-40681100 on Build GRCh38
rs202175341 Chr.22: 40681253-40681253 on Build GRCh38
rs764584039 Chr.22: 40681051-40681051 on Build GRCh38
rs766813535 Chr.22: 40681258-40681258 on Build GRCh38
rs376015795 Chr.22: 40681010-40681010 on Build GRCh38
rs201895311 Chr.22: 40681117-40681117 on Build GRCh38
rs574065876 Chr.22: 40681210-40681210 on Build GRCh38
rs369827677 Chr.22: 40681123-40681123 on Build GRCh38
rs199923574 Chr.22: 40681151-40681151 on Build GRCh38
rs886316956 Chr.22: 40681336-40681336 on Build GRCh38
rs751739288 Chr.22: 40681269-40681269 on Build GRCh38
rs773431836 Chr.22: 40681330-40681330 on Build GRCh38
rs201703342 Chr.22: 40681056-40681056 on Build GRCh38
rs201942191 Chr.22: 40680961-40680961 on Build GRCh38
rs765582325 Chr.22: 40680916-40680919 on Build GRCh38
rs770670064 Chr.22: 40681080-40681080 on Build GRCh38
rs200370529 Chr.22: 40681322-40681322 on Build GRCh38
rs750570821 Chr.22: 40681337-40681337 on Build GRCh38
rs148618462 Chr.22: 40681094-40681094 on Build GRCh38
rs201977776 Chr.22: 40681231-40681231 on Build GRCh38
rs149488799 Chr.22: 40681025-40681025 on Build GRCh38
rs762273865 Chr.22: 40681105-40681105 on Build GRCh38
rs770353095 Chr.22: 40681233-40681233 on Build GRCh38
rs201899738 Chr.22: 40680913-40680913 on Build GRCh38
rs757056931 Chr.22: 40681173-40681173 on Build GRCh38
rs963596917 Chr.22: 40680944-40680944 on Build GRCh38
rs777590947 Chr.22: 40681072-40681072 on Build GRCh38
rs948134494 Chr.22: 40681293-40681293 on Build GRCh38
rs760177693 Chr.22: 40681265-40681265 on Build GRCh38
rs763368168 Chr.22: 40681119-40681119 on Build GRCh38
rs766515276 Chr.22: 40680964-40680964 on Build GRCh38
rs776954401 Chr.22: 40681101-40681101 on Build GRCh38
rs201582299 Chr.22: 40681046-40681046 on Build GRCh38
rs200107865 Chr.22: 40681190-40681190 on Build GRCh38
rs764292808 Chr.22: 40681189-40681209 on Build GRCh38
rs755694362 Chr.22: 40681071-40681071 on Build GRCh38
rs754453025 Chr.22: 40680997-40680997 on Build GRCh38
rs368842613 Chr.22: 40681216-40681216 on Build GRCh38
rs781421715 Chr.22: 40681310-40681310 on Build GRCh38
rs750210146 Chr.22: 40681288-40681288 on Build GRCh38
rs771256039 Chr.22: 40681344-40681344 on Build GRCh38
rs200659135 Chr.22: 40681146-40681146 on Build GRCh38
rs11914085 Chr.22: 40680966-40680966 on Build GRCh38
rs775425755 Chr.22: 40681095-40681095 on Build GRCh38
rs979674247 Chr.22: 40681226-40681226 on Build GRCh38
rs145237086 Chr.22: 40681339-40681339 on Build GRCh38
rs917018246 Chr.22: 40681340-40681340 on Build GRCh38
rs776100044 Chr.22: 40680938-40680938 on Build GRCh38
rs757274861 Chr.22: 40681222-40681222 on Build GRCh38
rs779995029 Chr.22: 40681198-40681198 on Build GRCh38
rs765055919 Chr.22: 40681287-40681287 on Build GRCh38
rs188147970 Chr.22: 40681279-40681279 on Build GRCh38
rs199738395 Chr.22: 40681087-40681087 on Build GRCh38
rs199947845 Chr.22: 40681240-40681240 on Build GRCh38
rs775580961 Chr.22: 40680986-40680988 on Build GRCh38
rs774878903 Chr.22: 40681264-40681264 on Build GRCh38
rs771978984 Chr.22: 40681040-40681040 on Build GRCh38
rs146647078 Chr.22: 40681172-40681172 on Build GRCh38
rs375960571 Chr.22: 40681061-40681061 on Build GRCh38
rs200416969 Chr.22: 40681055-40681055 on Build GRCh38
rs779860748 Chr.22: 40681299-40681299 on Build GRCh38
rs372373026 Chr.22: 40681058-40681058 on Build GRCh38
rs201095452 Chr.22: 40681076-40681076 on Build GRCh38
rs199595704 Chr.22: 40680920-40680920 on Build GRCh38
rs140185504 Chr.22: 40681225-40681225 on Build GRCh38
rs560068779 Chr.22: 40681166-40681166 on Build GRCh38
rs763567756 Chr.22: 40681250-40681250 on Build GRCh38
rs139637639 Chr.22: 40681001-40681001 on Build GRCh38

View Full Product Details