Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 45193297-45193805 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA0930
Entrez Gene ID
23313
Gene Name
KIAA0930
Gene Aliases
C22orf9, LSC3
UniGene
Hs.592207
Gene Chromosome Location
Chr.22: 45192242-45240769 on Build GRCh38
Transcripts
NM_001009880.1 NM_015264.1 XM_011530017.2 XM_017028672.1

SNP Detail

SNP ID Chromosome Location
rs986900634 Chr.22: 45193428-45193431 on Build GRCh38
rs942893920 Chr.22: 45193464-45193464 on Build GRCh38
rs372412554 Chr.22: 45193484-45193484 on Build GRCh38
rs1039927555 Chr.22: 45193482-45193482 on Build GRCh38
rs944954167 Chr.22: 45193471-45193471 on Build GRCh38
rs757468159 Chr.22: 45193698-45193702 on Build GRCh38
rs748571113 Chr.22: 45193414-45193414 on Build GRCh38
rs961016287 Chr.22: 45193608-45193608 on Build GRCh38
rs886165403 Chr.22: 45193516-45193516 on Build GRCh38
rs563867374 Chr.22: 45193628-45193628 on Build GRCh38
rs939704806 Chr.22: 45193368-45193368 on Build GRCh38
rs964170853 Chr.22: 45193599-45193599 on Build GRCh38
rs1041014273 Chr.22: 45193732-45193732 on Build GRCh38
rs533127724 Chr.22: 45193765-45193766 on Build GRCh38
rs773809790 Chr.22: 45193451-45193451 on Build GRCh38
rs1023224013 Chr.22: 45193532-45193532 on Build GRCh38
rs910707668 Chr.22: 45193640-45193640 on Build GRCh38
rs957312331 Chr.22: 45193541-45193541 on Build GRCh38
rs893236256 Chr.22: 45193526-45193526 on Build GRCh38
rs930773525 Chr.22: 45193403-45193403 on Build GRCh38
rs529577629 Chr.22: 45193629-45193629 on Build GRCh38
rs190412632 Chr.22: 45193523-45193523 on Build GRCh38
rs1125398 Chr.22: 45193609-45193609 on Build GRCh38
rs1034261074 Chr.22: 45193537-45193537 on Build GRCh38
rs767250866 Chr.22: 45193644-45193644 on Build GRCh38
rs564164312 Chr.22: 45193573-45193573 on Build GRCh38
rs772436419 Chr.22: 45193424-45193424 on Build GRCh38
rs1012718832 Chr.22: 45193531-45193531 on Build GRCh38
rs774694574 Chr.22: 45193362-45193362 on Build GRCh38
rs906986401 Chr.22: 45193744-45193744 on Build GRCh38
rs182986294 Chr.22: 45193677-45193677 on Build GRCh38
rs967810323 Chr.22: 45193586-45193586 on Build GRCh38
rs539239005 Chr.22: 45193324-45193324 on Build GRCh38
rs967400782 Chr.22: 45193777-45193777 on Build GRCh38
rs893012855 Chr.22: 45193758-45193758 on Build GRCh38
rs531329565 Chr.22: 45193632-45193632 on Build GRCh38
rs563340918 Chr.22: 45193427-45193428 on Build GRCh38
rs762625257 Chr.22: 45193590-45193590 on Build GRCh38
rs946169933 Chr.22: 45193428-45193428 on Build GRCh38
rs903182981 Chr.22: 45193502-45193502 on Build GRCh38
rs147962181 Chr.22: 45193447-45193447 on Build GRCh38
rs889337957 Chr.22: 45193435-45193435 on Build GRCh38
rs187175405 Chr.22: 45193325-45193325 on Build GRCh38
rs541153395 Chr.22: 45193544-45193544 on Build GRCh38
rs1054394565 Chr.22: 45193769-45193769 on Build GRCh38
rs368999996 Chr.22: 45193372-45193372 on Build GRCh38
rs986827389 Chr.22: 45193587-45193587 on Build GRCh38
rs761116745 Chr.22: 45193456-45193456 on Build GRCh38
rs907524003 Chr.22: 45193663-45193663 on Build GRCh38
rs780921005 Chr.22: 45193765-45193765 on Build GRCh38
rs544036054 Chr.22: 45193623-45193623 on Build GRCh38
rs775151828 Chr.22: 45193534-45193534 on Build GRCh38
rs940362376 Chr.22: 45193689-45193689 on Build GRCh38
rs1015550325 Chr.22: 45193601-45193601 on Build GRCh38
rs1020295999 Chr.22: 45193581-45193581 on Build GRCh38
rs1048726094 Chr.22: 45193508-45193508 on Build GRCh38
rs1034292385 Chr.22: 45193406-45193406 on Build GRCh38
rs919695877 Chr.22: 45193384-45193384 on Build GRCh38
rs930273670 Chr.22: 45193488-45193488 on Build GRCh38
rs3810630 Chr.22: 45193521-45193521 on Build GRCh38
rs576258961 Chr.22: 45193457-45193457 on Build GRCh38
rs535699074 Chr.22: 45193493-45193493 on Build GRCh38
rs559517219 Chr.22: 45193755-45193755 on Build GRCh38
rs1040668472 Chr.22: 45193492-45193492 on Build GRCh38

View Full Product Details