Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 45202795-45203287 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA0930
Entrez Gene ID
23313
Gene Name
KIAA0930
Gene Aliases
C22orf9, LSC3
UniGene
Hs.592207
Gene Chromosome Location
Chr.22: 45192242-45240769 on Build GRCh38
Transcripts
NM_001009880.1 NM_015264.1 XM_011530017.2 XM_017028672.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6526651 c.779G>T p.R260L Chr.22: 45203063-45203063 on Build GRCh38
4695444 c.783G>A p.V261V Chr.22: 45203059-45203059 on Build GRCh38
6889238 c.787G>A p.V263I Chr.22: 45203070-45203070 on Build GRCh38
6005035 c.728G>A p.S243N Chr.22: 45203129-45203129 on Build GRCh38
6693511 c.794G>A p.R265Q Chr.22: 45203063-45203063 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1050967148 Chr.22: 45202828-45202828 on Build GRCh38
rs764883102 Chr.22: 45202967-45202968 on Build GRCh38
rs113662359 Chr.22: 45203173-45203173 on Build GRCh38
rs764724685 Chr.22: 45203006-45203006 on Build GRCh38
rs969548483 Chr.22: 45202901-45202901 on Build GRCh38
rs761743686 Chr.22: 45203207-45203207 on Build GRCh38
rs780499999 Chr.22: 45203040-45203040 on Build GRCh38
rs141939946 Chr.22: 45202992-45202992 on Build GRCh38
rs572724126 Chr.22: 45202924-45202924 on Build GRCh38
rs535576293 Chr.22: 45202872-45202872 on Build GRCh38
rs781727490 Chr.22: 45203178-45203178 on Build GRCh38
rs751314964 Chr.22: 45202916-45202916 on Build GRCh38
rs371999706 Chr.22: 45203194-45203194 on Build GRCh38
rs764746475 Chr.22: 45203156-45203156 on Build GRCh38
rs201951656 Chr.22: 45203083-45203083 on Build GRCh38
rs944517637 Chr.22: 45202965-45202965 on Build GRCh38
rs1041215340 Chr.22: 45203123-45203123 on Build GRCh38
rs762531719 Chr.22: 45203217-45203217 on Build GRCh38
rs866467744 Chr.22: 45202871-45202871 on Build GRCh38
rs747460442 Chr.22: 45202969-45202969 on Build GRCh38
rs756151601 Chr.22: 45203063-45203063 on Build GRCh38
rs765119685 Chr.22: 45203213-45203213 on Build GRCh38
rs372594026 Chr.22: 45202953-45202953 on Build GRCh38
rs139406514 Chr.22: 45203071-45203071 on Build GRCh38
rs972317689 Chr.22: 45202933-45202933 on Build GRCh38
rs920577304 Chr.22: 45202934-45202934 on Build GRCh38
rs1035482776 Chr.22: 45203200-45203200 on Build GRCh38
rs373830406 Chr.22: 45202967-45202967 on Build GRCh38
rs952160641 Chr.22: 45202949-45202949 on Build GRCh38
rs373057962 Chr.22: 45203225-45203225 on Build GRCh38
rs751242300 Chr.22: 45203220-45203220 on Build GRCh38
rs373895809 Chr.22: 45203152-45203152 on Build GRCh38
rs776307341 Chr.22: 45203206-45203206 on Build GRCh38
rs373645063 Chr.22: 45203082-45203082 on Build GRCh38
rs559426834 Chr.22: 45203227-45203227 on Build GRCh38
rs749906566 Chr.22: 45203167-45203167 on Build GRCh38
rs1139801 Chr.22: 45203005-45203005 on Build GRCh38
rs732373 Chr.22: 45202952-45202952 on Build GRCh38
rs761929586 Chr.22: 45202985-45202985 on Build GRCh38
rs570712471 Chr.22: 45203041-45203041 on Build GRCh38
rs1042233847 Chr.22: 45202858-45202858 on Build GRCh38
rs1012797658 Chr.22: 45203240-45203240 on Build GRCh38
rs1035136703 Chr.22: 45203126-45203126 on Build GRCh38
rs924423263 Chr.22: 45203229-45203229 on Build GRCh38
rs543737972 Chr.22: 45203143-45203143 on Build GRCh38
rs150986581 Chr.22: 45203149-45203149 on Build GRCh38
rs752278022 Chr.22: 45203148-45203150 on Build GRCh38
rs201472032 Chr.22: 45203029-45203029 on Build GRCh38
rs372186039 Chr.22: 45202996-45202996 on Build GRCh38
rs962260150 Chr.22: 45202928-45202928 on Build GRCh38
rs768430897 Chr.22: 45203203-45203203 on Build GRCh38
rs903105686 Chr.22: 45202842-45202842 on Build GRCh38
rs778608634 Chr.22: 45202958-45202958 on Build GRCh38
rs750474005 Chr.22: 45203036-45203036 on Build GRCh38
rs1023147233 Chr.22: 45202898-45202898 on Build GRCh38
rs962209201 Chr.22: 45203127-45203127 on Build GRCh38
rs755160878 Chr.22: 45203179-45203179 on Build GRCh38
rs774528653 Chr.22: 45203198-45203198 on Build GRCh38
rs773175119 Chr.22: 45203112-45203112 on Build GRCh38
rs754718085 Chr.22: 45202961-45202962 on Build GRCh38
rs774160004 Chr.22: 45202983-45202983 on Build GRCh38
rs369518786 Chr.22: 45203111-45203111 on Build GRCh38
rs753824996 Chr.22: 45203003-45203003 on Build GRCh38
rs763887785 Chr.22: 45202868-45202868 on Build GRCh38
rs1035188929 Chr.22: 45202927-45202927 on Build GRCh38
rs752490353 Chr.22: 45203002-45203002 on Build GRCh38
rs750081075 Chr.22: 45203018-45203018 on Build GRCh38
rs201416495 Chr.22: 45203197-45203197 on Build GRCh38
rs367567519 Chr.22: 45203199-45203199 on Build GRCh38
rs772484012 Chr.22: 45202966-45202966 on Build GRCh38
rs549767609 Chr.22: 45202850-45202850 on Build GRCh38
rs749457155 Chr.22: 45203128-45203128 on Build GRCh38
rs934491501 Chr.22: 45203242-45203242 on Build GRCh38
rs910594027 Chr.22: 45203209-45203209 on Build GRCh38
rs991488974 Chr.22: 45202848-45202848 on Build GRCh38
rs914588259 Chr.22: 45202849-45202849 on Build GRCh38
rs142644672 Chr.22: 45203080-45203080 on Build GRCh38
rs539318052 Chr.22: 45202836-45202836 on Build GRCh38
rs2840210 Chr.22: 45203218-45203218 on Build GRCh38
rs140037195 Chr.22: 45202991-45202991 on Build GRCh38
rs745551149 Chr.22: 45202961-45202961 on Build GRCh38
rs376843102 Chr.22: 45203196-45203196 on Build GRCh38
rs377371546 Chr.22: 45203154-45203154 on Build GRCh38
rs866692355 Chr.22: 45203072-45203072 on Build GRCh38
rs779333364 Chr.22: 45203224-45203224 on Build GRCh38
rs756592226 Chr.22: 45203235-45203235 on Build GRCh38
rs1040259549 Chr.22: 45202841-45202841 on Build GRCh38
rs944196960 Chr.22: 45202866-45202866 on Build GRCh38
rs370431927 Chr.22: 45203169-45203169 on Build GRCh38
rs944552237 Chr.22: 45202837-45202837 on Build GRCh38
rs997531250 Chr.22: 45202905-45202905 on Build GRCh38
rs751295257 Chr.22: 45203172-45203172 on Build GRCh38
rs763593646 Chr.22: 45202929-45202930 on Build GRCh38
rs570735818 Chr.22: 45202826-45202826 on Build GRCh38
rs368028093 Chr.22: 45202983-45202983 on Build GRCh38
rs555492251 Chr.22: 45202876-45202876 on Build GRCh38
rs554729459 Chr.22: 45202853-45202853 on Build GRCh38
rs373358378 Chr.22: 45202829-45202829 on Build GRCh38
rs772731065 Chr.22: 45203216-45203216 on Build GRCh38
rs756053877 Chr.22: 45202943-45202943 on Build GRCh38
rs766154885 Chr.22: 45203170-45203170 on Build GRCh38
rs569645086 Chr.22: 45202865-45202865 on Build GRCh38
rs771171042 Chr.22: 45203139-45203139 on Build GRCh38
rs958676893 Chr.22: 45202833-45202833 on Build GRCh38
rs1050081261 Chr.22: 45202911-45202911 on Build GRCh38
rs1013003128 Chr.22: 45203042-45203042 on Build GRCh38
rs769293437 Chr.22: 45202972-45202972 on Build GRCh38

View Full Product Details