Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 11137195-11137692 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HRH1
Entrez Gene ID
3269
Gene Name
histamine receptor H1
Gene Aliases
H1-R, H1R, HH1R, hisH1
UniGene
Hs.1570
Gene Chromosome Location
Chr.3: 11137093-11263253 on Build GRCh38
Transcripts
NM_001098213.1

SNP Detail

SNP ID Chromosome Location
rs967181574 Chr.3: 11137584-11137584 on Build GRCh38
rs905819791 Chr.3: 11137467-11137467 on Build GRCh38
rs955767929 Chr.3: 11137616-11137616 on Build GRCh38
rs752355947 Chr.3: 11137461-11137461 on Build GRCh38
rs775171547 Chr.3: 11137355-11137355 on Build GRCh38
rs761218800 Chr.3: 11137630-11137630 on Build GRCh38
rs978917987 Chr.3: 11137335-11137335 on Build GRCh38
rs917521312 Chr.3: 11137640-11137640 on Build GRCh38
rs966212521 Chr.3: 11137560-11137560 on Build GRCh38
rs920897674 Chr.3: 11137291-11137291 on Build GRCh38
rs543418406 Chr.3: 11137252-11137252 on Build GRCh38
rs975039258 Chr.3: 11137260-11137260 on Build GRCh38
rs925628417 Chr.3: 11137345-11137345 on Build GRCh38
rs73128473 Chr.3: 11137415-11137415 on Build GRCh38
rs866210201 Chr.3: 11137357-11137357 on Build GRCh38
rs991750404 Chr.3: 11137352-11137352 on Build GRCh38
rs767067085 Chr.3: 11137663-11137663 on Build GRCh38
rs1019097765 Chr.3: 11137526-11137526 on Build GRCh38
rs994887578 Chr.3: 11137469-11137469 on Build GRCh38
rs890371005 Chr.3: 11137499-11137499 on Build GRCh38
rs1044328266 Chr.3: 11137453-11137453 on Build GRCh38
rs917099691 Chr.3: 11137429-11137429 on Build GRCh38
rs564360286 Chr.3: 11137218-11137218 on Build GRCh38
rs978481071 Chr.3: 11137612-11137612 on Build GRCh38
rs1005087626 Chr.3: 11137232-11137232 on Build GRCh38
rs996795320 Chr.3: 11137588-11137588 on Build GRCh38
rs953837722 Chr.3: 11137610-11137610 on Build GRCh38
rs983378180 Chr.3: 11137657-11137657 on Build GRCh38
rs917470926 Chr.3: 11137372-11137372 on Build GRCh38
rs1020099541 Chr.3: 11137541-11137541 on Build GRCh38
rs958365786 Chr.3: 11137622-11137622 on Build GRCh38
rs966305466 Chr.3: 11137251-11137251 on Build GRCh38
rs1019193448 Chr.3: 11137249-11137249 on Build GRCh38
rs751586800 Chr.3: 11137605-11137605 on Build GRCh38
rs144533410 Chr.3: 11137644-11137644 on Build GRCh38
rs561497255 Chr.3: 11137347-11137347 on Build GRCh38
rs527876300 Chr.3: 11137427-11137427 on Build GRCh38
rs879661668 Chr.3: 11137435-11137435 on Build GRCh38
rs34080141 Chr.3: 11137646-11137646 on Build GRCh38
rs967141769 Chr.3: 11137320-11137320 on Build GRCh38
rs899856542 Chr.3: 11137464-11137464 on Build GRCh38
rs953743723 Chr.3: 11137314-11137314 on Build GRCh38
rs925688980 Chr.3: 11137614-11137614 on Build GRCh38
rs570795142 Chr.3: 11137482-11137482 on Build GRCh38
rs528897195 Chr.3: 11137367-11137367 on Build GRCh38
rs978449016 Chr.3: 11137334-11137334 on Build GRCh38
rs1009175416 Chr.3: 11137510-11137510 on Build GRCh38
rs1032729309 Chr.3: 11137618-11137618 on Build GRCh38
rs762892190 Chr.3: 11137480-11137480 on Build GRCh38
rs934484311 Chr.3: 11137408-11137408 on Build GRCh38
rs565770035 Chr.3: 11137419-11137419 on Build GRCh38
rs552776867 Chr.3: 11137489-11137489 on Build GRCh38
rs1038392183 Chr.3: 11137455-11137455 on Build GRCh38
rs79738984 Chr.3: 11137431-11137431 on Build GRCh38
rs886551038 Chr.3: 11137485-11137485 on Build GRCh38
rs954709191 Chr.3: 11137215-11137215 on Build GRCh38
rs184420692 Chr.3: 11137220-11137220 on Build GRCh38

View Full Product Details