Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 15076899-15077397 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RBSN
Entrez Gene ID
64145
Gene Name
rabenosyn, RAB effector
Gene Aliases
Rabenosyn-5, ZFYVE20
UniGene
Hs.475565
Gene Chromosome Location
Chr.3: 15070069-15099163 on Build GRCh38
Transcripts
NM_001302378.1 NM_022340.3 XM_005265384.4 XM_005265385.4 XM_011534001.2 XM_017007023.1 XM_017007024.1 XM_017007025.1 XM_017007026.1 XM_017007027.1 XM_017007028.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6316336 c.1101+8C>A p.? Chr.3: 15077054-15077054 on Build GRCh38
728689 c.1026C>A p.N342K Chr.3: 15077137-15077137 on Build GRCh38
5398417 c.1008C>T p.I336I Chr.3: 15077155-15077155 on Build GRCh38
3003758 c.1049G>A p.S350N Chr.3: 15077114-15077114 on Build GRCh38
3767103 c.1095T>C p.F365F Chr.3: 15077068-15077068 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs967694222 Chr.3: 15077286-15077286 on Build GRCh38
rs374498576 Chr.3: 15077303-15077303 on Build GRCh38
rs754844313 Chr.3: 15077020-15077020 on Build GRCh38
rs746883470 Chr.3: 15077014-15077014 on Build GRCh38
rs1047626544 Chr.3: 15077200-15077200 on Build GRCh38
rs757928090 Chr.3: 15077012-15077012 on Build GRCh38
rs764571121 Chr.3: 15076971-15076972 on Build GRCh38
rs111570844 Chr.3: 15077256-15077256 on Build GRCh38
rs571978529 Chr.3: 15077181-15077181 on Build GRCh38
rs987435704 Chr.3: 15077339-15077339 on Build GRCh38
rs752440317 Chr.3: 15077204-15077204 on Build GRCh38
rs916059984 Chr.3: 15076995-15076995 on Build GRCh38
rs769999609 Chr.3: 15077048-15077048 on Build GRCh38
rs375981231 Chr.3: 15077211-15077211 on Build GRCh38
rs773356650 Chr.3: 15077050-15077050 on Build GRCh38
rs750051860 Chr.3: 15077115-15077115 on Build GRCh38
rs150393317 Chr.3: 15077103-15077103 on Build GRCh38
rs200729473 Chr.3: 15077131-15077131 on Build GRCh38
rs747256391 Chr.3: 15077236-15077236 on Build GRCh38
rs529516599 Chr.3: 15077360-15077360 on Build GRCh38
rs935741977 Chr.3: 15076951-15076951 on Build GRCh38
rs1488096 Chr.3: 15077098-15077098 on Build GRCh38
rs746665 Chr.3: 15077226-15077226 on Build GRCh38
rs749393093 Chr.3: 15077053-15077053 on Build GRCh38
rs774878160 Chr.3: 15077064-15077064 on Build GRCh38
rs774795883 Chr.3: 15077022-15077022 on Build GRCh38
rs75933903 Chr.3: 15077309-15077309 on Build GRCh38
rs925762661 Chr.3: 15076938-15076938 on Build GRCh38
rs956018565 Chr.3: 15076999-15076999 on Build GRCh38
rs780829542 Chr.3: 15077040-15077040 on Build GRCh38
rs770958290 Chr.3: 15077063-15077063 on Build GRCh38
rs535136720 Chr.3: 15076964-15076964 on Build GRCh38
rs778707697 Chr.3: 15077114-15077115 on Build GRCh38
rs371816149 Chr.3: 15077190-15077190 on Build GRCh38
rs779838393 Chr.3: 15077106-15077106 on Build GRCh38
rs956278479 Chr.3: 15077329-15077329 on Build GRCh38
rs578206233 Chr.3: 15077284-15077284 on Build GRCh38
rs993551614 Chr.3: 15077144-15077144 on Build GRCh38
rs1031926036 Chr.3: 15077321-15077321 on Build GRCh38
rs201418042 Chr.3: 15077171-15077171 on Build GRCh38
rs770391152 Chr.3: 15077366-15077366 on Build GRCh38
rs758016107 Chr.3: 15077132-15077132 on Build GRCh38
rs746214908 Chr.3: 15077359-15077359 on Build GRCh38
rs781109932 Chr.3: 15077201-15077201 on Build GRCh38
rs748462260 Chr.3: 15077041-15077041 on Build GRCh38
rs977145808 Chr.3: 15077295-15077295 on Build GRCh38
rs1055918073 Chr.3: 15076957-15076957 on Build GRCh38
rs780166760 Chr.3: 15077013-15077013 on Build GRCh38
rs759935891 Chr.3: 15077070-15077070 on Build GRCh38
rs564783830 Chr.3: 15077264-15077264 on Build GRCh38
rs374570763 Chr.3: 15077073-15077073 on Build GRCh38
rs748304896 Chr.3: 15077159-15077159 on Build GRCh38
rs974576325 Chr.3: 15077125-15077125 on Build GRCh38
rs979404238 Chr.3: 15076927-15076927 on Build GRCh38
rs756142668 Chr.3: 15077105-15077105 on Build GRCh38
rs765959798 Chr.3: 15077162-15077162 on Build GRCh38

View Full Product Details