Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 97078703-97079203 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPYD
Entrez Gene ID
1806
Gene Name
dihydropyrimidine dehydrogenase
Gene Aliases
DHP, DHPDHASE, DPD
UniGene
Hs.335034
Gene Chromosome Location
Chr.1: 97077743-97921059 on Build GRCh38
Transcripts
NM_000110.3 XM_005270562.3 XM_017000507.1 XM_017000508.1 XM_017000509.1 XM_017000510.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3806188 c.2997C>T p.V999V Chr.1: 97079057-97079057 on Build GRCh38
3377320 c.3051A>G p.V1017V Chr.1: 97079003-97079003 on Build GRCh38
1688092 c.3012G>A p.M1004I Chr.1: 97079042-97079042 on Build GRCh38
5471142 c.3068C>T p.P1023L Chr.1: 97078986-97078986 on Build GRCh38
1345089 c.2909C>A p.A970D Chr.1: 97079145-97079145 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs886046573 Chr.1: 97078734-97078734 on Build GRCh38
rs918000563 Chr.1: 97078791-97078791 on Build GRCh38
rs72547602 Chr.1: 97079133-97079133 on Build GRCh38
rs768993607 Chr.1: 97079075-97079075 on Build GRCh38
rs757376267 Chr.1: 97079134-97079134 on Build GRCh38
rs201268750 Chr.1: 97079120-97079120 on Build GRCh38
rs672601272 Chr.1: 97078803-97078803 on Build GRCh38
rs376502495 Chr.1: 97079123-97079123 on Build GRCh38
rs770320798 Chr.1: 97078992-97078992 on Build GRCh38
rs763742591 Chr.1: 97078944-97078944 on Build GRCh38
rs757509506 Chr.1: 97079057-97079057 on Build GRCh38
rs140989814 Chr.1: 97079073-97079073 on Build GRCh38
rs879583305 Chr.1: 97079094-97079094 on Build GRCh38
rs140114515 Chr.1: 97079005-97079005 on Build GRCh38
rs184161277 Chr.1: 97078871-97078871 on Build GRCh38
rs766878892 Chr.1: 97079021-97079021 on Build GRCh38
rs371478594 Chr.1: 97079180-97079180 on Build GRCh38
rs545959800 Chr.1: 97079006-97079006 on Build GRCh38
rs753820482 Chr.1: 97079110-97079110 on Build GRCh38
rs913128155 Chr.1: 97078728-97078728 on Build GRCh38
rs749122978 Chr.1: 97078985-97078985 on Build GRCh38
rs778911905 Chr.1: 97079053-97079053 on Build GRCh38
rs938508528 Chr.1: 97078775-97078775 on Build GRCh38
rs753363470 Chr.1: 97078960-97078960 on Build GRCh38
rs886046574 Chr.1: 97078783-97078783 on Build GRCh38
rs746298645 Chr.1: 97078991-97078991 on Build GRCh38
rs148799944 Chr.1: 97078993-97078993 on Build GRCh38
rs1048815334 Chr.1: 97078795-97078795 on Build GRCh38
rs926969910 Chr.1: 97078761-97078761 on Build GRCh38
rs781348608 Chr.1: 97078990-97078990 on Build GRCh38
rs61757362 Chr.1: 97079106-97079106 on Build GRCh38
rs151074666 Chr.1: 97079056-97079056 on Build GRCh38
rs747228600 Chr.1: 97078824-97078824 on Build GRCh38
rs997814794 Chr.1: 97078968-97078968 on Build GRCh38
rs756182652 Chr.1: 97079153-97079153 on Build GRCh38
rs374367724 Chr.1: 97078962-97078962 on Build GRCh38
rs759562628 Chr.1: 97079113-97079113 on Build GRCh38
rs760411849 Chr.1: 97078852-97078853 on Build GRCh38
rs150499371 Chr.1: 97078869-97078869 on Build GRCh38
rs145529148 Chr.1: 97079139-97079139 on Build GRCh38
rs760485592 Chr.1: 97079026-97079026 on Build GRCh38
rs950022179 Chr.1: 97078734-97078734 on Build GRCh38
rs773868825 Chr.1: 97079007-97079007 on Build GRCh38
rs540257862 Chr.1: 97079149-97079149 on Build GRCh38
rs1047321478 Chr.1: 97078918-97078918 on Build GRCh38
rs937052223 Chr.1: 97078770-97078770 on Build GRCh38
rs760235888 Chr.1: 97079104-97079104 on Build GRCh38
rs672601273 Chr.1: 97079067-97079067 on Build GRCh38
rs1057387215 Chr.1: 97078776-97078776 on Build GRCh38
rs758640823 Chr.1: 97078986-97078986 on Build GRCh38
rs749954216 Chr.1: 97079024-97079024 on Build GRCh38
rs139459586 Chr.1: 97079076-97079076 on Build GRCh38
rs902088385 Chr.1: 97078933-97078933 on Build GRCh38
rs370005502 Chr.1: 97078924-97078924 on Build GRCh38
rs767121228 Chr.1: 97078943-97078943 on Build GRCh38
rs981070948 Chr.1: 97078757-97078757 on Build GRCh38
rs868519561 Chr.1: 97078857-97078857 on Build GRCh38
rs750312649 Chr.1: 97078956-97078956 on Build GRCh38
rs367558192 Chr.1: 97079178-97079178 on Build GRCh38
rs55938520 Chr.1: 97078817-97078817 on Build GRCh38
rs749736621 Chr.1: 97079168-97079168 on Build GRCh38
rs202144771 Chr.1: 97079077-97079077 on Build GRCh38
rs745982505 Chr.1: 97079062-97079062 on Build GRCh38
rs1801268 Chr.1: 97079071-97079071 on Build GRCh38
rs772544099 Chr.1: 97079100-97079100 on Build GRCh38
rs72547601 Chr.1: 97079121-97079121 on Build GRCh38
rs780163365 Chr.1: 97079164-97079164 on Build GRCh38
rs761584058 Chr.1: 97079020-97079020 on Build GRCh38
rs753873495 Chr.1: 97079030-97079030 on Build GRCh38
rs779927189 Chr.1: 97078969-97078969 on Build GRCh38
rs114096998 Chr.1: 97078987-97078987 on Build GRCh38
rs1045156985 Chr.1: 97078835-97078835 on Build GRCh38
rs956842768 Chr.1: 97079002-97079002 on Build GRCh38
rs534213499 Chr.1: 97078813-97078813 on Build GRCh38
rs1036554250 Chr.1: 97078885-97078885 on Build GRCh38
rs754786483 Chr.1: 97079044-97079044 on Build GRCh38
rs766438205 Chr.1: 97079029-97079029 on Build GRCh38
rs867143119 Chr.1: 97079042-97079042 on Build GRCh38

View Full Product Details